Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 100472832-100473331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERS3
Entrez Gene ID
204219
Gene Name
ceramide synthase 3
Gene Aliases
ARCI9, LASS3
UniGene
Hs.662371
Gene Chromosome Location
Chr.15: 100400395-100544982 on Build GRCh38
Transcripts
NM_001290341.2 NM_001290342.2 NM_001290343.1 NM_178842.4 XM_011521355.2 XM_011521357.2 XM_011521358.1 XM_017022002.1 XM_017022003.1 XM_017022004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5419386 c.674C>T p.A225V Chr.15: 100472988-100472988 on Build GRCh38
959555 c.721G>A p.A241T Chr.15: 100472941-100472941 on Build GRCh38
199218 c.686G>A p.R229H Chr.15: 100472976-100472976 on Build GRCh38
5040483 c.727A>T p.I243F Chr.15: 100472935-100472935 on Build GRCh38
282369 c.736G>A p.E246K Chr.15: 100472926-100472926 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777479343 Chr.15: 100472988-100472988 on Build GRCh38
rs754294287 Chr.15: 100472961-100472961 on Build GRCh38
rs760185474 Chr.15: 100473051-100473051 on Build GRCh38
rs373080717 Chr.15: 100472962-100472962 on Build GRCh38
rs547015224 Chr.15: 100472945-100472945 on Build GRCh38
rs1030047173 Chr.15: 100473272-100473272 on Build GRCh38
rs747714645 Chr.15: 100472884-100472884 on Build GRCh38
rs377393638 Chr.15: 100473063-100473063 on Build GRCh38
rs1566775 Chr.15: 100472918-100472918 on Build GRCh38
rs770842344 Chr.15: 100472906-100472906 on Build GRCh38
rs929501220 Chr.15: 100473114-100473114 on Build GRCh38
rs766572944 Chr.15: 100472857-100472857 on Build GRCh38
rs752230253 Chr.15: 100472976-100472976 on Build GRCh38
rs115411592 Chr.15: 100472953-100472953 on Build GRCh38
rs548362259 Chr.15: 100472973-100472973 on Build GRCh38
rs745640185 Chr.15: 100473096-100473096 on Build GRCh38
rs1003224432 Chr.15: 100473235-100473235 on Build GRCh38
rs564503711 Chr.15: 100472877-100472877 on Build GRCh38
rs576674543 Chr.15: 100472860-100472860 on Build GRCh38
rs368801224 Chr.15: 100472873-100472873 on Build GRCh38
rs754711117 Chr.15: 100473251-100473251 on Build GRCh38
rs942664952 Chr.15: 100473018-100473018 on Build GRCh38
rs975321337 Chr.15: 100473297-100473297 on Build GRCh38
rs536395650 Chr.15: 100473292-100473292 on Build GRCh38
rs55948136 Chr.15: 100473209-100473209 on Build GRCh38
rs774538474 Chr.15: 100472922-100472922 on Build GRCh38
rs765720613 Chr.15: 100473071-100473071 on Build GRCh38
rs773425420 Chr.15: 100473045-100473045 on Build GRCh38
rs371641623 Chr.15: 100472946-100472946 on Build GRCh38
rs746808569 Chr.15: 100472898-100472898 on Build GRCh38
rs199508614 Chr.15: 100473085-100473085 on Build GRCh38
rs919549618 Chr.15: 100473301-100473301 on Build GRCh38
rs375146995 Chr.15: 100473088-100473088 on Build GRCh38
rs575869683 Chr.15: 100473303-100473304 on Build GRCh38
rs967230702 Chr.15: 100472950-100472950 on Build GRCh38
rs781178842 Chr.15: 100473091-100473091 on Build GRCh38
rs967199639 Chr.15: 100473254-100473255 on Build GRCh38
rs769828785 Chr.15: 100473022-100473022 on Build GRCh38
rs748253520 Chr.15: 100473026-100473026 on Build GRCh38
rs1055540286 Chr.15: 100473211-100473211 on Build GRCh38
rs1017236698 Chr.15: 100473296-100473298 on Build GRCh38
rs964221735 Chr.15: 100473296-100473296 on Build GRCh38
rs751173964 Chr.15: 100473012-100473013 on Build GRCh38
rs970348645 Chr.15: 100473226-100473226 on Build GRCh38
rs1013208365 Chr.15: 100473111-100473111 on Build GRCh38
rs539013207 Chr.15: 100473089-100473089 on Build GRCh38
rs973608064 Chr.15: 100473300-100473300 on Build GRCh38
rs954616160 Chr.15: 100473289-100473289 on Build GRCh38
rs760085605 Chr.15: 100473066-100473066 on Build GRCh38
rs775958807 Chr.15: 100472899-100472899 on Build GRCh38
rs906646964 Chr.15: 100473188-100473188 on Build GRCh38
rs752217659 Chr.15: 100473073-100473073 on Build GRCh38
rs569753993 Chr.15: 100472991-100472991 on Build GRCh38
rs776646026 Chr.15: 100472908-100472908 on Build GRCh38
rs199763147 Chr.15: 100473016-100473016 on Build GRCh38
rs115551880 Chr.15: 100473025-100473025 on Build GRCh38
rs778558573 Chr.15: 100472883-100472883 on Build GRCh38
rs1046474015 Chr.15: 100473116-100473116 on Build GRCh38
rs772185465 Chr.15: 100473029-100473029 on Build GRCh38
rs757859652 Chr.15: 100473084-100473084 on Build GRCh38
rs183730041 Chr.15: 100473262-100473262 on Build GRCh38
rs758046951 Chr.15: 100472977-100472977 on Build GRCh38
rs765983911 Chr.15: 100473092-100473092 on Build GRCh38
rs116521824 Chr.15: 100472948-100472948 on Build GRCh38
rs777478755 Chr.15: 100472891-100472891 on Build GRCh38
rs959010454 Chr.15: 100472888-100472888 on Build GRCh38
rs764611319 Chr.15: 100472975-100472975 on Build GRCh38
rs757063256 Chr.15: 100472992-100472992 on Build GRCh38
rs372549585 Chr.15: 100472887-100472887 on Build GRCh38
rs923051911 Chr.15: 100472960-100472960 on Build GRCh38
rs1030735373 Chr.15: 100473250-100473250 on Build GRCh38
rs765868912 Chr.15: 100472914-100472914 on Build GRCh38
rs555101236 Chr.15: 100473294-100473294 on Build GRCh38
rs989709902 Chr.15: 100473270-100473270 on Build GRCh38
rs766689241 Chr.15: 100473054-100473054 on Build GRCh38
rs933999118 Chr.15: 100473031-100473031 on Build GRCh38
rs776949322 Chr.15: 100473062-100473062 on Build GRCh38
rs756184984 Chr.15: 100473098-100473098 on Build GRCh38
rs140343630 Chr.15: 100472994-100472994 on Build GRCh38
rs767763454 Chr.15: 100472947-100472947 on Build GRCh38
rs999126259 Chr.15: 100473254-100473254 on Build GRCh38
rs753276178 Chr.15: 100473105-100473105 on Build GRCh38
rs923695523 Chr.15: 100472963-100472963 on Build GRCh38
rs142542948 Chr.15: 100472872-100472872 on Build GRCh38

View Full Product Details