Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 100649329-100649823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB7
Entrez Gene ID
140460
Gene Name
ankyrin repeat and SOCS box containing 7
Gene Aliases
-
UniGene
Hs.31845
Gene Chromosome Location
Chr.15: 100602550-100651701 on Build GRCh38
Transcripts
NM_198243.2

SNP Detail

SNP ID Chromosome Location
rs576971784 Chr.15: 100649752-100649752 on Build GRCh38
rs902613462 Chr.15: 100649358-100649358 on Build GRCh38
rs79850994 Chr.15: 100649790-100649790 on Build GRCh38
rs756634104 Chr.15: 100649461-100649461 on Build GRCh38
rs900206475 Chr.15: 100649523-100649523 on Build GRCh38
rs780246301 Chr.15: 100649397-100649397 on Build GRCh38
rs768944359 Chr.15: 100649666-100649666 on Build GRCh38
rs1022210081 Chr.15: 100649552-100649552 on Build GRCh38
rs770423751 Chr.15: 100649733-100649733 on Build GRCh38
rs369018735 Chr.15: 100649362-100649362 on Build GRCh38
rs1016541646 Chr.15: 100649638-100649638 on Build GRCh38
rs776520922 Chr.15: 100649738-100649738 on Build GRCh38
rs749546152 Chr.15: 100649542-100649542 on Build GRCh38
rs2279594 Chr.15: 100649691-100649691 on Build GRCh38
rs751865257 Chr.15: 100649350-100649350 on Build GRCh38
rs935440080 Chr.15: 100649701-100649701 on Build GRCh38
rs775634665 Chr.15: 100649788-100649788 on Build GRCh38
rs12914456 Chr.15: 100649513-100649513 on Build GRCh38
rs531925413 Chr.15: 100649365-100649365 on Build GRCh38
rs149844367 Chr.15: 100649682-100649682 on Build GRCh38
rs761204112 Chr.15: 100649505-100649505 on Build GRCh38
rs182374876 Chr.15: 100649653-100649653 on Build GRCh38
rs546413230 Chr.15: 100649357-100649357 on Build GRCh38
rs558669882 Chr.15: 100649722-100649722 on Build GRCh38
rs749767312 Chr.15: 100649544-100649544 on Build GRCh38
rs771639312 Chr.15: 100649671-100649671 on Build GRCh38
rs991184491 Chr.15: 100649647-100649647 on Build GRCh38
rs867201014 Chr.15: 100649644-100649644 on Build GRCh38
rs1038598083 Chr.15: 100649438-100649438 on Build GRCh38
rs532795139 Chr.15: 100649459-100649459 on Build GRCh38
rs535867963 Chr.15: 100649627-100649627 on Build GRCh38
rs531382278 Chr.15: 100649543-100649543 on Build GRCh38
rs371832817 Chr.15: 100649524-100649524 on Build GRCh38
rs189796614 Chr.15: 100649540-100649540 on Build GRCh38
rs1002892169 Chr.15: 100649534-100649534 on Build GRCh38
rs983181829 Chr.15: 100649551-100649551 on Build GRCh38
rs185811920 Chr.15: 100649430-100649430 on Build GRCh38
rs757363663 Chr.15: 100649404-100649404 on Build GRCh38
rs968322757 Chr.15: 100649620-100649620 on Build GRCh38
rs759358154 Chr.15: 100649767-100649767 on Build GRCh38
rs765104361 Chr.15: 100649785-100649785 on Build GRCh38
rs1047186034 Chr.15: 100649519-100649519 on Build GRCh38
rs140003482 Chr.15: 100649468-100649468 on Build GRCh38
rs146163677 Chr.15: 100649425-100649425 on Build GRCh38

View Full Product Details