Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 101176546-101177091 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHSY1
Entrez Gene ID
22856
Gene Name
chondroitin sulfate synthase 1
Gene Aliases
CHSY, CSS1, ChSy-1, TPBS
UniGene
Hs.110488
Gene Chromosome Location
Chr.15: 101175723-101251932 on Build GRCh38
Transcripts
NM_014918.4 XM_006720435.3 XM_011521364.2 XM_017022011.1

SNP Detail

SNP ID Chromosome Location
rs28737322 Chr.15: 101176975-101176975 on Build GRCh38
rs3784524 Chr.15: 101176783-101176783 on Build GRCh38
rs1008478500 Chr.15: 101176812-101176812 on Build GRCh38
rs973579587 Chr.15: 101176702-101176702 on Build GRCh38
rs939551193 Chr.15: 101176664-101176664 on Build GRCh38
rs184438808 Chr.15: 101176630-101176630 on Build GRCh38
rs1049212457 Chr.15: 101177039-101177039 on Build GRCh38
rs996093075 Chr.15: 101176637-101176637 on Build GRCh38
rs145103094 Chr.15: 101176790-101176790 on Build GRCh38
rs576716148 Chr.15: 101176622-101176622 on Build GRCh38
rs772830207 Chr.15: 101176829-101176829 on Build GRCh38
rs1027640501 Chr.15: 101176852-101176852 on Build GRCh38
rs954359602 Chr.15: 101176642-101176642 on Build GRCh38
rs548084719 Chr.15: 101176964-101176964 on Build GRCh38
rs34904116 Chr.15: 101176966-101176967 on Build GRCh38
rs775543015 Chr.15: 101176608-101176608 on Build GRCh38
rs772933595 Chr.15: 101177024-101177024 on Build GRCh38
rs924941482 Chr.15: 101176976-101176979 on Build GRCh38
rs1027877931 Chr.15: 101176603-101176603 on Build GRCh38
rs1008531107 Chr.15: 101176657-101176657 on Build GRCh38
rs529864405 Chr.15: 101176638-101176638 on Build GRCh38
rs1008832147 Chr.15: 101176977-101176977 on Build GRCh38
rs559100661 Chr.15: 101176635-101176635 on Build GRCh38
rs943855940 Chr.15: 101177008-101177008 on Build GRCh38
rs903552261 Chr.15: 101176713-101176713 on Build GRCh38
rs536810585 Chr.15: 101176976-101176976 on Build GRCh38
rs754071268 Chr.15: 101176907-101176907 on Build GRCh38
rs566256771 Chr.15: 101176974-101176974 on Build GRCh38
rs1039574512 Chr.15: 101177017-101177017 on Build GRCh38
rs996453287 Chr.15: 101176895-101176895 on Build GRCh38
rs374348631 Chr.15: 101176689-101176689 on Build GRCh38
rs962038906 Chr.15: 101176998-101176998 on Build GRCh38
rs1014574957 Chr.15: 101176995-101176995 on Build GRCh38
rs563337847 Chr.15: 101176888-101176888 on Build GRCh38
rs769530999 Chr.15: 101176577-101176577 on Build GRCh38
rs969990859 Chr.15: 101176719-101176719 on Build GRCh38
rs550191571 Chr.15: 101176792-101176792 on Build GRCh38
rs3784523 Chr.15: 101176860-101176860 on Build GRCh38
rs116051847 Chr.15: 101176591-101176591 on Build GRCh38
rs935029395 Chr.15: 101176976-101176977 on Build GRCh38
rs796689764 Chr.15: 101176967-101176967 on Build GRCh38
rs774187627 Chr.15: 101176762-101176762 on Build GRCh38
rs3784525 Chr.15: 101176690-101176690 on Build GRCh38
rs904350093 Chr.15: 101176862-101176862 on Build GRCh38
rs749384044 Chr.15: 101176623-101176623 on Build GRCh38
rs760403246 Chr.15: 101177051-101177051 on Build GRCh38
rs201939118 Chr.15: 101176974-101176975 on Build GRCh38
rs755262623 Chr.15: 101177036-101177036 on Build GRCh38
rs960867221 Chr.15: 101176963-101176963 on Build GRCh38
rs920756440 Chr.15: 101176703-101176703 on Build GRCh38
rs935249088 Chr.15: 101176721-101176721 on Build GRCh38
rs904377170 Chr.15: 101176631-101176631 on Build GRCh38
rs554865684 Chr.15: 101176978-101176978 on Build GRCh38
rs775287163 Chr.15: 101176585-101176585 on Build GRCh38
rs193176744 Chr.15: 101176570-101176570 on Build GRCh38
rs969091298 Chr.15: 101176975-101176976 on Build GRCh38
rs766519228 Chr.15: 101176882-101176882 on Build GRCh38
rs980985873 Chr.15: 101176750-101176750 on Build GRCh38
rs961793051 Chr.15: 101176701-101176701 on Build GRCh38
rs761623559 Chr.15: 101176752-101176752 on Build GRCh38
rs946801677 Chr.15: 101176832-101176832 on Build GRCh38
rs539319307 Chr.15: 101176911-101176911 on Build GRCh38
rs3784526 Chr.15: 101176592-101176592 on Build GRCh38
rs994584084 Chr.15: 101177022-101177022 on Build GRCh38
rs564583524 Chr.15: 101176611-101176611 on Build GRCh38

View Full Product Details