Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 281654-282146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGDIG
Entrez Gene ID
398
Gene Name
Rho GDP dissociation inhibitor gamma
Gene Aliases
RHOGDI-3
UniGene
Hs.647388
Gene Chromosome Location
Chr.16: 280606-283003 on Build GRCh38
Transcripts
NM_001176.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
382808 c.336A>T p.T112T Chr.16: 282107-282107 on Build GRCh38
5949311 c.265C>A p.P89T Chr.16: 282036-282036 on Build GRCh38
3690982 c.195C>T p.D65D Chr.16: 281867-281867 on Build GRCh38
4060572 c.174G>T p.E58D Chr.16: 281846-281846 on Build GRCh38
392493 c.316delC p.V107fs*12 Chr.16: 282087-282087 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555974578 Chr.16: 281728-281728 on Build GRCh38
rs145631710 Chr.16: 281787-281787 on Build GRCh38
rs780512495 Chr.16: 281831-281831 on Build GRCh38
rs867453530 Chr.16: 282084-282084 on Build GRCh38
rs771842809 Chr.16: 281786-281786 on Build GRCh38
rs751866015 Chr.16: 281938-281938 on Build GRCh38
rs886364883 Chr.16: 282055-282055 on Build GRCh38
rs944397369 Chr.16: 282118-282118 on Build GRCh38
rs777046495 Chr.16: 281918-281918 on Build GRCh38
rs201719567 Chr.16: 281988-281988 on Build GRCh38
rs376514597 Chr.16: 282061-282061 on Build GRCh38
rs755466026 Chr.16: 281824-281824 on Build GRCh38
rs772708545 Chr.16: 281748-281748 on Build GRCh38
rs771861387 Chr.16: 281743-281743 on Build GRCh38
rs567751010 Chr.16: 281737-281737 on Build GRCh38
rs8063732 Chr.16: 281959-281959 on Build GRCh38
rs964452083 Chr.16: 282070-282070 on Build GRCh38
rs756794428 Chr.16: 281900-281900 on Build GRCh38
rs372112033 Chr.16: 282041-282041 on Build GRCh38
rs749641507 Chr.16: 281733-281733 on Build GRCh38
rs201323302 Chr.16: 281966-281966 on Build GRCh38
rs900313187 Chr.16: 282013-282013 on Build GRCh38
rs759661075 Chr.16: 281921-281921 on Build GRCh38
rs1011757083 Chr.16: 282065-282065 on Build GRCh38
rs1008720034 Chr.16: 281685-281685 on Build GRCh38
rs758886390 Chr.16: 281826-281826 on Build GRCh38
rs141891388 Chr.16: 281818-281818 on Build GRCh38
rs758435281 Chr.16: 282071-282071 on Build GRCh38
rs747751783 Chr.16: 281738-281738 on Build GRCh38
rs781200961 Chr.16: 281720-281720 on Build GRCh38
rs760748300 Chr.16: 281866-281866 on Build GRCh38
rs745387625 Chr.16: 281840-281840 on Build GRCh38
rs767124526 Chr.16: 281957-281957 on Build GRCh38
rs1031944519 Chr.16: 282033-282033 on Build GRCh38
rs78288331 Chr.16: 281777-281777 on Build GRCh38
rs775737017 Chr.16: 281931-281931 on Build GRCh38
rs765370268 Chr.16: 281922-281922 on Build GRCh38
rs1012995287 Chr.16: 281944-281944 on Build GRCh38
rs768293890 Chr.16: 281854-281854 on Build GRCh38
rs772037051 Chr.16: 281978-281978 on Build GRCh38
rs757503760 Chr.16: 281716-281716 on Build GRCh38
rs766759594 Chr.16: 281886-281887 on Build GRCh38
rs764496448 Chr.16: 281995-281995 on Build GRCh38
rs774145624 Chr.16: 281727-281727 on Build GRCh38
rs1038786328 Chr.16: 282011-282011 on Build GRCh38
rs763312757 Chr.16: 281933-281933 on Build GRCh38
rs1047590215 Chr.16: 281675-281675 on Build GRCh38
rs774217485 Chr.16: 281863-281863 on Build GRCh38
rs771042604 Chr.16: 281917-281917 on Build GRCh38
rs890285772 Chr.16: 281679-281679 on Build GRCh38
rs779626119 Chr.16: 281825-281825 on Build GRCh38
rs765989182 Chr.16: 282116-282116 on Build GRCh38
rs769333260 Chr.16: 281906-281906 on Build GRCh38
rs762257458 Chr.16: 282026-282026 on Build GRCh38
rs765821517 Chr.16: 281870-281870 on Build GRCh38
rs777454652 Chr.16: 281827-281827 on Build GRCh38
rs763159792 Chr.16: 281994-281994 on Build GRCh38
rs770390691 Chr.16: 281803-281803 on Build GRCh38
rs150651820 Chr.16: 281821-281821 on Build GRCh38
rs147346513 Chr.16: 282092-282092 on Build GRCh38
rs999044834 Chr.16: 282022-282022 on Build GRCh38
rs745359080 Chr.16: 281901-281901 on Build GRCh38
rs115646977 Chr.16: 282117-282117 on Build GRCh38
rs973789519 Chr.16: 281705-281705 on Build GRCh38
rs761083032 Chr.16: 282095-282095 on Build GRCh38
rs148414600 Chr.16: 282093-282093 on Build GRCh38
rs760173060 Chr.16: 281749-281749 on Build GRCh38
rs950809169 Chr.16: 281735-281735 on Build GRCh38
rs947007563 Chr.16: 281779-281779 on Build GRCh38
rs754790184 Chr.16: 281696-281696 on Build GRCh38
rs565538630 Chr.16: 282090-282090 on Build GRCh38
rs746605168 Chr.16: 281828-281828 on Build GRCh38
rs867998876 Chr.16: 281880-281880 on Build GRCh38
rs747869966 Chr.16: 281967-281967 on Build GRCh38
rs999097235 Chr.16: 281802-281802 on Build GRCh38
rs377627632 Chr.16: 281774-281774 on Build GRCh38
rs367906016 Chr.16: 282017-282017 on Build GRCh38
rs775199400 Chr.16: 281813-281813 on Build GRCh38
rs149498191 Chr.16: 281867-281867 on Build GRCh38
rs556875250 Chr.16: 281776-281776 on Build GRCh38
rs764978508 Chr.16: 281767-281767 on Build GRCh38
rs372294740 Chr.16: 282086-282086 on Build GRCh38
rs775646931 Chr.16: 281989-281989 on Build GRCh38
rs960856262 Chr.16: 281742-281742 on Build GRCh38
rs767819340 Chr.16: 282027-282027 on Build GRCh38
rs909143275 Chr.16: 282115-282115 on Build GRCh38
rs776484754 Chr.16: 281758-281758 on Build GRCh38
rs764669530 Chr.16: 281937-281937 on Build GRCh38
rs766085046 Chr.16: 282038-282038 on Build GRCh38
rs750503079 Chr.16: 281775-281775 on Build GRCh38
rs983535415 Chr.16: 281738-281739 on Build GRCh38
rs1004847941 Chr.16: 282059-282059 on Build GRCh38
rs147198180 Chr.16: 282045-282045 on Build GRCh38
rs750036782 Chr.16: 281958-281958 on Build GRCh38
rs117235505 Chr.16: 282066-282066 on Build GRCh38
rs748524908 Chr.16: 281785-281785 on Build GRCh38
rs894566263 Chr.16: 281914-281914 on Build GRCh38
rs773176723 Chr.16: 281983-281983 on Build GRCh38
rs894442219 Chr.16: 281773-281773 on Build GRCh38
rs755561283 Chr.16: 281781-281781 on Build GRCh38
rs140573990 Chr.16: 282082-282082 on Build GRCh38
rs754865357 Chr.16: 281965-281965 on Build GRCh38
rs200015949 Chr.16: 282028-282028 on Build GRCh38
rs762793508 Chr.16: 281817-281817 on Build GRCh38
rs746042485 Chr.16: 281984-281984 on Build GRCh38
rs775840106 Chr.16: 282089-282089 on Build GRCh38
rs776238542 Chr.16: 281804-281804 on Build GRCh38
rs941891602 Chr.16: 281891-281891 on Build GRCh38
rs763283100 Chr.16: 281781-281789 on Build GRCh38
rs751821634 Chr.16: 281699-281699 on Build GRCh38
rs114176225 Chr.16: 281850-281850 on Build GRCh38
rs757699880 Chr.16: 281948-281948 on Build GRCh38
rs199591071 Chr.16: 281769-281769 on Build GRCh38
rs371718533 Chr.16: 281909-281909 on Build GRCh38
rs765174306 Chr.16: 281811-281811 on Build GRCh38
rs928071033 Chr.16: 281741-281741 on Build GRCh38
rs769719632 Chr.16: 281849-281849 on Build GRCh38
rs766318916 Chr.16: 281822-281822 on Build GRCh38
rs753300269 Chr.16: 281881-281881 on Build GRCh38
rs746508423 Chr.16: 281799-281799 on Build GRCh38
rs759298517 Chr.16: 281760-281760 on Build GRCh38
rs531387216 Chr.16: 281915-281915 on Build GRCh38
rs1052796043 Chr.16: 281796-281796 on Build GRCh38
rs758978302 Chr.16: 281892-281892 on Build GRCh38
rs201597842 Chr.16: 281851-281851 on Build GRCh38
rs144552315 Chr.16: 282083-282083 on Build GRCh38
rs752103994 Chr.16: 281961-281961 on Build GRCh38
rs929197834 Chr.16: 281864-281864 on Build GRCh38
rs535922142 Chr.16: 281694-281694 on Build GRCh38
rs764282089 Chr.16: 281698-281698 on Build GRCh38
rs756171428 Chr.16: 282104-282104 on Build GRCh38
rs748832283 Chr.16: 281961-281961 on Build GRCh38
rs373658766 Chr.16: 281912-281912 on Build GRCh38
rs777953367 Chr.16: 281893-281893 on Build GRCh38
rs759475334 Chr.16: 281810-281810 on Build GRCh38
rs1038477127 Chr.16: 281751-281751 on Build GRCh38
rs766330406 Chr.16: 281755-281755 on Build GRCh38
rs1023764690 Chr.16: 282082-282083 on Build GRCh38

View Full Product Details