Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 281877-282382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGDIG
Entrez Gene ID
398
Gene Name
Rho GDP dissociation inhibitor gamma
Gene Aliases
RHOGDI-3
UniGene
Hs.647388
Gene Chromosome Location
Chr.16: 280606-283003 on Build GRCh38
Transcripts
NM_001176.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
382808 c.336A>T p.T112T Chr.16: 282107-282107 on Build GRCh38
5990348 c.357G>A p.K119K Chr.16: 282316-282316 on Build GRCh38
5949311 c.265C>A p.P89T Chr.16: 282036-282036 on Build GRCh38
392493 c.316delC p.V107fs*12 Chr.16: 282087-282087 on Build GRCh38
396432 c.300G>T p.S100S Chr.16: 282071-282071 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867453530 Chr.16: 282084-282084 on Build GRCh38
rs777925710 Chr.16: 282143-282143 on Build GRCh38
rs751866015 Chr.16: 281938-281938 on Build GRCh38
rs776991799 Chr.16: 282332-282332 on Build GRCh38
rs886364883 Chr.16: 282055-282055 on Build GRCh38
rs944397369 Chr.16: 282118-282118 on Build GRCh38
rs777046495 Chr.16: 281918-281918 on Build GRCh38
rs762523264 Chr.16: 282352-282352 on Build GRCh38
rs201719567 Chr.16: 281988-281988 on Build GRCh38
rs775334163 Chr.16: 282350-282350 on Build GRCh38
rs376514597 Chr.16: 282061-282061 on Build GRCh38
rs746288496 Chr.16: 282340-282340 on Build GRCh38
rs8063732 Chr.16: 281959-281959 on Build GRCh38
rs964452083 Chr.16: 282070-282070 on Build GRCh38
rs759146295 Chr.16: 282136-282136 on Build GRCh38
rs756794428 Chr.16: 281900-281900 on Build GRCh38
rs372112033 Chr.16: 282041-282041 on Build GRCh38
rs868311072 Chr.16: 282305-282305 on Build GRCh38
rs201323302 Chr.16: 281966-281966 on Build GRCh38
rs900313187 Chr.16: 282013-282013 on Build GRCh38
rs772551912 Chr.16: 282250-282250 on Build GRCh38
rs759661075 Chr.16: 281921-281921 on Build GRCh38
rs751588062 Chr.16: 282180-282180 on Build GRCh38
rs1011757083 Chr.16: 282065-282065 on Build GRCh38
rs554184565 Chr.16: 282216-282216 on Build GRCh38
rs758435281 Chr.16: 282071-282071 on Build GRCh38
rs769474713 Chr.16: 282344-282344 on Build GRCh38
rs767124526 Chr.16: 281957-281957 on Build GRCh38
rs1031944519 Chr.16: 282033-282033 on Build GRCh38
rs775737017 Chr.16: 281931-281931 on Build GRCh38
rs765370268 Chr.16: 281922-281922 on Build GRCh38
rs780880385 Chr.16: 282155-282155 on Build GRCh38
rs200937476 Chr.16: 282270-282270 on Build GRCh38
rs1012995287 Chr.16: 281944-281944 on Build GRCh38
rs772037051 Chr.16: 281978-281978 on Build GRCh38
rs764496448 Chr.16: 281995-281995 on Build GRCh38
rs1038786328 Chr.16: 282011-282011 on Build GRCh38
rs150946433 Chr.16: 282329-282329 on Build GRCh38
rs763312757 Chr.16: 281933-281933 on Build GRCh38
rs777550189 Chr.16: 282178-282178 on Build GRCh38
rs536785569 Chr.16: 282229-282229 on Build GRCh38
rs771042604 Chr.16: 281917-281917 on Build GRCh38
rs765989182 Chr.16: 282116-282116 on Build GRCh38
rs1001017192 Chr.16: 282190-282190 on Build GRCh38
rs769333260 Chr.16: 281906-281906 on Build GRCh38
rs767967129 Chr.16: 282161-282161 on Build GRCh38
rs762257458 Chr.16: 282026-282026 on Build GRCh38
rs921664670 Chr.16: 282258-282258 on Build GRCh38
rs763159792 Chr.16: 281994-281994 on Build GRCh38
rs750187935 Chr.16: 282290-282290 on Build GRCh38
rs755458844 Chr.16: 282139-282142 on Build GRCh38
rs764760374 Chr.16: 282138-282138 on Build GRCh38
rs752164615 Chr.16: 282139-282139 on Build GRCh38
rs940602313 Chr.16: 282151-282151 on Build GRCh38
rs147346513 Chr.16: 282092-282092 on Build GRCh38
rs999044834 Chr.16: 282022-282022 on Build GRCh38
rs745359080 Chr.16: 281901-281901 on Build GRCh38
rs115646977 Chr.16: 282117-282117 on Build GRCh38
rs761083032 Chr.16: 282095-282095 on Build GRCh38
rs758345150 Chr.16: 282141-282141 on Build GRCh38
rs950333827 Chr.16: 282276-282276 on Build GRCh38
rs148414600 Chr.16: 282093-282093 on Build GRCh38
rs200441576 Chr.16: 282293-282293 on Build GRCh38
rs776886528 Chr.16: 282263-282263 on Build GRCh38
rs565538630 Chr.16: 282090-282090 on Build GRCh38
rs376247669 Chr.16: 282145-282145 on Build GRCh38
rs747869966 Chr.16: 281967-281967 on Build GRCh38
rs367906016 Chr.16: 282017-282017 on Build GRCh38
rs372294740 Chr.16: 282086-282086 on Build GRCh38
rs775646931 Chr.16: 281989-281989 on Build GRCh38
rs767819340 Chr.16: 282027-282027 on Build GRCh38
rs554741124 Chr.16: 282252-282252 on Build GRCh38
rs759172417 Chr.16: 282269-282269 on Build GRCh38
rs909143275 Chr.16: 282115-282115 on Build GRCh38
rs571979563 Chr.16: 282169-282169 on Build GRCh38
rs751588124 Chr.16: 282275-282275 on Build GRCh38
rs903937618 Chr.16: 282186-282186 on Build GRCh38
rs764669530 Chr.16: 281937-281937 on Build GRCh38
rs766085046 Chr.16: 282038-282038 on Build GRCh38
rs771290643 Chr.16: 282254-282254 on Build GRCh38
rs1004847941 Chr.16: 282059-282059 on Build GRCh38
rs772174910 Chr.16: 282301-282301 on Build GRCh38
rs147198180 Chr.16: 282045-282045 on Build GRCh38
rs373716691 Chr.16: 282291-282291 on Build GRCh38
rs117235505 Chr.16: 282066-282066 on Build GRCh38
rs750036782 Chr.16: 281958-281958 on Build GRCh38
rs894566263 Chr.16: 281914-281914 on Build GRCh38
rs773176723 Chr.16: 281983-281983 on Build GRCh38
rs140573990 Chr.16: 282082-282082 on Build GRCh38
rs111793116 Chr.16: 282271-282271 on Build GRCh38
rs754865357 Chr.16: 281965-281965 on Build GRCh38
rs1019861125 Chr.16: 282223-282223 on Build GRCh38
rs200015949 Chr.16: 282028-282028 on Build GRCh38
rs539290566 Chr.16: 282203-282203 on Build GRCh38
rs749477550 Chr.16: 282156-282156 on Build GRCh38
rs746042485 Chr.16: 281984-281984 on Build GRCh38
rs775840106 Chr.16: 282089-282089 on Build GRCh38
rs369485603 Chr.16: 282343-282343 on Build GRCh38
rs763648457 Chr.16: 282357-282357 on Build GRCh38
rs778982863 Chr.16: 282247-282247 on Build GRCh38
rs372918790 Chr.16: 282159-282159 on Build GRCh38
rs757699880 Chr.16: 281948-281948 on Build GRCh38
rs1016554903 Chr.16: 282347-282347 on Build GRCh38
rs371718533 Chr.16: 281909-281909 on Build GRCh38
rs907191509 Chr.16: 282202-282202 on Build GRCh38
rs1017762514 Chr.16: 282231-282231 on Build GRCh38
rs142556476 Chr.16: 282325-282325 on Build GRCh38
rs748291024 Chr.16: 282248-282248 on Build GRCh38
rs900698884 Chr.16: 282251-282251 on Build GRCh38
rs777858497 Chr.16: 282311-282311 on Build GRCh38
rs531387216 Chr.16: 281915-281915 on Build GRCh38
rs144552315 Chr.16: 282083-282083 on Build GRCh38
rs752103994 Chr.16: 281961-281961 on Build GRCh38
rs940528772 Chr.16: 282278-282278 on Build GRCh38
rs764101027 Chr.16: 282274-282274 on Build GRCh38
rs75289488 Chr.16: 282148-282148 on Build GRCh38
rs572337864 Chr.16: 282228-282228 on Build GRCh38
rs756171428 Chr.16: 282104-282104 on Build GRCh38
rs780983164 Chr.16: 282281-282281 on Build GRCh38
rs748832283 Chr.16: 281961-281961 on Build GRCh38
rs373658766 Chr.16: 281912-281912 on Build GRCh38
rs574851094 Chr.16: 282279-282279 on Build GRCh38
rs1031598718 Chr.16: 282192-282192 on Build GRCh38
rs1056424527 Chr.16: 282153-282153 on Build GRCh38
rs1023764690 Chr.16: 282082-282083 on Build GRCh38
rs956921102 Chr.16: 282199-282199 on Build GRCh38

View Full Product Details