Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 287362-287856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AXIN1
Entrez Gene ID
8312
Gene Name
axin 1
Gene Aliases
AXIN, PPP1R49
UniGene
Hs.592082
Gene Chromosome Location
Chr.16: 287440-355226 on Build GRCh38
Transcripts
NM_003502.3 NM_181050.2 NR_134879.1 XM_011522682.2 XM_011522683.2 XM_011522684.2 XM_011522686.1 XM_017023743.1 XM_017023744.1 XM_017023745.1 XM_017023746.1

SNP Detail

SNP ID Chromosome Location
rs973718176 Chr.16: 287769-287769 on Build GRCh38
rs45603234 Chr.16: 287820-287820 on Build GRCh38
rs571884326 Chr.16: 287480-287480 on Build GRCh38
rs200725501 Chr.16: 287826-287827 on Build GRCh38
rs1005731081 Chr.16: 287697-287697 on Build GRCh38
rs1055612598 Chr.16: 287435-287435 on Build GRCh38
rs937282466 Chr.16: 287423-287423 on Build GRCh38
rs1043014467 Chr.16: 287835-287835 on Build GRCh38
rs553717791 Chr.16: 287457-287457 on Build GRCh38
rs755386222 Chr.16: 287494-287497 on Build GRCh38
rs537865043 Chr.16: 287413-287413 on Build GRCh38
rs984233113 Chr.16: 287420-287420 on Build GRCh38
rs1002735731 Chr.16: 287524-287524 on Build GRCh38
rs942691373 Chr.16: 287776-287776 on Build GRCh38
rs968890363 Chr.16: 287417-287417 on Build GRCh38
rs565801186 Chr.16: 287735-287735 on Build GRCh38
rs975469420 Chr.16: 287640-287640 on Build GRCh38
rs542383418 Chr.16: 287490-287490 on Build GRCh38
rs1016569946 Chr.16: 287699-287699 on Build GRCh38
rs752184161 Chr.16: 287639-287639 on Build GRCh38
rs983788711 Chr.16: 287594-287594 on Build GRCh38
rs575819356 Chr.16: 287705-287705 on Build GRCh38
rs772269532 Chr.16: 287631-287631 on Build GRCh38
rs556476403 Chr.16: 287414-287414 on Build GRCh38
rs893819075 Chr.16: 287649-287649 on Build GRCh38
rs1032791979 Chr.16: 287528-287528 on Build GRCh38
rs1017098515 Chr.16: 287823-287823 on Build GRCh38
rs397977450 Chr.16: 287549-287550 on Build GRCh38
rs931104992 Chr.16: 287500-287500 on Build GRCh38
rs929646771 Chr.16: 287399-287399 on Build GRCh38
rs991383284 Chr.16: 287703-287703 on Build GRCh38
rs756745324 Chr.16: 287394-287394 on Build GRCh38
rs148341718 Chr.16: 287765-287765 on Build GRCh38
rs895669044 Chr.16: 287440-287440 on Build GRCh38
rs553867836 Chr.16: 287706-287706 on Build GRCh38
rs878949036 Chr.16: 287382-287382 on Build GRCh38
rs969195767 Chr.16: 287589-287589 on Build GRCh38
rs541098940 Chr.16: 287615-287615 on Build GRCh38
rs1022130210 Chr.16: 287416-287416 on Build GRCh38
rs971337527 Chr.16: 287711-287711 on Build GRCh38
rs918217795 Chr.16: 287398-287398 on Build GRCh38
rs951165279 Chr.16: 287577-287577 on Build GRCh38
rs543053302 Chr.16: 287564-287564 on Build GRCh38
rs538782407 Chr.16: 287700-287700 on Build GRCh38
rs1018414467 Chr.16: 287553-287553 on Build GRCh38
rs559064493 Chr.16: 287432-287432 on Build GRCh38
rs395545 Chr.16: 287750-287750 on Build GRCh38
rs193056425 Chr.16: 287830-287830 on Build GRCh38
rs754831777 Chr.16: 287827-287828 on Build GRCh38
rs575891478 Chr.16: 287548-287548 on Build GRCh38
rs777554680 Chr.16: 287658-287658 on Build GRCh38
rs3833914 Chr.16: 287547-287548 on Build GRCh38
rs938519749 Chr.16: 287678-287679 on Build GRCh38
rs999532397 Chr.16: 287523-287523 on Build GRCh38
rs1005268873 Chr.16: 287821-287821 on Build GRCh38
rs879224462 Chr.16: 287739-287755 on Build GRCh38
rs1039711507 Chr.16: 287784-287784 on Build GRCh38
rs34665498 Chr.16: 287691-287691 on Build GRCh38
rs1024008534 Chr.16: 287834-287834 on Build GRCh38
rs541552430 Chr.16: 287692-287692 on Build GRCh38
rs539112038 Chr.16: 287441-287441 on Build GRCh38
rs998416540 Chr.16: 287805-287805 on Build GRCh38
rs377220 Chr.16: 287421-287421 on Build GRCh38
rs397840896 Chr.16: 287496-287499 on Build GRCh38
rs780918641 Chr.16: 287696-287696 on Build GRCh38
rs558296289 Chr.16: 287807-287807 on Build GRCh38
rs942353498 Chr.16: 287476-287476 on Build GRCh38
rs958521839 Chr.16: 287826-287826 on Build GRCh38
rs935407582 Chr.16: 287757-287757 on Build GRCh38
rs1039435414 Chr.16: 287489-287490 on Build GRCh38
rs1054286406 Chr.16: 287764-287764 on Build GRCh38
rs549561878 Chr.16: 287656-287656 on Build GRCh38
rs1041614332 Chr.16: 287634-287634 on Build GRCh38
rs183298341 Chr.16: 287576-287576 on Build GRCh38
rs912225792 Chr.16: 287453-287454 on Build GRCh38
rs370513758 Chr.16: 287497-287500 on Build GRCh38
rs965524812 Chr.16: 287554-287554 on Build GRCh38
rs45530432 Chr.16: 287655-287655 on Build GRCh38
rs575975905 Chr.16: 287770-287770 on Build GRCh38
rs34396403 Chr.16: 287828-287829 on Build GRCh38
rs191434623 Chr.16: 287517-287517 on Build GRCh38
rs3833913 Chr.16: 287492-287495 on Build GRCh38
rs982899680 Chr.16: 287783-287784 on Build GRCh38
rs976431536 Chr.16: 287566-287566 on Build GRCh38
rs935442933 Chr.16: 287645-287645 on Build GRCh38
rs529831203 Chr.16: 287682-287682 on Build GRCh38
rs977558179 Chr.16: 287610-287610 on Build GRCh38
rs61433087 Chr.16: 287836-287836 on Build GRCh38
rs9939865 Chr.16: 287613-287613 on Build GRCh38
rs543687441 Chr.16: 287788-287788 on Build GRCh38
rs1047316100 Chr.16: 287504-287504 on Build GRCh38
rs1047194052 Chr.16: 287818-287818 on Build GRCh38
rs894045759 Chr.16: 287532-287532 on Build GRCh38
rs746350083 Chr.16: 287650-287651 on Build GRCh38
rs999338725 Chr.16: 287721-287721 on Build GRCh38
rs1046526541 Chr.16: 287514-287514 on Build GRCh38
rs866649485 Chr.16: 287612-287612 on Build GRCh38
rs954821359 Chr.16: 287736-287736 on Build GRCh38
rs559462815 Chr.16: 287646-287646 on Build GRCh38
rs879366710 Chr.16: 287836-287837 on Build GRCh38
rs393521 Chr.16: 287678-287678 on Build GRCh38
rs764852968 Chr.16: 287616-287616 on Build GRCh38
rs987762801 Chr.16: 287452-287452 on Build GRCh38
rs1021683682 Chr.16: 287587-287587 on Build GRCh38

View Full Product Details