Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 309895-310340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AXIN1
Entrez Gene ID
8312
Gene Name
axin 1
Gene Aliases
AXIN, PPP1R49
UniGene
Hs.592082
Gene Chromosome Location
Chr.16: 287440-355226 on Build GRCh38
Transcripts
NM_003502.3 NM_181050.2 NR_134879.1 XM_011522682.2 XM_011522683.2 XM_011522684.2 XM_011522686.1 XM_017023743.1 XM_017023744.1 XM_017023745.1 XM_017023746.1 XM_017023747.1 XM_017023748.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1377326 c.1116+1G>A p.? Chr.16: 309972-309972 on Build GRCh38
7339193 c.1054C>A p.H352N Chr.16: 310035-310035 on Build GRCh38
233878 c.1020-9C>T p.? Chr.16: 310078-310078 on Build GRCh38
143745 c.1058G>A p.R353H Chr.16: 310031-310031 on Build GRCh38
6220270 c.1064_1065insG p.M356fs*68 Chr.16: 310024-310025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756893611 Chr.16: 310245-310245 on Build GRCh38
rs55931984 Chr.16: 310148-310148 on Build GRCh38
rs970119890 Chr.16: 309993-309993 on Build GRCh38
rs550737152 Chr.16: 310232-310232 on Build GRCh38
rs761401617 Chr.16: 310112-310112 on Build GRCh38
rs745589774 Chr.16: 309956-309956 on Build GRCh38
rs116234849 Chr.16: 309955-309955 on Build GRCh38
rs763869998 Chr.16: 310025-310025 on Build GRCh38
rs563996799 Chr.16: 310035-310035 on Build GRCh38
rs373190155 Chr.16: 310155-310155 on Build GRCh38
rs796665119 Chr.16: 310032-310032 on Build GRCh38
rs143161189 Chr.16: 310143-310143 on Build GRCh38
rs1048146513 Chr.16: 309957-309957 on Build GRCh38
rs930547361 Chr.16: 310279-310279 on Build GRCh38
rs965375893 Chr.16: 310299-310299 on Build GRCh38
rs930619034 Chr.16: 310133-310133 on Build GRCh38
rs369325988 Chr.16: 310044-310044 on Build GRCh38
rs546820473 Chr.16: 310181-310181 on Build GRCh38
rs751315416 Chr.16: 310240-310240 on Build GRCh38
rs1049456793 Chr.16: 310286-310286 on Build GRCh38
rs181442175 Chr.16: 310270-310270 on Build GRCh38
rs764741140 Chr.16: 309943-309943 on Build GRCh38
rs551213610 Chr.16: 310271-310271 on Build GRCh38
rs760776188 Chr.16: 310000-310000 on Build GRCh38
rs755837370 Chr.16: 310051-310051 on Build GRCh38
rs746540296 Chr.16: 309951-309951 on Build GRCh38
rs1038931805 Chr.16: 309915-309915 on Build GRCh38
rs763891787 Chr.16: 310192-310192 on Build GRCh38
rs386787549 Chr.16: 309953-309955 on Build GRCh38
rs930147023 Chr.16: 310152-310152 on Build GRCh38
rs749063966 Chr.16: 310068-310068 on Build GRCh38
rs1056553552 Chr.16: 310302-310302 on Build GRCh38
rs201756048 Chr.16: 309925-309925 on Build GRCh38
rs985462754 Chr.16: 310106-310106 on Build GRCh38
rs777137467 Chr.16: 310005-310005 on Build GRCh38
rs1018256425 Chr.16: 310289-310289 on Build GRCh38
rs137897147 Chr.16: 310023-310023 on Build GRCh38
rs529104744 Chr.16: 310230-310230 on Build GRCh38
rs191442698 Chr.16: 309932-309932 on Build GRCh38
rs773681463 Chr.16: 309928-309928 on Build GRCh38
rs2301522 Chr.16: 309953-309953 on Build GRCh38
rs771058655 Chr.16: 310078-310078 on Build GRCh38
rs370794582 Chr.16: 310092-310092 on Build GRCh38
rs763227952 Chr.16: 310093-310093 on Build GRCh38
rs771361430 Chr.16: 310001-310001 on Build GRCh38
rs534964404 Chr.16: 309940-309940 on Build GRCh38
rs561178597 Chr.16: 310308-310308 on Build GRCh38
rs918721273 Chr.16: 310151-310151 on Build GRCh38
rs900447510 Chr.16: 310132-310132 on Build GRCh38
rs944583768 Chr.16: 310052-310052 on Build GRCh38
rs768667099 Chr.16: 310196-310198 on Build GRCh38
rs759808424 Chr.16: 310011-310011 on Build GRCh38
rs895519011 Chr.16: 310188-310188 on Build GRCh38
rs907829268 Chr.16: 310291-310291 on Build GRCh38
rs951639530 Chr.16: 310310-310310 on Build GRCh38
rs145226052 Chr.16: 309982-309982 on Build GRCh38
rs528447263 Chr.16: 310156-310156 on Build GRCh38
rs747033703 Chr.16: 310074-310074 on Build GRCh38
rs779951904 Chr.16: 310055-310055 on Build GRCh38
rs937110441 Chr.16: 310170-310170 on Build GRCh38
rs760137026 Chr.16: 310087-310087 on Build GRCh38
rs769686933 Chr.16: 309963-309963 on Build GRCh38
rs900502133 Chr.16: 309919-309919 on Build GRCh38
rs925627763 Chr.16: 310020-310020 on Build GRCh38
rs369249132 Chr.16: 309922-309922 on Build GRCh38
rs949937185 Chr.16: 310197-310197 on Build GRCh38
rs902999505 Chr.16: 310228-310228 on Build GRCh38
rs748445916 Chr.16: 309981-309981 on Build GRCh38
rs568675596 Chr.16: 310248-310248 on Build GRCh38
rs752268532 Chr.16: 310015-310015 on Build GRCh38
rs755376498 Chr.16: 309941-309941 on Build GRCh38
rs36045489 Chr.16: 310043-310043 on Build GRCh38
rs777975737 Chr.16: 310071-310071 on Build GRCh38
rs940710999 Chr.16: 310301-310301 on Build GRCh38
rs921777447 Chr.16: 310272-310272 on Build GRCh38
rs763777610 Chr.16: 310100-310100 on Build GRCh38
rs3842949 Chr.16: 310303-310303 on Build GRCh38
rs751638186 Chr.16: 310031-310031 on Build GRCh38
rs1023933056 Chr.16: 309989-309989 on Build GRCh38
rs576025706 Chr.16: 310126-310126 on Build GRCh38
rs984248086 Chr.16: 310153-310153 on Build GRCh38
rs779000870 Chr.16: 309964-309964 on Build GRCh38
rs562210631 Chr.16: 310206-310206 on Build GRCh38
rs3842950 Chr.16: 310317-310317 on Build GRCh38
rs751311645 Chr.16: 310110-310110 on Build GRCh38
rs1055461664 Chr.16: 310180-310180 on Build GRCh38
rs200837956 Chr.16: 309995-309995 on Build GRCh38
rs587776627 Chr.16: 309973-310004 on Build GRCh38
rs765672618 Chr.16: 310012-310012 on Build GRCh38
rs777090301 Chr.16: 310086-310086 on Build GRCh38
rs372286097 Chr.16: 309944-309944 on Build GRCh38
rs374214679 Chr.16: 309946-309946 on Build GRCh38
rs980105282 Chr.16: 310179-310179 on Build GRCh38
rs1012454663 Chr.16: 310154-310154 on Build GRCh38
rs977012294 Chr.16: 310149-310149 on Build GRCh38

View Full Product Details