Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 647699-648193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCRIP2
Entrez Gene ID
84331
Gene Name
MAPK regulated corepressor interacting protein 2
Gene Aliases
C16orf14, FAM195A, c349E10.1
UniGene
Hs.706105
Gene Chromosome Location
Chr.16: 641849-648474 on Build GRCh38
Transcripts
NM_138418.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3717022 c.446C>T p.A149V Chr.16: 648153-648153 on Build GRCh38
3957760 c.362G>T p.G121V Chr.16: 647834-647834 on Build GRCh38
6215808 c.345C>T p.G115G Chr.16: 647817-647817 on Build GRCh38
4129246 c.319C>A p.Q107K Chr.16: 647791-647791 on Build GRCh38
6386843 c.407T>C p.L136P Chr.16: 647879-647879 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774790118 Chr.16: 648040-648040 on Build GRCh38
rs749073945 Chr.16: 648121-648121 on Build GRCh38
rs201848182 Chr.16: 647780-647780 on Build GRCh38
rs756748600 Chr.16: 648095-648095 on Build GRCh38
rs770896163 Chr.16: 647751-647751 on Build GRCh38
rs553476185 Chr.16: 648073-648073 on Build GRCh38
rs753106108 Chr.16: 647918-647918 on Build GRCh38
rs200227390 Chr.16: 647778-647778 on Build GRCh38
rs1024647923 Chr.16: 647984-647984 on Build GRCh38
rs1054945491 Chr.16: 647964-647964 on Build GRCh38
rs776819332 Chr.16: 648154-648154 on Build GRCh38
rs760751146 Chr.16: 647893-647893 on Build GRCh38
rs756649619 Chr.16: 647781-647781 on Build GRCh38
rs753124748 Chr.16: 648168-648168 on Build GRCh38
rs571059299 Chr.16: 648035-648035 on Build GRCh38
rs534783437 Chr.16: 648058-648058 on Build GRCh38
rs917887509 Chr.16: 648067-648067 on Build GRCh38
rs1048014188 Chr.16: 647943-647943 on Build GRCh38
rs371119487 Chr.16: 647758-647758 on Build GRCh38
rs770996271 Chr.16: 648145-648145 on Build GRCh38
rs923331589 Chr.16: 648026-648026 on Build GRCh38
rs745857285 Chr.16: 648096-648096 on Build GRCh38
rs766801904 Chr.16: 647907-647907 on Build GRCh38
rs769527227 Chr.16: 647861-647861 on Build GRCh38
rs940820976 Chr.16: 648055-648055 on Build GRCh38
rs377027034 Chr.16: 647750-647750 on Build GRCh38
rs1024653785 Chr.16: 647767-647767 on Build GRCh38
rs540352621 Chr.16: 647978-647978 on Build GRCh38
rs757147620 Chr.16: 647953-647953 on Build GRCh38
rs887647308 Chr.16: 647957-647957 on Build GRCh38
rs924842148 Chr.16: 647784-647784 on Build GRCh38
rs145625595 Chr.16: 648142-648142 on Build GRCh38
rs570314949 Chr.16: 648013-648013 on Build GRCh38
rs763306329 Chr.16: 648169-648169 on Build GRCh38
rs772197690 Chr.16: 648130-648130 on Build GRCh38
rs367641794 Chr.16: 648143-648143 on Build GRCh38
rs371488689 Chr.16: 647797-647797 on Build GRCh38
rs984665858 Chr.16: 648041-648041 on Build GRCh38
rs943327684 Chr.16: 647862-647862 on Build GRCh38
rs955870053 Chr.16: 647814-647814 on Build GRCh38
rs779744955 Chr.16: 648015-648015 on Build GRCh38
rs374043940 Chr.16: 647823-647823 on Build GRCh38
rs754359005 Chr.16: 647908-647908 on Build GRCh38
rs745691518 Chr.16: 647859-647859 on Build GRCh38
rs200729583 Chr.16: 648111-648111 on Build GRCh38
rs1056476445 Chr.16: 648059-648059 on Build GRCh38
rs774512747 Chr.16: 647769-647769 on Build GRCh38
rs767779448 Chr.16: 647881-647881 on Build GRCh38
rs746514508 Chr.16: 647836-647836 on Build GRCh38
rs13380608 Chr.16: 647963-647963 on Build GRCh38
rs552460180 Chr.16: 648021-648021 on Build GRCh38
rs776343645 Chr.16: 647756-647756 on Build GRCh38
rs771669521 Chr.16: 647734-647734 on Build GRCh38
rs1023846298 Chr.16: 647766-647766 on Build GRCh38
rs765735457 Chr.16: 647910-647910 on Build GRCh38
rs571089180 Chr.16: 647922-647922 on Build GRCh38
rs867240664 Chr.16: 647816-647816 on Build GRCh38
rs762065954 Chr.16: 647875-647875 on Build GRCh38
rs777421830 Chr.16: 647933-647933 on Build GRCh38
rs770783166 Chr.16: 647842-647842 on Build GRCh38
rs182370986 Chr.16: 647971-647971 on Build GRCh38
rs761691015 Chr.16: 648115-648116 on Build GRCh38
rs1032772707 Chr.16: 647804-647805 on Build GRCh38
rs1032241274 Chr.16: 648005-648005 on Build GRCh38
rs772755663 Chr.16: 647744-647744 on Build GRCh38
rs948044627 Chr.16: 648098-648098 on Build GRCh38
rs868056381 Chr.16: 647822-647822 on Build GRCh38
rs867794421 Chr.16: 647786-647786 on Build GRCh38
rs531252054 Chr.16: 648014-648014 on Build GRCh38
rs957846635 Chr.16: 647808-647808 on Build GRCh38
rs768437684 Chr.16: 648034-648034 on Build GRCh38
rs964966251 Chr.16: 648019-648019 on Build GRCh38
rs779895686 Chr.16: 648115-648115 on Build GRCh38
rs776561608 Chr.16: 647853-647853 on Build GRCh38
rs761906698 Chr.16: 647770-647770 on Build GRCh38
rs775246231 Chr.16: 647868-647868 on Build GRCh38
rs1013225254 Chr.16: 647764-647764 on Build GRCh38
rs765177328 Chr.16: 648122-648123 on Build GRCh38
rs528939024 Chr.16: 647991-647991 on Build GRCh38
rs965961730 Chr.16: 647832-647832 on Build GRCh38
rs903990973 Chr.16: 647968-647968 on Build GRCh38
rs907964294 Chr.16: 647891-647891 on Build GRCh38
rs999314795 Chr.16: 647989-647989 on Build GRCh38
rs966496005 Chr.16: 647771-647771 on Build GRCh38
rs1018652126 Chr.16: 648165-648165 on Build GRCh38
rs773226458 Chr.16: 647892-647892 on Build GRCh38
rs1045314567 Chr.16: 648106-648106 on Build GRCh38
rs930495162 Chr.16: 647878-647878 on Build GRCh38
rs550368708 Chr.16: 647981-647981 on Build GRCh38
rs550605194 Chr.16: 648008-648008 on Build GRCh38
rs983578166 Chr.16: 648068-648068 on Build GRCh38
rs200384428 Chr.16: 647829-647829 on Build GRCh38
rs929369473 Chr.16: 648029-648029 on Build GRCh38
rs541715975 Chr.16: 647833-647833 on Build GRCh38
rs556874707 Chr.16: 647896-647896 on Build GRCh38
rs902256855 Chr.16: 647988-647988 on Build GRCh38
rs367609553 Chr.16: 647824-647824 on Build GRCh38
rs754166759 Chr.16: 647800-647800 on Build GRCh38
rs556960361 Chr.16: 648167-648167 on Build GRCh38
rs374404215 Chr.16: 647763-647763 on Build GRCh38
rs868184520 Chr.16: 647805-647805 on Build GRCh38
rs769158333 Chr.16: 647997-647997 on Build GRCh38
rs575182270 Chr.16: 647909-647909 on Build GRCh38

View Full Product Details