Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 791054-791551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHTF18
Entrez Gene ID
63922
Gene Name
chromosome transmission fidelity factor 18
Gene Aliases
C16orf41, C321D2.2, C321D2.3, C321D2.4, CHL12, Ctf18, RUVBL
UniGene
Hs.153850
Gene Chromosome Location
Chr.16: 788622-798074 on Build GRCh38
Transcripts
NM_022092.2 XM_005255470.1 XM_005255471.3 XM_011522572.1 XM_011522573.1 XM_017023532.1 XM_017023533.1 XM_017023534.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6434554 c.1004G>A p.S335N Chr.16: 791270-791270 on Build GRCh38
7097856 c.1627G>A p.E543K Chr.16: 791308-791308 on Build GRCh38
6434552 c.1589G>A p.S530N Chr.16: 791270-791270 on Build GRCh38
7002740 c.1049C>T p.S350F Chr.16: 791231-791231 on Build GRCh38
7233807 c.967C>G p.R323G Chr.16: 791233-791233 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs939871355 Chr.16: 791487-791487 on Build GRCh38
rs919824166 Chr.16: 791427-791427 on Build GRCh38
rs752484196 Chr.16: 791316-791316 on Build GRCh38
rs765362941 Chr.16: 791372-791372 on Build GRCh38
rs781507569 Chr.16: 791150-791150 on Build GRCh38
rs777061042 Chr.16: 791112-791112 on Build GRCh38
rs181101102 Chr.16: 791477-791477 on Build GRCh38
rs2063911 Chr.16: 791175-791175 on Build GRCh38
rs768346736 Chr.16: 791223-791223 on Build GRCh38
rs758915562 Chr.16: 791147-791147 on Build GRCh38
rs746096068 Chr.16: 791154-791154 on Build GRCh38
rs750628374 Chr.16: 791212-791212 on Build GRCh38
rs151271197 Chr.16: 791238-791238 on Build GRCh38
rs1033584217 Chr.16: 791429-791429 on Build GRCh38
rs986936884 Chr.16: 791517-791517 on Build GRCh38
rs778839776 Chr.16: 791225-791225 on Build GRCh38
rs764352026 Chr.16: 791417-791417 on Build GRCh38
rs540462504 Chr.16: 791530-791530 on Build GRCh38
rs551612573 Chr.16: 791420-791420 on Build GRCh38
rs1057282499 Chr.16: 791474-791474 on Build GRCh38
rs755389605 Chr.16: 791090-791090 on Build GRCh38
rs753072586 Chr.16: 791178-791178 on Build GRCh38
rs756317834 Chr.16: 791263-791263 on Build GRCh38
rs759477822 Chr.16: 791400-791400 on Build GRCh38
rs772511994 Chr.16: 791393-791393 on Build GRCh38
rs746510810 Chr.16: 791281-791281 on Build GRCh38
rs528377032 Chr.16: 791176-791176 on Build GRCh38
rs113712337 Chr.16: 791518-791518 on Build GRCh38
rs747817344 Chr.16: 791437-791437 on Build GRCh38
rs752931732 Chr.16: 791373-791373 on Build GRCh38
rs199742669 Chr.16: 791258-791258 on Build GRCh38
rs777275083 Chr.16: 791351-791365 on Build GRCh38
rs759144007 Chr.16: 791290-791290 on Build GRCh38
rs372944800 Chr.16: 791396-791396 on Build GRCh38
rs557715203 Chr.16: 791120-791120 on Build GRCh38
rs113406233 Chr.16: 791077-791077 on Build GRCh38
rs956484143 Chr.16: 791310-791310 on Build GRCh38
rs377044412 Chr.16: 791106-791106 on Build GRCh38
rs771522450 Chr.16: 791232-791232 on Build GRCh38
rs781456747 Chr.16: 791203-791203 on Build GRCh38
rs771964364 Chr.16: 791277-791277 on Build GRCh38
rs375995023 Chr.16: 791198-791198 on Build GRCh38
rs748908080 Chr.16: 791376-791376 on Build GRCh38
rs573119207 Chr.16: 791525-791525 on Build GRCh38
rs568608547 Chr.16: 791262-791262 on Build GRCh38
rs533940723 Chr.16: 791433-791433 on Build GRCh38
rs956257711 Chr.16: 791432-791432 on Build GRCh38
rs759726281 Chr.16: 791364-791364 on Build GRCh38
rs776542587 Chr.16: 791361-791361 on Build GRCh38
rs555585580 Chr.16: 791476-791476 on Build GRCh38
rs564775730 Chr.16: 791524-791524 on Build GRCh38
rs921995203 Chr.16: 791463-791463 on Build GRCh38
rs376920307 Chr.16: 791170-791170 on Build GRCh38
rs754194228 Chr.16: 791089-791089 on Build GRCh38
rs201924399 Chr.16: 791412-791412 on Build GRCh38
rs769018984 Chr.16: 791292-791292 on Build GRCh38
rs569093423 Chr.16: 791116-791116 on Build GRCh38
rs749093068 Chr.16: 791221-791221 on Build GRCh38
rs771155658 Chr.16: 791108-791108 on Build GRCh38
rs774560775 Chr.16: 791296-791296 on Build GRCh38
rs952382696 Chr.16: 791138-791138 on Build GRCh38
rs968310302 Chr.16: 791492-791492 on Build GRCh38
rs756385419 Chr.16: 791218-791218 on Build GRCh38
rs374003888 Chr.16: 791166-791166 on Build GRCh38
rs946698680 Chr.16: 791228-791228 on Build GRCh38
rs537935315 Chr.16: 791498-791498 on Build GRCh38
rs757471554 Chr.16: 791149-791149 on Build GRCh38
rs758248563 Chr.16: 791317-791317 on Build GRCh38
rs569233413 Chr.16: 791404-791404 on Build GRCh38
rs752021490 Chr.16: 791192-791192 on Build GRCh38
rs775649713 Chr.16: 791406-791406 on Build GRCh38
rs766957989 Chr.16: 791360-791360 on Build GRCh38
rs373518407 Chr.16: 791514-791514 on Build GRCh38
rs916062999 Chr.16: 791151-791151 on Build GRCh38
rs535432028 Chr.16: 791107-791107 on Build GRCh38
rs545556243 Chr.16: 791086-791086 on Build GRCh38
rs778594129 Chr.16: 791267-791267 on Build GRCh38
rs375178408 Chr.16: 791381-791381 on Build GRCh38
rs769724607 Chr.16: 791115-791115 on Build GRCh38
rs755893851 Chr.16: 791340-791341 on Build GRCh38
rs757123635 Chr.16: 791385-791385 on Build GRCh38
rs756752592 Chr.16: 791331-791331 on Build GRCh38
rs768137200 Chr.16: 791301-791301 on Build GRCh38
rs770568072 Chr.16: 791234-791234 on Build GRCh38
rs1053176278 Chr.16: 791110-791110 on Build GRCh38
rs1021242366 Chr.16: 791490-791490 on Build GRCh38
rs112651326 Chr.16: 791511-791511 on Build GRCh38
rs1027332957 Chr.16: 791131-791131 on Build GRCh38
rs748976957 Chr.16: 791392-791392 on Build GRCh38
rs981462716 Chr.16: 791088-791088 on Build GRCh38
rs1001498936 Chr.16: 791501-791501 on Build GRCh38
rs751450855 Chr.16: 791384-791384 on Build GRCh38
rs961000540 Chr.16: 791188-791188 on Build GRCh38
rs375203153 Chr.16: 791307-791307 on Build GRCh38
rs547884181 Chr.16: 791506-791506 on Build GRCh38
rs745437526 Chr.16: 791333-791333 on Build GRCh38
rs763295767 Chr.16: 791183-791183 on Build GRCh38
rs764651155 Chr.16: 791184-791184 on Build GRCh38
rs1036936586 Chr.16: 791421-791421 on Build GRCh38
rs563655428 Chr.16: 791349-791349 on Build GRCh38
rs754039243 Chr.16: 791308-791308 on Build GRCh38
rs755710461 Chr.16: 791389-791389 on Build GRCh38
rs372267710 Chr.16: 791113-791113 on Build GRCh38
rs369884054 Chr.16: 791123-791123 on Build GRCh38
rs540340115 Chr.16: 791125-791125 on Build GRCh38
rs779329938 Chr.16: 791091-791091 on Build GRCh38
rs566552327 Chr.16: 791426-791426 on Build GRCh38
rs931206342 Chr.16: 791436-791436 on Build GRCh38
rs780261830 Chr.16: 791264-791264 on Build GRCh38
rs765535375 Chr.16: 791312-791312 on Build GRCh38
rs1035202789 Chr.16: 791351-791351 on Build GRCh38
rs772811355 Chr.16: 791171-791171 on Build GRCh38
rs748463556 Chr.16: 791348-791348 on Build GRCh38
rs761282835 Chr.16: 791357-791357 on Build GRCh38
rs745367765 Chr.16: 791388-791388 on Build GRCh38
rs372528223 Chr.16: 791329-791329 on Build GRCh38
rs752221385 Chr.16: 791247-791248 on Build GRCh38
rs771337295 Chr.16: 791395-791395 on Build GRCh38
rs370569554 Chr.16: 791216-791216 on Build GRCh38
rs777929912 Chr.16: 791102-791102 on Build GRCh38
rs369717724 Chr.16: 791336-791336 on Build GRCh38
rs773811024 Chr.16: 791352-791352 on Build GRCh38
rs377505482 Chr.16: 791330-791330 on Build GRCh38
rs200880754 Chr.16: 791311-791311 on Build GRCh38
rs748658836 Chr.16: 791098-791098 on Build GRCh38
rs550288774 Chr.16: 791249-791249 on Build GRCh38
rs755264725 Chr.16: 791137-791137 on Build GRCh38
rs199878079 Chr.16: 791233-791233 on Build GRCh38
rs776281537 Chr.16: 791288-791288 on Build GRCh38
rs760342767 Chr.16: 791174-791174 on Build GRCh38
rs774210020 Chr.16: 791163-791163 on Build GRCh38
rs372208068 Chr.16: 791287-791287 on Build GRCh38
rs753094586 Chr.16: 791145-791145 on Build GRCh38
rs373267846 Chr.16: 791148-791148 on Build GRCh38
rs911932119 Chr.16: 791172-791172 on Build GRCh38
rs767182487 Chr.16: 791102-791103 on Build GRCh38
rs753796694 Chr.16: 791222-791222 on Build GRCh38
rs905491457 Chr.16: 791455-791455 on Build GRCh38
rs777752422 Chr.16: 791278-791278 on Build GRCh38
rs891928135 Chr.16: 791531-791531 on Build GRCh38
rs532156558 Chr.16: 791242-791242 on Build GRCh38
rs369540346 Chr.16: 791259-791259 on Build GRCh38
rs769869083 Chr.16: 791403-791403 on Build GRCh38
rs763320442 Chr.16: 791246-791246 on Build GRCh38
rs780594242 Chr.16: 791332-791332 on Build GRCh38
rs996755739 Chr.16: 791128-791128 on Build GRCh38
rs539700061 Chr.16: 791119-791119 on Build GRCh38
rs370688299 Chr.16: 791391-791391 on Build GRCh38
rs773866071 Chr.16: 791303-791303 on Build GRCh38
rs369021522 Chr.16: 791140-791140 on Build GRCh38
rs1000336565 Chr.16: 791520-791520 on Build GRCh38
rs374149034 Chr.16: 791194-791194 on Build GRCh38
rs747984664 Chr.16: 791274-791274 on Build GRCh38
rs754758324 Chr.16: 791265-791265 on Build GRCh38
rs751328531 Chr.16: 791418-791418 on Build GRCh38
rs544258068 Chr.16: 791168-791168 on Build GRCh38
rs769676245 Chr.16: 791156-791156 on Build GRCh38
rs781235003 Chr.16: 791386-791386 on Build GRCh38
rs572396867 Chr.16: 791500-791500 on Build GRCh38
rs188666865 Chr.16: 791157-791157 on Build GRCh38
rs1044447507 Chr.16: 791516-791516 on Build GRCh38
rs762982444 Chr.16: 791118-791118 on Build GRCh38
rs373409796 Chr.16: 791337-791337 on Build GRCh38
rs368568398 Chr.16: 791254-791254 on Build GRCh38
rs944705541 Chr.16: 791347-791347 on Build GRCh38
rs535883233 Chr.16: 791394-791394 on Build GRCh38
rs1002628831 Chr.16: 791462-791462 on Build GRCh38
rs748062791 Chr.16: 791227-791227 on Build GRCh38
rs199983682 Chr.16: 791315-791315 on Build GRCh38
rs961216450 Chr.16: 791521-791521 on Build GRCh38
rs374015756 Chr.16: 791220-791220 on Build GRCh38
rs768632231 Chr.16: 791161-791161 on Build GRCh38
rs758368224 Chr.16: 791100-791100 on Build GRCh38
rs184417663 Chr.16: 791122-791122 on Build GRCh38
rs758123915 Chr.16: 791379-791379 on Build GRCh38

View Full Product Details