Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1153866-1154362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1H
Entrez Gene ID
8912
Gene Name
calcium voltage-gated channel subunit alpha1 H
Gene Aliases
CACNA1HB, Cav3.2, ECA6, EIG6
UniGene
Hs.459642
Gene Chromosome Location
Chr.16: 1153241-1221772 on Build GRCh38
Transcripts
NM_001005407.1 NM_021098.2 XM_005255652.4 XM_006720963.3 XM_006720964.3 XM_006720965.3 XM_006720967.3 XM_006720968.3 XM_011522727.2 XM_017023819.1 XM_017023820.1 XM_017023821.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6046152 c.153C>T p.S51S Chr.16: 1153890-1153890 on Build GRCh38
6046154 c.153C>T p.S51S Chr.16: 1153890-1153890 on Build GRCh38
5728157 c.252T>C p.G84G Chr.16: 1153989-1153989 on Build GRCh38
4651233 c.177C>A p.G59G Chr.16: 1153914-1153914 on Build GRCh38
4651235 c.177C>A p.G59G Chr.16: 1153914-1153914 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925110145 Chr.16: 1154302-1154302 on Build GRCh38
rs969994726 Chr.16: 1154188-1154188 on Build GRCh38
rs996002880 Chr.16: 1154246-1154246 on Build GRCh38
rs974426701 Chr.16: 1154252-1154252 on Build GRCh38
rs900768678 Chr.16: 1153967-1153967 on Build GRCh38
rs761812532 Chr.16: 1153953-1153953 on Build GRCh38
rs868339661 Chr.16: 1154333-1154333 on Build GRCh38
rs1012233171 Chr.16: 1154172-1154172 on Build GRCh38
rs935195220 Chr.16: 1154306-1154306 on Build GRCh38
rs911639825 Chr.16: 1154112-1154112 on Build GRCh38
rs765181693 Chr.16: 1154036-1154036 on Build GRCh38
rs994984264 Chr.16: 1153974-1153974 on Build GRCh38
rs759273300 Chr.16: 1154020-1154020 on Build GRCh38
rs547911587 Chr.16: 1154273-1154273 on Build GRCh38
rs878980599 Chr.16: 1154091-1154091 on Build GRCh38
rs1024087466 Chr.16: 1154173-1154173 on Build GRCh38
rs970406044 Chr.16: 1154045-1154045 on Build GRCh38
rs545660975 Chr.16: 1154338-1154338 on Build GRCh38
rs1039133104 Chr.16: 1154118-1154118 on Build GRCh38
rs374841130 Chr.16: 1154258-1154258 on Build GRCh38
rs562710782 Chr.16: 1154110-1154110 on Build GRCh38
rs915820138 Chr.16: 1153889-1153889 on Build GRCh38
rs966216601 Chr.16: 1154247-1154247 on Build GRCh38
rs976224496 Chr.16: 1153994-1153994 on Build GRCh38
rs565242327 Chr.16: 1154154-1154154 on Build GRCh38
rs949341963 Chr.16: 1154330-1154330 on Build GRCh38
rs757925753 Chr.16: 1154047-1154047 on Build GRCh38
rs921726348 Chr.16: 1153999-1153999 on Build GRCh38
rs1022859623 Chr.16: 1154041-1154041 on Build GRCh38
rs978586823 Chr.16: 1153899-1153899 on Build GRCh38
rs879053417 Chr.16: 1154128-1154128 on Build GRCh38
rs879196739 Chr.16: 1154230-1154230 on Build GRCh38
rs770277527 Chr.16: 1153913-1153913 on Build GRCh38
rs1045040818 Chr.16: 1154334-1154334 on Build GRCh38
rs979191693 Chr.16: 1154300-1154300 on Build GRCh38
rs557303824 Chr.16: 1153980-1153980 on Build GRCh38
rs573776247 Chr.16: 1154083-1154083 on Build GRCh38
rs753718656 Chr.16: 1154017-1154017 on Build GRCh38
rs539924701 Chr.16: 1154050-1154050 on Build GRCh38
rs896478139 Chr.16: 1154178-1154178 on Build GRCh38
rs1005061315 Chr.16: 1154162-1154162 on Build GRCh38
rs766061463 Chr.16: 1154016-1154016 on Build GRCh38
rs1027114085 Chr.16: 1154251-1154251 on Build GRCh38
rs921630107 Chr.16: 1154255-1154255 on Build GRCh38
rs879240111 Chr.16: 1154110-1154110 on Build GRCh38
rs1052288263 Chr.16: 1154325-1154325 on Build GRCh38
rs188293742 Chr.16: 1154130-1154130 on Build GRCh38
rs890167407 Chr.16: 1153941-1153941 on Build GRCh38
rs865917855 Chr.16: 1154146-1154146 on Build GRCh38
rs773413660 Chr.16: 1153997-1153997 on Build GRCh38
rs769155464 Chr.16: 1153928-1153928 on Build GRCh38
rs1043511168 Chr.16: 1154010-1154010 on Build GRCh38
rs1052712888 Chr.16: 1153932-1153932 on Build GRCh38
rs752583405 Chr.16: 1154044-1154044 on Build GRCh38
rs772150091 Chr.16: 1153977-1153977 on Build GRCh38
rs1032894798 Chr.16: 1154190-1154190 on Build GRCh38
rs966081215 Chr.16: 1154267-1154267 on Build GRCh38
rs777490298 Chr.16: 1154086-1154086 on Build GRCh38
rs1045493128 Chr.16: 1154227-1154227 on Build GRCh38
rs1019906289 Chr.16: 1154266-1154266 on Build GRCh38
rs879102520 Chr.16: 1154202-1154202 on Build GRCh38
rs527719120 Chr.16: 1154194-1154194 on Build GRCh38
rs940817491 Chr.16: 1154301-1154301 on Build GRCh38
rs746716155 Chr.16: 1153886-1153886 on Build GRCh38
rs112034409 Chr.16: 1154116-1154116 on Build GRCh38
rs995600485 Chr.16: 1154341-1154341 on Build GRCh38
rs749735503 Chr.16: 1153917-1153917 on Build GRCh38
rs147910127 Chr.16: 1154052-1154072 on Build GRCh38
rs886565487 Chr.16: 1154157-1154157 on Build GRCh38
rs1013614732 Chr.16: 1154211-1154211 on Build GRCh38
rs539324457 Chr.16: 1153908-1153908 on Build GRCh38
rs542496956 Chr.16: 1154109-1154109 on Build GRCh38
rs1052336679 Chr.16: 1154168-1154168 on Build GRCh38
rs764707005 Chr.16: 1154275-1154275 on Build GRCh38
rs988622480 Chr.16: 1154200-1154200 on Build GRCh38
rs748591413 Chr.16: 1154047-1154053 on Build GRCh38
rs1012676685 Chr.16: 1154034-1154034 on Build GRCh38
rs975749661 Chr.16: 1154270-1154270 on Build GRCh38
rs905248497 Chr.16: 1154235-1154235 on Build GRCh38
rs1049253328 Chr.16: 1154131-1154131 on Build GRCh38
rs911627014 Chr.16: 1154221-1154221 on Build GRCh38
rs951562635 Chr.16: 1154255-1154255 on Build GRCh38
rs1039258079 Chr.16: 1153958-1153958 on Build GRCh38
rs934440398 Chr.16: 1153929-1153929 on Build GRCh38
rs917820301 Chr.16: 1154328-1154328 on Build GRCh38
rs960703593 Chr.16: 1154019-1154019 on Build GRCh38
rs577235589 Chr.16: 1154007-1154007 on Build GRCh38
rs922195483 Chr.16: 1154119-1154119 on Build GRCh38
rs553478910 Chr.16: 1154051-1154051 on Build GRCh38
rs953118855 Chr.16: 1154294-1154294 on Build GRCh38
rs948554848 Chr.16: 1153891-1153891 on Build GRCh38
rs775960864 Chr.16: 1153914-1153914 on Build GRCh38
rs774748739 Chr.16: 1153949-1153949 on Build GRCh38
rs1035103368 Chr.16: 1154008-1154008 on Build GRCh38
rs1056521483 Chr.16: 1154163-1154163 on Build GRCh38
rs868008563 Chr.16: 1154063-1154063 on Build GRCh38
rs777123569 Chr.16: 1154052-1154058 on Build GRCh38

View Full Product Details