Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1195460-1195955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1H
Entrez Gene ID
8912
Gene Name
calcium voltage-gated channel subunit alpha1 H
Gene Aliases
CACNA1HB, Cav3.2, ECA6, EIG6
UniGene
Hs.459642
Gene Chromosome Location
Chr.16: 1153241-1221772 on Build GRCh38
Transcripts
NM_001005407.1 NM_021098.2 XM_005255652.4 XM_006720963.3 XM_006720964.3 XM_006720965.3 XM_006720967.3 XM_006720968.3 XM_011522724.2 XM_011522727.2 XM_017023819.1 XM_017023820.1 XM_017023821.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6618182 c.561G>A p.S187S Chr.16: 1195941-1195941 on Build GRCh38
7197328 c.451A>G p.M151V Chr.16: 1195471-1195471 on Build GRCh38
5701310 c.543_544insG p.M183fs*33 Chr.16: 1195563-1195564 on Build GRCh38
3957112 c.445G>T p.V149L Chr.16: 1195465-1195465 on Build GRCh38
4494985 c.443C>T p.A148V Chr.16: 1195463-1195463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536319083 Chr.16: 1195725-1195725 on Build GRCh38
rs954223968 Chr.16: 1195913-1195913 on Build GRCh38
rs60593994 Chr.16: 1195509-1195509 on Build GRCh38
rs922615065 Chr.16: 1195780-1195780 on Build GRCh38
rs757048691 Chr.16: 1195562-1195562 on Build GRCh38
rs980602127 Chr.16: 1195580-1195580 on Build GRCh38
rs1025077195 Chr.16: 1195739-1195739 on Build GRCh38
rs1017986326 Chr.16: 1195841-1195841 on Build GRCh38
rs976754165 Chr.16: 1195774-1195774 on Build GRCh38
rs748867788 Chr.16: 1195575-1195575 on Build GRCh38
rs535337528 Chr.16: 1195821-1195821 on Build GRCh38
rs570245223 Chr.16: 1195764-1195764 on Build GRCh38
rs377538101 Chr.16: 1195594-1195594 on Build GRCh38
rs372122003 Chr.16: 1195719-1195719 on Build GRCh38
rs936116544 Chr.16: 1195801-1195801 on Build GRCh38
rs760642532 Chr.16: 1195498-1195498 on Build GRCh38
rs148444212 Chr.16: 1195784-1195784 on Build GRCh38
rs35509671 Chr.16: 1195916-1195916 on Build GRCh38
rs977100988 Chr.16: 1195905-1195905 on Build GRCh38
rs145298036 Chr.16: 1195823-1195823 on Build GRCh38
rs369016752 Chr.16: 1195921-1195921 on Build GRCh38
rs935274303 Chr.16: 1195832-1195832 on Build GRCh38
rs956395086 Chr.16: 1195554-1195554 on Build GRCh38
rs376566207 Chr.16: 1195932-1195932 on Build GRCh38
rs61700478 Chr.16: 1195935-1195935 on Build GRCh38
rs1033461942 Chr.16: 1195749-1195749 on Build GRCh38
rs564878042 Chr.16: 1195646-1195646 on Build GRCh38
rs914627859 Chr.16: 1195815-1195815 on Build GRCh38
rs538387091 Chr.16: 1195747-1195747 on Build GRCh38
rs373468095 Chr.16: 1195748-1195748 on Build GRCh38
rs962355151 Chr.16: 1195750-1195750 on Build GRCh38
rs773672916 Chr.16: 1195519-1195519 on Build GRCh38
rs999386572 Chr.16: 1195716-1195716 on Build GRCh38
rs775863287 Chr.16: 1195923-1195923 on Build GRCh38
rs974113206 Chr.16: 1195754-1195754 on Build GRCh38
rs368216156 Chr.16: 1195911-1195911 on Build GRCh38
rs766955535 Chr.16: 1195528-1195528 on Build GRCh38
rs771440171 Chr.16: 1195599-1195599 on Build GRCh38
rs531432645 Chr.16: 1195582-1195582 on Build GRCh38
rs777285771 Chr.16: 1195530-1195530 on Build GRCh38
rs1055873165 Chr.16: 1195688-1195688 on Build GRCh38
rs939984306 Chr.16: 1195628-1195628 on Build GRCh38
rs544851523 Chr.16: 1195850-1195850 on Build GRCh38
rs552627906 Chr.16: 1195567-1195567 on Build GRCh38
rs569269448 Chr.16: 1195694-1195694 on Build GRCh38
rs754454536 Chr.16: 1195914-1195914 on Build GRCh38
rs922681307 Chr.16: 1195667-1195667 on Build GRCh38
rs892057896 Chr.16: 1195717-1195717 on Build GRCh38
rs760157365 Chr.16: 1195603-1195603 on Build GRCh38
rs761145870 Chr.16: 1195526-1195526 on Build GRCh38
rs119454947 Chr.16: 1195503-1195503 on Build GRCh38
rs1049900719 Chr.16: 1195669-1195669 on Build GRCh38
rs867317332 Chr.16: 1195852-1195852 on Build GRCh38
rs575823388 Chr.16: 1195838-1195838 on Build GRCh38
rs867084712 Chr.16: 1195504-1195504 on Build GRCh38
rs750325482 Chr.16: 1195907-1195907 on Build GRCh38
rs767363393 Chr.16: 1195906-1195906 on Build GRCh38
rs572601906 Chr.16: 1195817-1195817 on Build GRCh38
rs779875096 Chr.16: 1195912-1195912 on Build GRCh38
rs930167876 Chr.16: 1195651-1195651 on Build GRCh38
rs746919701 Chr.16: 1195498-1195498 on Build GRCh38
rs926108931 Chr.16: 1195798-1195798 on Build GRCh38
rs1043659476 Chr.16: 1195713-1195713 on Build GRCh38
rs765201603 Chr.16: 1195606-1195606 on Build GRCh38
rs902400992 Chr.16: 1195715-1195715 on Build GRCh38
rs770289037 Chr.16: 1195922-1195922 on Build GRCh38
rs888204371 Chr.16: 1195842-1195842 on Build GRCh38
rs58142268 Chr.16: 1195518-1195518 on Build GRCh38
rs562279659 Chr.16: 1195833-1195833 on Build GRCh38
rs118057853 Chr.16: 1195830-1195830 on Build GRCh38
rs755570501 Chr.16: 1195568-1195568 on Build GRCh38
rs758688396 Chr.16: 1195546-1195546 on Build GRCh38
rs751792904 Chr.16: 1195883-1195883 on Build GRCh38
rs374608011 Chr.16: 1195490-1195490 on Build GRCh38
rs375982567 Chr.16: 1195917-1195917 on Build GRCh38
rs547690343 Chr.16: 1195915-1195915 on Build GRCh38
rs991609784 Chr.16: 1195809-1195809 on Build GRCh38
rs1053290820 Chr.16: 1195814-1195814 on Build GRCh38
rs564747288 Chr.16: 1195853-1195853 on Build GRCh38
rs201635357 Chr.16: 1195600-1195600 on Build GRCh38
rs759634467 Chr.16: 1195536-1195536 on Build GRCh38
rs746339730 Chr.16: 1195920-1195920 on Build GRCh38
rs774987924 Chr.16: 1195512-1195512 on Build GRCh38
rs1011546545 Chr.16: 1195709-1195709 on Build GRCh38
rs1046288068 Chr.16: 1195655-1195655 on Build GRCh38
rs530140454 Chr.16: 1195918-1195918 on Build GRCh38
rs368785828 Chr.16: 1195557-1195557 on Build GRCh38
rs757136301 Chr.16: 1195500-1195500 on Build GRCh38
rs952582946 Chr.16: 1195763-1195763 on Build GRCh38
rs961604674 Chr.16: 1195766-1195766 on Build GRCh38
rs910918155 Chr.16: 1195598-1195598 on Build GRCh38
rs762972911 Chr.16: 1195611-1195611 on Build GRCh38
rs754882726 Chr.16: 1195721-1195721 on Build GRCh38
rs527682250 Chr.16: 1195658-1195658 on Build GRCh38
rs749032674 Chr.16: 1195513-1195513 on Build GRCh38
rs781049180 Chr.16: 1195563-1195563 on Build GRCh38
rs551248143 Chr.16: 1195593-1195593 on Build GRCh38
rs777692032 Chr.16: 1195491-1195491 on Build GRCh38
rs1029429259 Chr.16: 1195564-1195564 on Build GRCh38
rs771105384 Chr.16: 1195737-1195737 on Build GRCh38
rs937430505 Chr.16: 1195684-1195684 on Build GRCh38
rs1045818263 Chr.16: 1195483-1195483 on Build GRCh38
rs112895303 Chr.16: 1195836-1195836 on Build GRCh38
rs763776082 Chr.16: 1195548-1195548 on Build GRCh38
rs202153414 Chr.16: 1195879-1195879 on Build GRCh38
rs1053406289 Chr.16: 1195701-1195701 on Build GRCh38
rs904719565 Chr.16: 1195674-1195674 on Build GRCh38
rs779651648 Chr.16: 1195574-1195574 on Build GRCh38
rs573604767 Chr.16: 1195828-1195828 on Build GRCh38
rs562316147 Chr.16: 1195537-1195537 on Build GRCh38
rs775690682 Chr.16: 1195610-1195610 on Build GRCh38
rs768303354 Chr.16: 1195585-1195585 on Build GRCh38
rs374374399 Chr.16: 1195891-1195891 on Build GRCh38
rs112165476 Chr.16: 1195765-1195765 on Build GRCh38
rs866396554 Chr.16: 1195666-1195666 on Build GRCh38
rs906258550 Chr.16: 1195746-1195746 on Build GRCh38
rs932744635 Chr.16: 1195668-1195668 on Build GRCh38
rs985575194 Chr.16: 1195791-1195791 on Build GRCh38
rs374558503 Chr.16: 1195555-1195555 on Build GRCh38

View Full Product Details