Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1436358-1436852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC154
Entrez Gene ID
645811
Gene Name
coiled-coil domain containing 154
Gene Aliases
C16orf29
UniGene
Hs.355232
Gene Chromosome Location
Chr.16: 1434388-1444489 on Build GRCh38
Transcripts
NM_001143980.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4255969 c.1489A>G p.R497G Chr.16: 1436470-1436470 on Build GRCh38
6274564 c.1511C>A p.A504D Chr.16: 1436448-1436448 on Build GRCh38
5528224 c.1499C>T p.A500V Chr.16: 1436460-1436460 on Build GRCh38
6220641 c.1489A>C p.R497R Chr.16: 1436470-1436470 on Build GRCh38
329229 c.1498G>A p.A500T Chr.16: 1436461-1436461 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766828357 Chr.16: 1436731-1436731 on Build GRCh38
rs759709659 Chr.16: 1436647-1436648 on Build GRCh38
rs948190401 Chr.16: 1436571-1436571 on Build GRCh38
rs899008985 Chr.16: 1436664-1436664 on Build GRCh38
rs981368758 Chr.16: 1436558-1436558 on Build GRCh38
rs902881017 Chr.16: 1436427-1436427 on Build GRCh38
rs997156570 Chr.16: 1436437-1436437 on Build GRCh38
rs977954514 Chr.16: 1436813-1436813 on Build GRCh38
rs968149139 Chr.16: 1436720-1436720 on Build GRCh38
rs888779016 Chr.16: 1436636-1436636 on Build GRCh38
rs928730244 Chr.16: 1436560-1436560 on Build GRCh38
rs906423935 Chr.16: 1436725-1436725 on Build GRCh38
rs141015418 Chr.16: 1436536-1436536 on Build GRCh38
rs1038519011 Chr.16: 1436802-1436802 on Build GRCh38
rs978554617 Chr.16: 1436729-1436729 on Build GRCh38
rs533329414 Chr.16: 1436506-1436506 on Build GRCh38
rs769855361 Chr.16: 1436461-1436464 on Build GRCh38
rs948345742 Chr.16: 1436792-1436792 on Build GRCh38
rs886076421 Chr.16: 1436625-1436625 on Build GRCh38
rs760863704 Chr.16: 1436519-1436519 on Build GRCh38
rs947529521 Chr.16: 1436585-1436585 on Build GRCh38
rs112015572 Chr.16: 1436699-1436699 on Build GRCh38
rs1056621503 Chr.16: 1436541-1436541 on Build GRCh38
rs757251838 Chr.16: 1436615-1436615 on Build GRCh38
rs995020439 Chr.16: 1436605-1436605 on Build GRCh38
rs35583599 Chr.16: 1436461-1436461 on Build GRCh38
rs549594902 Chr.16: 1436669-1436669 on Build GRCh38
rs1018886446 Chr.16: 1436482-1436482 on Build GRCh38
rs1038977144 Chr.16: 1436591-1436591 on Build GRCh38
rs1007351130 Chr.16: 1436643-1436643 on Build GRCh38
rs1008585185 Chr.16: 1436647-1436647 on Build GRCh38
rs763994634 Chr.16: 1436535-1436535 on Build GRCh38
rs114304323 Chr.16: 1436394-1436394 on Build GRCh38
rs1037841343 Chr.16: 1436645-1436645 on Build GRCh38
rs1012390051 Chr.16: 1436412-1436412 on Build GRCh38
rs773221815 Chr.16: 1436514-1436517 on Build GRCh38
rs369531400 Chr.16: 1436803-1436803 on Build GRCh38
rs964516808 Chr.16: 1436502-1436502 on Build GRCh38
rs898917182 Chr.16: 1436449-1436449 on Build GRCh38
rs200290023 Chr.16: 1436507-1436507 on Build GRCh38
rs539385865 Chr.16: 1436829-1436829 on Build GRCh38
rs958680092 Chr.16: 1436758-1436758 on Build GRCh38
rs538546813 Chr.16: 1436739-1436739 on Build GRCh38
rs766420843 Chr.16: 1436419-1436419 on Build GRCh38
rs545124415 Chr.16: 1436441-1436441 on Build GRCh38
rs765564071 Chr.16: 1436753-1436753 on Build GRCh38
rs541611072 Chr.16: 1436400-1436400 on Build GRCh38
rs987412939 Chr.16: 1436462-1436462 on Build GRCh38
rs758266311 Chr.16: 1436772-1436772 on Build GRCh38
rs745314448 Chr.16: 1436545-1436545 on Build GRCh38
rs76876166 Chr.16: 1436597-1436597 on Build GRCh38
rs752544978 Chr.16: 1436501-1436501 on Build GRCh38
rs916823439 Chr.16: 1436773-1436773 on Build GRCh38
rs1026618911 Chr.16: 1436542-1436542 on Build GRCh38
rs991980350 Chr.16: 1436575-1436575 on Build GRCh38
rs769499035 Chr.16: 1436559-1436559 on Build GRCh38
rs188759974 Chr.16: 1436819-1436819 on Build GRCh38
rs73492006 Chr.16: 1436383-1436383 on Build GRCh38
rs1016134033 Chr.16: 1436510-1436510 on Build GRCh38
rs184119365 Chr.16: 1436504-1436504 on Build GRCh38
rs146803807 Chr.16: 1436399-1436399 on Build GRCh38
rs947701341 Chr.16: 1436418-1436418 on Build GRCh38
rs991617593 Chr.16: 1436776-1436776 on Build GRCh38
rs375262245 Chr.16: 1436686-1436686 on Build GRCh38
rs759469892 Chr.16: 1436745-1436745 on Build GRCh38
rs570758553 Chr.16: 1436505-1436505 on Build GRCh38
rs1051396816 Chr.16: 1436620-1436620 on Build GRCh38
rs978113950 Chr.16: 1436513-1436513 on Build GRCh38
rs574311061 Chr.16: 1436754-1436754 on Build GRCh38
rs974981740 Chr.16: 1436503-1436503 on Build GRCh38
rs6600145 Chr.16: 1436397-1436397 on Build GRCh38
rs914770937 Chr.16: 1436584-1436584 on Build GRCh38
rs923974721 Chr.16: 1436515-1436515 on Build GRCh38
rs1042789161 Chr.16: 1436384-1436384 on Build GRCh38
rs774936760 Chr.16: 1436563-1436563 on Build GRCh38
rs561677923 Chr.16: 1436458-1436458 on Build GRCh38
rs964628949 Chr.16: 1436667-1436667 on Build GRCh38
rs754095869 Chr.16: 1436456-1436456 on Build GRCh38
rs748613307 Chr.16: 1436666-1436666 on Build GRCh38

View Full Product Details