Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1614631-1615132 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRAMP1
Entrez Gene ID
57585
Gene Name
cramped chromatin regulator homolog 1
Gene Aliases
CRAMP1L, HN1L, TCE4
UniGene
Hs.603676
Gene Chromosome Location
Chr.16: 1614640-1677908 on Build GRCh38
Transcripts
NM_020825.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6398864 c.314G>A p.G105D Chr.16: 1614953-1614953 on Build GRCh38
5684574 c.208T>C p.S70P Chr.16: 1614847-1614847 on Build GRCh38
3754712 c.291T>C p.D97D Chr.16: 1614930-1614930 on Build GRCh38
3749336 c.203C>T p.A68V Chr.16: 1614842-1614842 on Build GRCh38
6532187 c.346G>C p.G116R Chr.16: 1614985-1614985 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1011131956 Chr.16: 1614845-1614845 on Build GRCh38
rs879615489 Chr.16: 1615020-1615020 on Build GRCh38
rs923089487 Chr.16: 1614996-1614996 on Build GRCh38
rs553852632 Chr.16: 1614798-1614798 on Build GRCh38
rs963285123 Chr.16: 1615051-1615051 on Build GRCh38
rs375219531 Chr.16: 1615093-1615093 on Build GRCh38
rs959779059 Chr.16: 1614944-1614944 on Build GRCh38
rs994523558 Chr.16: 1614791-1614813 on Build GRCh38
rs778126645 Chr.16: 1614791-1614791 on Build GRCh38
rs1001291682 Chr.16: 1614865-1614865 on Build GRCh38
rs1024445444 Chr.16: 1614846-1614846 on Build GRCh38
rs919127061 Chr.16: 1615070-1615070 on Build GRCh38
rs1034098447 Chr.16: 1614895-1614895 on Build GRCh38
rs774605333 Chr.16: 1614951-1614951 on Build GRCh38
rs1034044494 Chr.16: 1615108-1615108 on Build GRCh38
rs776510604 Chr.16: 1614817-1614817 on Build GRCh38
rs112844895 Chr.16: 1614830-1614830 on Build GRCh38
rs976412658 Chr.16: 1614972-1614972 on Build GRCh38
rs754153222 Chr.16: 1615012-1615012 on Build GRCh38
rs769727202 Chr.16: 1614885-1614885 on Build GRCh38
rs971976818 Chr.16: 1614848-1614848 on Build GRCh38
rs2301458 Chr.16: 1615109-1615109 on Build GRCh38
rs957023853 Chr.16: 1614920-1614920 on Build GRCh38
rs531070701 Chr.16: 1615094-1615094 on Build GRCh38
rs941322877 Chr.16: 1615027-1615027 on Build GRCh38
rs189020189 Chr.16: 1614842-1614842 on Build GRCh38
rs868231334 Chr.16: 1614745-1614745 on Build GRCh38
rs926923881 Chr.16: 1614695-1614695 on Build GRCh38
rs1049831809 Chr.16: 1615018-1615018 on Build GRCh38
rs181418270 Chr.16: 1614933-1614933 on Build GRCh38
rs778881967 Chr.16: 1614720-1614720 on Build GRCh38
rs990019394 Chr.16: 1614777-1614777 on Build GRCh38
rs576208802 Chr.16: 1615017-1615017 on Build GRCh38
rs991122903 Chr.16: 1614998-1614998 on Build GRCh38
rs770853672 Chr.16: 1614806-1614806 on Build GRCh38
rs1049843132 Chr.16: 1614820-1614820 on Build GRCh38
rs980268947 Chr.16: 1614678-1614678 on Build GRCh38
rs1017576837 Chr.16: 1614831-1614831 on Build GRCh38
rs1005589431 Chr.16: 1614825-1614825 on Build GRCh38
rs1024453985 Chr.16: 1614663-1614663 on Build GRCh38
rs994393222 Chr.16: 1615054-1615054 on Build GRCh38
rs915549382 Chr.16: 1615019-1615019 on Build GRCh38
rs766748223 Chr.16: 1614941-1614941 on Build GRCh38
rs761391734 Chr.16: 1614938-1614938 on Build GRCh38
rs867649752 Chr.16: 1614959-1614959 on Build GRCh38
rs774899859 Chr.16: 1614910-1614910 on Build GRCh38
rs897159014 Chr.16: 1615039-1615039 on Build GRCh38
rs929126024 Chr.16: 1615074-1615074 on Build GRCh38
rs1026967852 Chr.16: 1614819-1614830 on Build GRCh38
rs1007968040 Chr.16: 1614824-1614824 on Build GRCh38
rs1038353991 Chr.16: 1615033-1615033 on Build GRCh38
rs763376463 Chr.16: 1614924-1614924 on Build GRCh38
rs527826310 Chr.16: 1615110-1615110 on Build GRCh38
rs758590705 Chr.16: 1614780-1614780 on Build GRCh38
rs762499648 Chr.16: 1614923-1614923 on Build GRCh38
rs7201813 Chr.16: 1614930-1614930 on Build GRCh38
rs971145177 Chr.16: 1614673-1614673 on Build GRCh38
rs1046444959 Chr.16: 1615061-1615061 on Build GRCh38
rs1040184115 Chr.16: 1614813-1614813 on Build GRCh38
rs748220918 Chr.16: 1614738-1614738 on Build GRCh38
rs951018716 Chr.16: 1614833-1614850 on Build GRCh38
rs755456226 Chr.16: 1614691-1614691 on Build GRCh38
rs1051893277 Chr.16: 1615082-1615082 on Build GRCh38
rs957076005 Chr.16: 1614748-1614748 on Build GRCh38
rs973277473 Chr.16: 1615064-1615064 on Build GRCh38
rs754351402 Chr.16: 1614684-1614684 on Build GRCh38

View Full Product Details