Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1772720-1773222 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EME2
Entrez Gene ID
197342
Gene Name
essential meiotic structure-specific endonuclease subunit 2
Gene Aliases
SLX2B, gs125
UniGene
Hs.7247
Gene Chromosome Location
Chr.16: 1773222-1776714 on Build GRCh38
Transcripts
XM_017023025.1 XM_017023026.1 NM_001300900.1 NM_023936.1 XM_017023595.1

Gene Symbol
MRPS34
Entrez Gene ID
65993
Gene Name
mitochondrial ribosomal protein S34
Gene Aliases
MRP-S12, MRP-S34, MRPS12
UniGene
Hs.720388
Gene Chromosome Location
Chr.16: 1771889-1773151 on Build GRCh38
Transcripts
XM_017023025.1 XM_017023026.1 NM_001300900.1 NM_023936.1 XM_017023595.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7276356 c.241A>G p.T81A Chr.16: 1772879-1772879 on Build GRCh38
4958187 c.189C>A p.R63R Chr.16: 1772931-1772931 on Build GRCh38
5499239 c.230G>A p.G77D Chr.16: 1772890-1772890 on Build GRCh38
5732310 c.22C>T p.P8S Chr.16: 1773098-1773098 on Build GRCh38
5433483 c.78C>T p.N26N Chr.16: 1773042-1773042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038067053 Chr.16: 1773013-1773013 on Build GRCh38
rs1025882021 Chr.16: 1772783-1772783 on Build GRCh38
rs888638893 Chr.16: 1773168-1773168 on Build GRCh38
rs759517804 Chr.16: 1773040-1773040 on Build GRCh38
rs915544717 Chr.16: 1773089-1773089 on Build GRCh38
rs763672163 Chr.16: 1773026-1773026 on Build GRCh38
rs1021652235 Chr.16: 1772976-1772976 on Build GRCh38
rs867256648 Chr.16: 1772795-1772796 on Build GRCh38
rs892560987 Chr.16: 1773114-1773114 on Build GRCh38
rs11552432 Chr.16: 1773053-1773053 on Build GRCh38
rs548748973 Chr.16: 1772923-1772923 on Build GRCh38
rs994323145 Chr.16: 1772905-1772905 on Build GRCh38
rs929641538 Chr.16: 1773124-1773124 on Build GRCh38
rs538094386 Chr.16: 1772999-1772999 on Build GRCh38
rs765217192 Chr.16: 1772915-1772915 on Build GRCh38
rs758711405 Chr.16: 1773105-1773105 on Build GRCh38
rs778248787 Chr.16: 1773109-1773109 on Build GRCh38
rs33993627 Chr.16: 1772797-1772798 on Build GRCh38
rs975567721 Chr.16: 1772884-1772884 on Build GRCh38
rs912341085 Chr.16: 1773159-1773159 on Build GRCh38
rs755456304 Chr.16: 1773092-1773092 on Build GRCh38
rs763000565 Chr.16: 1773156-1773156 on Build GRCh38
rs749995544 Chr.16: 1773074-1773074 on Build GRCh38
rs1012370099 Chr.16: 1772746-1772746 on Build GRCh38
rs898270739 Chr.16: 1772902-1772902 on Build GRCh38
rs915831791 Chr.16: 1772990-1772990 on Build GRCh38
rs977924736 Chr.16: 1772827-1772827 on Build GRCh38
rs762655490 Chr.16: 1773046-1773046 on Build GRCh38
rs891304369 Chr.16: 1772945-1772945 on Build GRCh38
rs780648399 Chr.16: 1773058-1773058 on Build GRCh38
rs749593629 Chr.16: 1772751-1772751 on Build GRCh38
rs752478974 Chr.16: 1772840-1772840 on Build GRCh38
rs914681096 Chr.16: 1772813-1772813 on Build GRCh38
rs11552431 Chr.16: 1773023-1773023 on Build GRCh38
rs199579121 Chr.16: 1772801-1772801 on Build GRCh38
rs528871514 Chr.16: 1772885-1772885 on Build GRCh38
rs969365847 Chr.16: 1773115-1773115 on Build GRCh38
rs767171526 Chr.16: 1773194-1773194 on Build GRCh38
rs531329366 Chr.16: 1772786-1772786 on Build GRCh38
rs916517837 Chr.16: 1773003-1773003 on Build GRCh38
rs1000988298 Chr.16: 1773005-1773005 on Build GRCh38
rs201469362 Chr.16: 1772796-1772796 on Build GRCh38
rs11552433 Chr.16: 1772811-1772811 on Build GRCh38
rs964306396 Chr.16: 1772869-1772869 on Build GRCh38
rs771344174 Chr.16: 1773111-1773111 on Build GRCh38
rs773310882 Chr.16: 1773018-1773018 on Build GRCh38
rs947691777 Chr.16: 1773006-1773006 on Build GRCh38
rs977891012 Chr.16: 1772989-1772989 on Build GRCh38
rs892494470 Chr.16: 1773011-1773011 on Build GRCh38
rs34595082 Chr.16: 1772800-1772801 on Build GRCh38
rs748505165 Chr.16: 1773104-1773104 on Build GRCh38
rs759607447 Chr.16: 1772830-1772830 on Build GRCh38
rs952467574 Chr.16: 1772944-1772944 on Build GRCh38
rs894769666 Chr.16: 1772849-1772849 on Build GRCh38
rs908269410 Chr.16: 1773072-1773072 on Build GRCh38
rs1052918975 Chr.16: 1773113-1773113 on Build GRCh38
rs960799129 Chr.16: 1773099-1773099 on Build GRCh38
rs1035747244 Chr.16: 1773098-1773098 on Build GRCh38
rs554225447 Chr.16: 1773122-1773122 on Build GRCh38
rs765079347 Chr.16: 1772833-1772833 on Build GRCh38
rs534007365 Chr.16: 1773094-1773094 on Build GRCh38
rs749869159 Chr.16: 1773199-1773199 on Build GRCh38
rs1056386574 Chr.16: 1772844-1772844 on Build GRCh38
rs1046823772 Chr.16: 1773133-1773133 on Build GRCh38
rs775315534 Chr.16: 1773126-1773126 on Build GRCh38
rs773293309 Chr.16: 1772794-1772794 on Build GRCh38
rs964367525 Chr.16: 1772972-1772972 on Build GRCh38
rs760869237 Chr.16: 1772795-1772795 on Build GRCh38
rs1027180981 Chr.16: 1773163-1773163 on Build GRCh38
rs758051458 Chr.16: 1772864-1772864 on Build GRCh38
rs923851255 Chr.16: 1772834-1772834 on Build GRCh38
rs919673500 Chr.16: 1773015-1773015 on Build GRCh38
rs986798358 Chr.16: 1772791-1772791 on Build GRCh38
rs944193192 Chr.16: 1773161-1773161 on Build GRCh38
rs990190809 Chr.16: 1772974-1772974 on Build GRCh38
rs1038503314 Chr.16: 1772865-1772865 on Build GRCh38
rs769701527 Chr.16: 1773123-1773123 on Build GRCh38
rs1018543927 Chr.16: 1772956-1772956 on Build GRCh38
rs560022978 Chr.16: 1772793-1772793 on Build GRCh38
rs2006150 Chr.16: 1772770-1772770 on Build GRCh38
rs776919892 Chr.16: 1773037-1773037 on Build GRCh38
rs4027362 Chr.16: 1772796-1772797 on Build GRCh38
rs527902565 Chr.16: 1772760-1772760 on Build GRCh38
rs780734714 Chr.16: 1772948-1772948 on Build GRCh38
rs967352546 Chr.16: 1773093-1773093 on Build GRCh38
rs761444109 Chr.16: 1773178-1773178 on Build GRCh38
rs891271151 Chr.16: 1773028-1773028 on Build GRCh38
rs753841145 Chr.16: 1772825-1772825 on Build GRCh38
rs749789584 Chr.16: 1772967-1772967 on Build GRCh38
rs923737009 Chr.16: 1772995-1772995 on Build GRCh38
rs866553498 Chr.16: 1772766-1772766 on Build GRCh38
rs1057072435 Chr.16: 1772982-1772982 on Build GRCh38
rs774644160 Chr.16: 1772777-1772777 on Build GRCh38
rs752492016 Chr.16: 1773043-1773043 on Build GRCh38
rs776835146 Chr.16: 1773112-1773112 on Build GRCh38
rs1025859109 Chr.16: 1772924-1772924 on Build GRCh38
rs779312899 Chr.16: 1772997-1772997 on Build GRCh38
rs867780521 Chr.16: 1772835-1772835 on Build GRCh38
rs944104381 Chr.16: 1773117-1773117 on Build GRCh38
rs1076695 Chr.16: 1772946-1772946 on Build GRCh38
rs772298168 Chr.16: 1773000-1773000 on Build GRCh38
rs1004356275 Chr.16: 1773174-1773174 on Build GRCh38
rs756968636 Chr.16: 1773056-1773056 on Build GRCh38
rs567695582 Chr.16: 1773202-1773202 on Build GRCh38
rs947756957 Chr.16: 1772860-1772860 on Build GRCh38
rs967468654 Chr.16: 1772983-1772983 on Build GRCh38
rs763769567 Chr.16: 1773169-1773169 on Build GRCh38
rs898633725 Chr.16: 1772765-1772765 on Build GRCh38

View Full Product Details