Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1773602-1774099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EME2
Entrez Gene ID
197342
Gene Name
essential meiotic structure-specific endonuclease subunit 2
Gene Aliases
SLX2B, gs125
UniGene
Hs.7247
Gene Chromosome Location
Chr.16: 1773222-1776714 on Build GRCh38
Transcripts
NM_001257370.1 XM_017023025.1 XM_017023026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6299653 c.314G>A p.R105H Chr.16: 1773771-1773771 on Build GRCh38
6397129 c.344T>A p.L115Q Chr.16: 1773801-1773801 on Build GRCh38
7366182 c.304T>C p.C102R Chr.16: 1773761-1773761 on Build GRCh38
4852963 c.319G>C p.E107Q Chr.16: 1773776-1773776 on Build GRCh38
5740515 c.286G>A p.A96T Chr.16: 1773743-1773743 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755764387 Chr.16: 1773694-1773694 on Build GRCh38
rs920562527 Chr.16: 1773909-1773909 on Build GRCh38
rs767363303 Chr.16: 1773763-1773763 on Build GRCh38
rs763128723 Chr.16: 1773734-1773734 on Build GRCh38
rs1040519625 Chr.16: 1773950-1773950 on Build GRCh38
rs776920899 Chr.16: 1773720-1773720 on Build GRCh38
rs186546615 Chr.16: 1773650-1773650 on Build GRCh38
rs774425457 Chr.16: 1773873-1773873 on Build GRCh38
rs778471994 Chr.16: 1773810-1773810 on Build GRCh38
rs904174627 Chr.16: 1774000-1774000 on Build GRCh38
rs777251842 Chr.16: 1773964-1773964 on Build GRCh38
rs920676787 Chr.16: 1773752-1773752 on Build GRCh38
rs558383127 Chr.16: 1773682-1773682 on Build GRCh38
rs1028820864 Chr.16: 1773662-1773662 on Build GRCh38
rs547679861 Chr.16: 1773839-1773839 on Build GRCh38
rs886910511 Chr.16: 1773789-1773789 on Build GRCh38
rs76650990 Chr.16: 1773692-1773692 on Build GRCh38
rs552104285 Chr.16: 1774013-1774013 on Build GRCh38
rs886944827 Chr.16: 1773627-1773627 on Build GRCh38
rs751652697 Chr.16: 1773746-1773746 on Build GRCh38
rs797017582 Chr.16: 1773935-1773935 on Build GRCh38
rs199761703 Chr.16: 1773727-1773727 on Build GRCh38
rs574916642 Chr.16: 1774068-1774068 on Build GRCh38
rs192331602 Chr.16: 1773921-1773921 on Build GRCh38
rs996937332 Chr.16: 1773688-1773688 on Build GRCh38
rs1024795198 Chr.16: 1774023-1774023 on Build GRCh38
rs1024857410 Chr.16: 1774052-1774052 on Build GRCh38
rs957610618 Chr.16: 1774062-1774062 on Build GRCh38
rs1031403402 Chr.16: 1773853-1773853 on Build GRCh38
rs1027925351 Chr.16: 1774072-1774072 on Build GRCh38
rs999843405 Chr.16: 1774006-1774006 on Build GRCh38
rs901086909 Chr.16: 1773980-1773980 on Build GRCh38
rs770118349 Chr.16: 1773836-1773836 on Build GRCh38
rs573283501 Chr.16: 1773827-1773827 on Build GRCh38
rs1040947078 Chr.16: 1773800-1773800 on Build GRCh38
rs567315180 Chr.16: 1773864-1773864 on Build GRCh38
rs878929511 Chr.16: 1773663-1773663 on Build GRCh38
rs980734819 Chr.16: 1774070-1774070 on Build GRCh38
rs761833968 Chr.16: 1773874-1773874 on Build GRCh38
rs945752344 Chr.16: 1773758-1773758 on Build GRCh38
rs971793389 Chr.16: 1773925-1773925 on Build GRCh38
rs557282646 Chr.16: 1773737-1773738 on Build GRCh38
rs1031369405 Chr.16: 1774012-1774012 on Build GRCh38
rs990325272 Chr.16: 1773937-1773937 on Build GRCh38
rs538643966 Chr.16: 1773987-1773987 on Build GRCh38
rs948735777 Chr.16: 1773733-1773733 on Build GRCh38
rs930642630 Chr.16: 1773913-1773913 on Build GRCh38
rs763227586 Chr.16: 1773845-1773845 on Build GRCh38
rs746179767 Chr.16: 1773832-1773832 on Build GRCh38
rs765401210 Chr.16: 1773723-1773723 on Build GRCh38
rs572452172 Chr.16: 1773645-1773645 on Build GRCh38
rs150946324 Chr.16: 1774015-1774015 on Build GRCh38
rs569854840 Chr.16: 1773975-1773975 on Build GRCh38
rs374315507 Chr.16: 1773766-1773766 on Build GRCh38
rs754653019 Chr.16: 1773794-1773794 on Build GRCh38
rs956189417 Chr.16: 1773855-1773855 on Build GRCh38
rs531318985 Chr.16: 1773702-1773702 on Build GRCh38
rs996456546 Chr.16: 1773659-1773659 on Build GRCh38
rs1033476 Chr.16: 1774074-1774074 on Build GRCh38
rs201737681 Chr.16: 1773739-1773739 on Build GRCh38
rs1026860895 Chr.16: 1773626-1773626 on Build GRCh38
rs1028445254 Chr.16: 1773823-1773823 on Build GRCh38
rs970261231 Chr.16: 1774057-1774057 on Build GRCh38
rs1014800723 Chr.16: 1773992-1773992 on Build GRCh38
rs750205139 Chr.16: 1773777-1773777 on Build GRCh38
rs747546404 Chr.16: 1773715-1773715 on Build GRCh38
rs945854436 Chr.16: 1773940-1773940 on Build GRCh38
rs753436339 Chr.16: 1773990-1773990 on Build GRCh38
rs1016955096 Chr.16: 1773670-1773670 on Build GRCh38
rs936210710 Chr.16: 1773936-1773936 on Build GRCh38
rs779884399 Chr.16: 1773698-1773698 on Build GRCh38
rs913350647 Chr.16: 1773938-1773938 on Build GRCh38
rs529926617 Chr.16: 1773897-1773897 on Build GRCh38
rs965996635 Chr.16: 1773658-1773658 on Build GRCh38
rs759897366 Chr.16: 1773722-1773722 on Build GRCh38
rs917501982 Chr.16: 1773881-1773881 on Build GRCh38
rs1040041297 Chr.16: 1773771-1773771 on Build GRCh38
rs555262939 Chr.16: 1774067-1774067 on Build GRCh38
rs568538388 Chr.16: 1774027-1774028 on Build GRCh38
rs970757175 Chr.16: 1773893-1773893 on Build GRCh38
rs183713932 Chr.16: 1774039-1774039 on Build GRCh38
rs761755726 Chr.16: 1773762-1773762 on Build GRCh38
rs780043716 Chr.16: 1773786-1773786 on Build GRCh38
rs983478800 Chr.16: 1774077-1774077 on Build GRCh38
rs951872776 Chr.16: 1773664-1773664 on Build GRCh38
rs766809953 Chr.16: 1773622-1773623 on Build GRCh38
rs997126589 Chr.16: 1773983-1773983 on Build GRCh38
rs931628931 Chr.16: 1773952-1773952 on Build GRCh38
rs999981929 Chr.16: 1773631-1773631 on Build GRCh38
rs887706885 Chr.16: 1773969-1773969 on Build GRCh38
rs773774708 Chr.16: 1773714-1773714 on Build GRCh38
rs901114865 Chr.16: 1773816-1773816 on Build GRCh38
rs1049639012 Chr.16: 1773993-1773993 on Build GRCh38
rs980373995 Chr.16: 1773932-1773932 on Build GRCh38
rs574989090 Chr.16: 1773657-1773657 on Build GRCh38
rs768190872 Chr.16: 1773708-1773708 on Build GRCh38
rs961733474 Chr.16: 1773684-1773684 on Build GRCh38
rs755542062 Chr.16: 1773818-1773818 on Build GRCh38
rs907979650 Chr.16: 1773926-1773926 on Build GRCh38
rs965048708 Chr.16: 1773651-1773651 on Build GRCh38
rs924742128 Chr.16: 1773711-1773711 on Build GRCh38
rs781684913 Chr.16: 1773828-1773828 on Build GRCh38
rs771398630 Chr.16: 1773716-1773716 on Build GRCh38
rs901538131 Chr.16: 1773951-1773951 on Build GRCh38
rs755997861 Chr.16: 1773785-1773785 on Build GRCh38
rs984196932 Chr.16: 1773666-1773666 on Build GRCh38
rs756465784 Chr.16: 1773738-1773746 on Build GRCh38
rs34551631 Chr.16: 1773892-1773893 on Build GRCh38
rs996083815 Chr.16: 1773819-1773819 on Build GRCh38
rs993072488 Chr.16: 1773721-1773721 on Build GRCh38
rs917201339 Chr.16: 1773693-1773693 on Build GRCh38
rs753510592 Chr.16: 1773790-1773790 on Build GRCh38
rs558812921 Chr.16: 1773635-1773635 on Build GRCh38
rs539783783 Chr.16: 1773646-1773646 on Build GRCh38
rs767515317 Chr.16: 1773888-1773888 on Build GRCh38

View Full Product Details