Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 1984640-1985139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFER
Entrez Gene ID
2671
Gene Name
growth factor, augmenter of liver regeneration
Gene Aliases
ALR, ERV1, HERV1, HPO, HPO1, HPO2, HSS
UniGene
Hs.27184
Gene Chromosome Location
Chr.16: 1984149-1987749 on Build GRCh38
Transcripts
NM_005262.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6895365 c.427G>C p.E143Q Chr.16: 1984915-1984915 on Build GRCh38
1242309 c.278A>G p.E93G Chr.16: 1984766-1984766 on Build GRCh38
7155905 c.298G>A p.E100K Chr.16: 1984786-1984786 on Build GRCh38
6555346 c.280G>T p.D94Y Chr.16: 1984768-1984768 on Build GRCh38
1678758 c.410C>A p.S137Y Chr.16: 1984898-1984898 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991504262 Chr.16: 1985043-1985043 on Build GRCh38
rs545266207 Chr.16: 1984683-1984683 on Build GRCh38
rs929832047 Chr.16: 1985068-1985068 on Build GRCh38
rs376478434 Chr.16: 1984995-1984995 on Build GRCh38
rs750605745 Chr.16: 1984988-1984988 on Build GRCh38
rs781395247 Chr.16: 1984837-1984837 on Build GRCh38
rs962089141 Chr.16: 1984803-1984803 on Build GRCh38
rs863224026 Chr.16: 1984942-1984942 on Build GRCh38
rs756436660 Chr.16: 1984991-1984991 on Build GRCh38
rs760336801 Chr.16: 1984964-1984964 on Build GRCh38
rs771378387 Chr.16: 1985089-1985089 on Build GRCh38
rs770384334 Chr.16: 1984970-1984970 on Build GRCh38
rs938705432 Chr.16: 1984725-1984725 on Build GRCh38
rs745746499 Chr.16: 1984738-1984738 on Build GRCh38
rs770846566 Chr.16: 1984719-1984719 on Build GRCh38
rs373446345 Chr.16: 1984910-1984910 on Build GRCh38
rs758618799 Chr.16: 1984696-1984696 on Build GRCh38
rs1039357489 Chr.16: 1985109-1985109 on Build GRCh38
rs201603991 Chr.16: 1984840-1984840 on Build GRCh38
rs532850984 Chr.16: 1984691-1984691 on Build GRCh38
rs747266786 Chr.16: 1984716-1984716 on Build GRCh38
rs774235374 Chr.16: 1984856-1984856 on Build GRCh38
rs928741231 Chr.16: 1985015-1985015 on Build GRCh38
rs765722914 Chr.16: 1984881-1984881 on Build GRCh38
rs763786791 Chr.16: 1984770-1984770 on Build GRCh38
rs756203483 Chr.16: 1984925-1984925 on Build GRCh38
rs534308773 Chr.16: 1985056-1985056 on Build GRCh38
rs538091471 Chr.16: 1984976-1984976 on Build GRCh38
rs569048447 Chr.16: 1984952-1984952 on Build GRCh38
rs980288156 Chr.16: 1984876-1984876 on Build GRCh38
rs768661163 Chr.16: 1984951-1984951 on Build GRCh38
rs895073717 Chr.16: 1984771-1984771 on Build GRCh38
rs753132528 Chr.16: 1984884-1984884 on Build GRCh38
rs771712671 Chr.16: 1984960-1984960 on Build GRCh38
rs528813454 Chr.16: 1984887-1984887 on Build GRCh38
rs766898922 Chr.16: 1984792-1984792 on Build GRCh38
rs2075812 Chr.16: 1984660-1984660 on Build GRCh38
rs931891068 Chr.16: 1985022-1985022 on Build GRCh38
rs778160972 Chr.16: 1984961-1984961 on Build GRCh38
rs916469728 Chr.16: 1985045-1985045 on Build GRCh38
rs1038714161 Chr.16: 1985058-1985058 on Build GRCh38
rs564232412 Chr.16: 1984685-1984685 on Build GRCh38
rs946166808 Chr.16: 1984686-1984686 on Build GRCh38
rs763545328 Chr.16: 1984909-1984909 on Build GRCh38
rs549270205 Chr.16: 1984949-1984949 on Build GRCh38
rs776799405 Chr.16: 1984731-1984731 on Build GRCh38
rs749260043 Chr.16: 1984848-1984848 on Build GRCh38
rs1056390126 Chr.16: 1984727-1984727 on Build GRCh38
rs750515991 Chr.16: 1984917-1984917 on Build GRCh38
rs898349806 Chr.16: 1984836-1984836 on Build GRCh38
rs1046764757 Chr.16: 1985069-1985069 on Build GRCh38
rs369886365 Chr.16: 1984933-1984933 on Build GRCh38
rs761533080 Chr.16: 1984780-1984780 on Build GRCh38
rs899526838 Chr.16: 1985115-1985115 on Build GRCh38
rs760018110 Chr.16: 1984799-1984799 on Build GRCh38
rs780196676 Chr.16: 1984843-1984843 on Build GRCh38
rs757388602 Chr.16: 1984911-1984911 on Build GRCh38
rs757519057 Chr.16: 1984833-1984833 on Build GRCh38
rs370700293 Chr.16: 1984850-1984850 on Build GRCh38
rs571384984 Chr.16: 1985046-1985046 on Build GRCh38
rs753058591 Chr.16: 1984800-1984800 on Build GRCh38
rs898325421 Chr.16: 1985065-1985065 on Build GRCh38
rs772682634 Chr.16: 1984868-1984868 on Build GRCh38
rs760247120 Chr.16: 1984877-1984877 on Build GRCh38
rs947853655 Chr.16: 1984817-1984817 on Build GRCh38
rs768270980 Chr.16: 1984849-1984849 on Build GRCh38
rs778201726 Chr.16: 1984822-1984822 on Build GRCh38
rs763341760 Chr.16: 1984978-1984978 on Build GRCh38
rs863224024 Chr.16: 1984758-1984758 on Build GRCh38
rs781628694 Chr.16: 1984913-1984913 on Build GRCh38
rs771809901 Chr.16: 1984861-1984861 on Build GRCh38
rs762295209 Chr.16: 1984986-1984986 on Build GRCh38
rs764697486 Chr.16: 1984980-1984980 on Build GRCh38
rs769900445 Chr.16: 1984749-1984749 on Build GRCh38
rs748018775 Chr.16: 1984955-1984955 on Build GRCh38
rs768871809 Chr.16: 1984702-1984702 on Build GRCh38
rs762907502 Chr.16: 1984765-1984765 on Build GRCh38
rs923068897 Chr.16: 1985015-1985016 on Build GRCh38
rs3177436 Chr.16: 1984945-1984945 on Build GRCh38
rs745983562 Chr.16: 1984839-1984839 on Build GRCh38
rs1007826566 Chr.16: 1985106-1985106 on Build GRCh38
rs368889722 Chr.16: 1984823-1984823 on Build GRCh38
rs887684795 Chr.16: 1985091-1985091 on Build GRCh38
rs777926923 Chr.16: 1984708-1984708 on Build GRCh38
rs758750318 Chr.16: 1984818-1984818 on Build GRCh38
rs754277823 Chr.16: 1984668-1984668 on Build GRCh38
rs758983119 Chr.16: 1984776-1984776 on Build GRCh38
rs199651600 Chr.16: 1984757-1984757 on Build GRCh38
rs747357298 Chr.16: 1985086-1985086 on Build GRCh38
rs1006300352 Chr.16: 1984786-1984786 on Build GRCh38
rs780287282 Chr.16: 1984946-1984946 on Build GRCh38
rs938632533 Chr.16: 1985016-1985016 on Build GRCh38
rs749958872 Chr.16: 1984798-1984798 on Build GRCh38

View Full Product Details