Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2211793-2212302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRICD5
Entrez Gene ID
283870
Gene Name
BRICHOS domain containing 5
Gene Aliases
C16orf79
UniGene
Hs.647399
Gene Chromosome Location
Chr.16: 2209253-2213037 on Build GRCh38
Transcripts
XM_005255263.4 NM_001042371.2

Gene Symbol
PGP
Entrez Gene ID
283871
Gene Name
phosphoglycolate phosphatase
Gene Aliases
AUM, G3PP, PGPase
UniGene
Hs.442634
Gene Chromosome Location
Chr.16: 2211602-2214827 on Build GRCh38
Transcripts
XM_005255263.4 NM_001042371.2

SNP Detail

SNP ID Chromosome Location
rs76732812 Chr.16: 2211882-2211882 on Build GRCh38
rs772638517 Chr.16: 2212053-2212053 on Build GRCh38
rs112972541 Chr.16: 2211937-2211937 on Build GRCh38
rs768228372 Chr.16: 2211973-2211973 on Build GRCh38
rs115079696 Chr.16: 2211822-2211822 on Build GRCh38
rs11403 Chr.16: 2212065-2212065 on Build GRCh38
rs902794938 Chr.16: 2212195-2212195 on Build GRCh38
rs144974338 Chr.16: 2211927-2211927 on Build GRCh38
rs990810468 Chr.16: 2211936-2211936 on Build GRCh38
rs1010613410 Chr.16: 2212165-2212165 on Build GRCh38
rs887320407 Chr.16: 2211907-2211907 on Build GRCh38
rs777833180 Chr.16: 2212062-2212062 on Build GRCh38
rs1126 Chr.16: 2212112-2212112 on Build GRCh38
rs919207557 Chr.16: 2212098-2212098 on Build GRCh38
rs1052087548 Chr.16: 2212140-2212140 on Build GRCh38
rs528334328 Chr.16: 2212089-2212089 on Build GRCh38
rs558229688 Chr.16: 2212268-2212268 on Build GRCh38
rs549628055 Chr.16: 2212230-2212230 on Build GRCh38
rs149895819 Chr.16: 2211862-2211862 on Build GRCh38
rs160546 Chr.16: 2212092-2212092 on Build GRCh38
rs368222893 Chr.16: 2211849-2211849 on Build GRCh38
rs924490209 Chr.16: 2211835-2211835 on Build GRCh38
rs895320245 Chr.16: 2212250-2212250 on Build GRCh38
rs962694558 Chr.16: 2212079-2212079 on Build GRCh38
rs1031697391 Chr.16: 2211974-2211974 on Build GRCh38
rs1042039318 Chr.16: 2212176-2212176 on Build GRCh38
rs780265386 Chr.16: 2212147-2212147 on Build GRCh38
rs544642886 Chr.16: 2212027-2212027 on Build GRCh38
rs563831594 Chr.16: 2212125-2212125 on Build GRCh38
rs536153053 Chr.16: 2212004-2212004 on Build GRCh38
rs111273330 Chr.16: 2212271-2212271 on Build GRCh38
rs571490950 Chr.16: 2212026-2212026 on Build GRCh38
rs890473735 Chr.16: 2212114-2212119 on Build GRCh38
rs761417962 Chr.16: 2212049-2212049 on Build GRCh38
rs959468415 Chr.16: 2211931-2211931 on Build GRCh38
rs546835026 Chr.16: 2212164-2212164 on Build GRCh38
rs928079957 Chr.16: 2211930-2211930 on Build GRCh38
rs373165504 Chr.16: 2212187-2212187 on Build GRCh38
rs941342335 Chr.16: 2211946-2211946 on Build GRCh38
rs910387507 Chr.16: 2211854-2211854 on Build GRCh38
rs1043220920 Chr.16: 2212268-2212269 on Build GRCh38
rs561321204 Chr.16: 2212110-2212110 on Build GRCh38
rs949019833 Chr.16: 2212262-2212262 on Build GRCh38
rs1004145341 Chr.16: 2212061-2212061 on Build GRCh38
rs1003813865 Chr.16: 2212202-2212202 on Build GRCh38
rs929059281 Chr.16: 2212056-2212056 on Build GRCh38
rs948867864 Chr.16: 2212090-2212090 on Build GRCh38
rs141582521 Chr.16: 2212245-2212245 on Build GRCh38
rs567066501 Chr.16: 2212190-2212190 on Build GRCh38
rs779553644 Chr.16: 2211964-2211964 on Build GRCh38
rs929165458 Chr.16: 2211958-2211958 on Build GRCh38
rs773945914 Chr.16: 2212046-2212046 on Build GRCh38
rs990884102 Chr.16: 2212071-2212071 on Build GRCh38
rs939050769 Chr.16: 2212256-2212256 on Build GRCh38
rs1027463293 Chr.16: 2212192-2212192 on Build GRCh38
rs769689410 Chr.16: 2211947-2211947 on Build GRCh38
rs1047394072 Chr.16: 2211899-2211899 on Build GRCh38
rs893550701 Chr.16: 2212162-2212162 on Build GRCh38
rs114996223 Chr.16: 2212070-2212070 on Build GRCh38
rs376725307 Chr.16: 2211956-2211956 on Build GRCh38
rs558266565 Chr.16: 2212028-2212028 on Build GRCh38
rs927662415 Chr.16: 2212062-2212063 on Build GRCh38
rs998955590 Chr.16: 2212103-2212103 on Build GRCh38
rs201312081 Chr.16: 2211961-2211961 on Build GRCh38
rs927631399 Chr.16: 2212255-2212255 on Build GRCh38
rs117181636 Chr.16: 2212024-2212024 on Build GRCh38
rs142776533 Chr.16: 2212260-2212260 on Build GRCh38
rs1017267418 Chr.16: 2211843-2211843 on Build GRCh38
rs1038777615 Chr.16: 2211988-2211988 on Build GRCh38
rs749012193 Chr.16: 2211965-2211965 on Build GRCh38
rs943119185 Chr.16: 2211858-2211858 on Build GRCh38
rs535743749 Chr.16: 2212219-2212219 on Build GRCh38
rs910365473 Chr.16: 2212000-2212000 on Build GRCh38
rs954579200 Chr.16: 2211987-2211987 on Build GRCh38
rs140873535 Chr.16: 2211885-2211885 on Build GRCh38
rs1038808210 Chr.16: 2212156-2212156 on Build GRCh38
rs964940541 Chr.16: 2212042-2212042 on Build GRCh38
rs909932087 Chr.16: 2212078-2212078 on Build GRCh38
rs752707230 Chr.16: 2212258-2212258 on Build GRCh38
rs553154452 Chr.16: 2212128-2212128 on Build GRCh38
rs1007321656 Chr.16: 2211980-2211980 on Build GRCh38
rs147954828 Chr.16: 2212037-2212037 on Build GRCh38
rs964665539 Chr.16: 2212144-2212144 on Build GRCh38
rs530489493 Chr.16: 2211869-2211869 on Build GRCh38
rs957662438 Chr.16: 2211852-2211852 on Build GRCh38
rs182633436 Chr.16: 2212143-2212143 on Build GRCh38
rs765458777 Chr.16: 2212163-2212163 on Build GRCh38
rs550284464 Chr.16: 2211872-2211872 on Build GRCh38
rs1007660419 Chr.16: 2211827-2211827 on Build GRCh38

View Full Product Details