Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2238383-2238883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNASE1L2
Entrez Gene ID
1775
Gene Name
deoxyribonuclease I-like 2
Gene Aliases
DNAS1L2
UniGene
Hs.103503
Gene Chromosome Location
Chr.16: 2236252-2238711 on Build GRCh38
Transcripts
NM_001301680.1 NM_001374.2 XM_011522399.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1204229 c.868C>G p.P290A Chr.16: 2238386-2238386 on Build GRCh38
3817544 c.885C>G p.L295L Chr.16: 2238403-2238403 on Build GRCh38
6647453 c.895C>T p.R299* Chr.16: 2238413-2238413 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1039586776 Chr.16: 2238719-2238719 on Build GRCh38
rs563281136 Chr.16: 2238697-2238697 on Build GRCh38
rs892634927 Chr.16: 2238460-2238460 on Build GRCh38
rs979144597 Chr.16: 2238823-2238823 on Build GRCh38
rs989091674 Chr.16: 2238776-2238776 on Build GRCh38
rs923074729 Chr.16: 2238583-2238583 on Build GRCh38
rs892688765 Chr.16: 2238615-2238615 on Build GRCh38
rs751748992 Chr.16: 2238445-2238446 on Build GRCh38
rs952693616 Chr.16: 2238698-2238698 on Build GRCh38
rs961944239 Chr.16: 2238722-2238722 on Build GRCh38
rs759034902 Chr.16: 2238464-2238464 on Build GRCh38
rs200375931 Chr.16: 2238447-2238447 on Build GRCh38
rs769030883 Chr.16: 2238445-2238445 on Build GRCh38
rs78961807 Chr.16: 2238488-2238488 on Build GRCh38
rs546802071 Chr.16: 2238522-2238522 on Build GRCh38
rs576005895 Chr.16: 2238413-2238413 on Build GRCh38
rs367824378 Chr.16: 2238856-2238856 on Build GRCh38
rs189957083 Chr.16: 2238427-2238427 on Build GRCh38
rs146173984 Chr.16: 2238582-2238582 on Build GRCh38
rs539484003 Chr.16: 2238678-2238678 on Build GRCh38
rs1053331197 Chr.16: 2238507-2238507 on Build GRCh38
rs773796074 Chr.16: 2238458-2238458 on Build GRCh38
rs1034093332 Chr.16: 2238537-2238537 on Build GRCh38
rs369564280 Chr.16: 2238414-2238414 on Build GRCh38
rs1053917197 Chr.16: 2238459-2238459 on Build GRCh38
rs1050140918 Chr.16: 2238590-2238590 on Build GRCh38
rs7199539 Chr.16: 2238588-2238588 on Build GRCh38
rs900554113 Chr.16: 2238548-2238548 on Build GRCh38
rs994938089 Chr.16: 2238561-2238561 on Build GRCh38
rs752363403 Chr.16: 2238468-2238468 on Build GRCh38
rs912246075 Chr.16: 2238857-2238857 on Build GRCh38
rs767083489 Chr.16: 2238465-2238465 on Build GRCh38
rs772841224 Chr.16: 2238664-2238664 on Build GRCh38
rs181964818 Chr.16: 2238811-2238811 on Build GRCh38
rs1002232838 Chr.16: 2238530-2238530 on Build GRCh38
rs983388997 Chr.16: 2238415-2238415 on Build GRCh38
rs910212905 Chr.16: 2238592-2238592 on Build GRCh38
rs1025069653 Chr.16: 2238492-2238492 on Build GRCh38
rs914177210 Chr.16: 2238785-2238785 on Build GRCh38
rs780278307 Chr.16: 2238431-2238432 on Build GRCh38
rs762346037 Chr.16: 2238450-2238450 on Build GRCh38
rs1009665282 Chr.16: 2238471-2238471 on Build GRCh38
rs565833535 Chr.16: 2238550-2238550 on Build GRCh38
rs530927808 Chr.16: 2238409-2238409 on Build GRCh38
rs906656614 Chr.16: 2238665-2238665 on Build GRCh38
rs536634078 Chr.16: 2238600-2238600 on Build GRCh38
rs754497892 Chr.16: 2238576-2238576 on Build GRCh38
rs779916878 Chr.16: 2238528-2238528 on Build GRCh38
rs570373828 Chr.16: 2238650-2238650 on Build GRCh38
rs934896038 Chr.16: 2238496-2238496 on Build GRCh38
rs370539824 Chr.16: 2238481-2238481 on Build GRCh38
rs1035036127 Chr.16: 2238777-2238777 on Build GRCh38
rs1017777141 Chr.16: 2238538-2238538 on Build GRCh38
rs1002690317 Chr.16: 2238696-2238696 on Build GRCh38
rs564410995 Chr.16: 2238457-2238457 on Build GRCh38
rs1046981429 Chr.16: 2238652-2238652 on Build GRCh38
rs956460586 Chr.16: 2238833-2238833 on Build GRCh38
rs543484622 Chr.16: 2238601-2238601 on Build GRCh38
rs909090486 Chr.16: 2238554-2238554 on Build GRCh38
rs923733769 Chr.16: 2238829-2238829 on Build GRCh38
rs576043935 Chr.16: 2238797-2238797 on Build GRCh38
rs144019338 Chr.16: 2238767-2238767 on Build GRCh38
rs746987312 Chr.16: 2238442-2238442 on Build GRCh38
rs984714201 Chr.16: 2238553-2238553 on Build GRCh38
rs753381751 Chr.16: 2238549-2238549 on Build GRCh38
rs1012184181 Chr.16: 2238813-2238813 on Build GRCh38
rs753791066 Chr.16: 2238406-2238406 on Build GRCh38
rs1050502982 Chr.16: 2238422-2238422 on Build GRCh38
rs755756378 Chr.16: 2238515-2238515 on Build GRCh38
rs186347579 Chr.16: 2238820-2238820 on Build GRCh38
rs971204958 Chr.16: 2238529-2238529 on Build GRCh38
rs994908465 Chr.16: 2238725-2238725 on Build GRCh38
rs549028368 Chr.16: 2238807-2238807 on Build GRCh38
rs936352292 Chr.16: 2238771-2238771 on Build GRCh38
rs954521783 Chr.16: 2238741-2238741 on Build GRCh38
rs772360045 Chr.16: 2238421-2238421 on Build GRCh38
rs373512956 Chr.16: 2238444-2238444 on Build GRCh38
rs62040663 Chr.16: 2238520-2238520 on Build GRCh38
rs35631920 Chr.16: 2238465-2238465 on Build GRCh38
rs28586887 Chr.16: 2238702-2238702 on Build GRCh38
rs544260811 Chr.16: 2238815-2238815 on Build GRCh38
rs374477903 Chr.16: 2238761-2238761 on Build GRCh38
rs116420388 Chr.16: 2238735-2238735 on Build GRCh38
rs780407888 Chr.16: 2238437-2238437 on Build GRCh38
rs528279633 Chr.16: 2238559-2238559 on Build GRCh38
rs772659693 Chr.16: 2238701-2238701 on Build GRCh38
rs879550849 Chr.16: 2238625-2238625 on Build GRCh38
rs162301 Chr.16: 2238740-2238740 on Build GRCh38
rs1004794792 Chr.16: 2238587-2238587 on Build GRCh38

View Full Product Details