Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2244316-2244808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECI1
Entrez Gene ID
1632
Gene Name
enoyl-CoA delta isomerase 1
Gene Aliases
DCI
UniGene
Hs.403436
Gene Chromosome Location
Chr.16: 2239872-2251601 on Build GRCh38
Transcripts
NM_001178029.1 NM_001919.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7417750 c.335A>T p.E112V Chr.16: 2244512-2244512 on Build GRCh38
4602755 c.382G>A p.V128I Chr.16: 2244465-2244465 on Build GRCh38
7365819 c.421G>A p.V141M Chr.16: 2244426-2244426 on Build GRCh38
968679 c.298C>T p.R100C Chr.16: 2244549-2244549 on Build GRCh38
4748656 c.355G>A p.A119T Chr.16: 2244492-2244492 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760423250 Chr.16: 2244445-2244445 on Build GRCh38
rs781561935 Chr.16: 2244414-2244414 on Build GRCh38
rs368708095 Chr.16: 2244372-2244372 on Build GRCh38
rs1040214223 Chr.16: 2244761-2244761 on Build GRCh38
rs891257091 Chr.16: 2244553-2244553 on Build GRCh38
rs140753801 Chr.16: 2244439-2244439 on Build GRCh38
rs544132422 Chr.16: 2244533-2244533 on Build GRCh38
rs160555 Chr.16: 2244354-2244354 on Build GRCh38
rs370772682 Chr.16: 2244572-2244572 on Build GRCh38
rs150691746 Chr.16: 2244405-2244405 on Build GRCh38
rs571640971 Chr.16: 2244700-2244700 on Build GRCh38
rs930232433 Chr.16: 2244662-2244662 on Build GRCh38
rs772989313 Chr.16: 2244502-2244502 on Build GRCh38
rs909540211 Chr.16: 2244383-2244383 on Build GRCh38
rs533910308 Chr.16: 2244715-2244715 on Build GRCh38
rs977958254 Chr.16: 2244576-2244576 on Build GRCh38
rs113602974 Chr.16: 2244577-2244577 on Build GRCh38
rs745357478 Chr.16: 2244431-2244431 on Build GRCh38
rs774973333 Chr.16: 2244442-2244442 on Build GRCh38
rs753073208 Chr.16: 2244473-2244473 on Build GRCh38
rs538123254 Chr.16: 2244466-2244466 on Build GRCh38
rs571196670 Chr.16: 2244661-2244661 on Build GRCh38
rs749096740 Chr.16: 2244603-2244603 on Build GRCh38
rs372668766 Chr.16: 2244532-2244532 on Build GRCh38
rs148548119 Chr.16: 2244493-2244493 on Build GRCh38
rs762972307 Chr.16: 2244508-2244508 on Build GRCh38
rs544509062 Chr.16: 2244352-2244352 on Build GRCh38
rs748618022 Chr.16: 2244415-2244415 on Build GRCh38
rs181505389 Chr.16: 2244559-2244559 on Build GRCh38
rs373778276 Chr.16: 2244592-2244592 on Build GRCh38
rs200130130 Chr.16: 2244465-2244465 on Build GRCh38
rs1016717088 Chr.16: 2244545-2244545 on Build GRCh38
rs756529820 Chr.16: 2244474-2244474 on Build GRCh38
rs778709886 Chr.16: 2244406-2244406 on Build GRCh38
rs1042424043 Chr.16: 2244587-2244587 on Build GRCh38
rs377274938 Chr.16: 2244483-2244483 on Build GRCh38
rs771681200 Chr.16: 2244438-2244438 on Build GRCh38
rs375300423 Chr.16: 2244528-2244528 on Build GRCh38
rs777373321 Chr.16: 2244589-2244589 on Build GRCh38
rs771262093 Chr.16: 2244436-2244436 on Build GRCh38
rs142982644 Chr.16: 2244457-2244457 on Build GRCh38
rs376899686 Chr.16: 2244549-2244549 on Build GRCh38
rs771459648 Chr.16: 2244480-2244480 on Build GRCh38
rs768335613 Chr.16: 2244538-2244538 on Build GRCh38
rs199757340 Chr.16: 2244513-2244513 on Build GRCh38
rs750430412 Chr.16: 2244451-2244451 on Build GRCh38
rs560267973 Chr.16: 2244564-2244564 on Build GRCh38
rs770294190 Chr.16: 2244426-2244426 on Build GRCh38
rs767625370 Chr.16: 2244558-2244558 on Build GRCh38
rs755277651 Chr.16: 2244412-2244412 on Build GRCh38
rs199619754 Chr.16: 2244365-2244365 on Build GRCh38
rs374092458 Chr.16: 2244562-2244562 on Build GRCh38
rs899899647 Chr.16: 2244775-2244775 on Build GRCh38
rs765276090 Chr.16: 2244449-2244449 on Build GRCh38
rs371123821 Chr.16: 2244554-2244554 on Build GRCh38
rs764019052 Chr.16: 2244377-2244377 on Build GRCh38
rs373423708 Chr.16: 2244566-2244566 on Build GRCh38
rs747764348 Chr.16: 2244585-2244585 on Build GRCh38
rs528954080 Chr.16: 2244567-2244567 on Build GRCh38
rs369319156 Chr.16: 2244529-2244529 on Build GRCh38
rs760907228 Chr.16: 2244561-2244561 on Build GRCh38
rs779143999 Chr.16: 2244440-2244445 on Build GRCh38
rs1023928639 Chr.16: 2244724-2244724 on Build GRCh38
rs939079701 Chr.16: 2244507-2244507 on Build GRCh38
rs201346166 Chr.16: 2244447-2244447 on Build GRCh38
rs763165861 Chr.16: 2244641-2244641 on Build GRCh38
rs746287914 Chr.16: 2244453-2244453 on Build GRCh38
rs765368036 Chr.16: 2244400-2244400 on Build GRCh38
rs532962324 Chr.16: 2244357-2244357 on Build GRCh38
rs144791724 Chr.16: 2244446-2244446 on Build GRCh38
rs373850326 Chr.16: 2244341-2244341 on Build GRCh38
rs756441804 Chr.16: 2244523-2244523 on Build GRCh38
rs757282014 Chr.16: 2244395-2244395 on Build GRCh38
rs1025716252 Chr.16: 2244421-2244421 on Build GRCh38
rs971079960 Chr.16: 2244730-2244730 on Build GRCh38
rs145712373 Chr.16: 2244515-2244515 on Build GRCh38
rs373950696 Chr.16: 2244514-2244514 on Build GRCh38
rs755796344 Chr.16: 2244588-2244588 on Build GRCh38
rs551647547 Chr.16: 2244643-2244643 on Build GRCh38
rs376657107 Chr.16: 2244492-2244492 on Build GRCh38
rs533971343 Chr.16: 2244484-2244484 on Build GRCh38
rs529477231 Chr.16: 2244407-2244407 on Build GRCh38
rs775271968 Chr.16: 2244370-2244370 on Build GRCh38
rs149722709 Chr.16: 2244544-2244544 on Build GRCh38
rs1020918518 Chr.16: 2244339-2244339 on Build GRCh38
rs372341953 Chr.16: 2244479-2244479 on Build GRCh38
rs926926146 Chr.16: 2244776-2244776 on Build GRCh38
rs562961829 Chr.16: 2244636-2244636 on Build GRCh38
rs753802399 Chr.16: 2244385-2244385 on Build GRCh38
rs775162611 Chr.16: 2244464-2244471 on Build GRCh38
rs145543160 Chr.16: 2244548-2244548 on Build GRCh38
rs531978368 Chr.16: 2244638-2244638 on Build GRCh38
rs769775669 Chr.16: 2244496-2244496 on Build GRCh38
rs773793866 Chr.16: 2244430-2244430 on Build GRCh38
rs1022662691 Chr.16: 2244570-2244570 on Build GRCh38
rs1047271187 Chr.16: 2244702-2244702 on Build GRCh38
rs200369056 Chr.16: 2244358-2244358 on Build GRCh38
rs368064307 Chr.16: 2244581-2244581 on Build GRCh38
rs770918670 Chr.16: 2244550-2244550 on Build GRCh38
rs944185792 Chr.16: 2244719-2244719 on Build GRCh38
rs764286026 Chr.16: 2244695-2244695 on Build GRCh38
rs771690070 Chr.16: 2244458-2244462 on Build GRCh38
rs994278121 Chr.16: 2244417-2244417 on Build GRCh38
rs762935665 Chr.16: 2244455-2244455 on Build GRCh38
rs761771407 Chr.16: 2244448-2244448 on Build GRCh38
rs778249875 Chr.16: 2244475-2244475 on Build GRCh38
rs774213990 Chr.16: 2244552-2244552 on Build GRCh38
rs969488917 Chr.16: 2244642-2244642 on Build GRCh38

View Full Product Details