Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2246671-2247080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECI1
Entrez Gene ID
1632
Gene Name
enoyl-CoA delta isomerase 1
Gene Aliases
DCI
UniGene
Hs.403436
Gene Chromosome Location
Chr.16: 2239872-2251601 on Build GRCh38
Transcripts
NM_001178029.1 NM_001919.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4680082 c.276C>T p.R92R Chr.16: 2246877-2246877 on Build GRCh38
6450960 c.274C>T p.R92C Chr.16: 2246879-2246879 on Build GRCh38
7365820 c.244C>G p.L82V Chr.16: 2246909-2246909 on Build GRCh38
325127 c.275G>A p.R92H Chr.16: 2246878-2246878 on Build GRCh38
6078850 c.268A>T p.S90C Chr.16: 2246885-2246885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759007674 Chr.16: 2246855-2246855 on Build GRCh38
rs562081321 Chr.16: 2247054-2247054 on Build GRCh38
rs748337504 Chr.16: 2246985-2246985 on Build GRCh38
rs766980032 Chr.16: 2246954-2246954 on Build GRCh38
rs986195060 Chr.16: 2246774-2246774 on Build GRCh38
rs767811781 Chr.16: 2247014-2247015 on Build GRCh38
rs936689695 Chr.16: 2246794-2246794 on Build GRCh38
rs376093416 Chr.16: 2247005-2247005 on Build GRCh38
rs763497833 Chr.16: 2246939-2246939 on Build GRCh38
rs201995306 Chr.16: 2246945-2246945 on Build GRCh38
rs72766670 Chr.16: 2246981-2246981 on Build GRCh38
rs577583578 Chr.16: 2246888-2246888 on Build GRCh38
rs923475641 Chr.16: 2246762-2246762 on Build GRCh38
rs576184725 Chr.16: 2246878-2246878 on Build GRCh38
rs984414564 Chr.16: 2246711-2246711 on Build GRCh38
rs753290457 Chr.16: 2247038-2247038 on Build GRCh38
rs141206306 Chr.16: 2246876-2246876 on Build GRCh38
rs146513443 Chr.16: 2246877-2246877 on Build GRCh38
rs776799008 Chr.16: 2246791-2246791 on Build GRCh38
rs762885059 Chr.16: 2246956-2246956 on Build GRCh38
rs770144171 Chr.16: 2246928-2246928 on Build GRCh38
rs750942541 Chr.16: 2246852-2246852 on Build GRCh38
rs199680856 Chr.16: 2246843-2246843 on Build GRCh38
rs770962592 Chr.16: 2246788-2246788 on Build GRCh38
rs763564542 Chr.16: 2246961-2246961 on Build GRCh38
rs776775804 Chr.16: 2246833-2246833 on Build GRCh38
rs746958924 Chr.16: 2246982-2246982 on Build GRCh38
rs774317342 Chr.16: 2246814-2246814 on Build GRCh38
rs369676219 Chr.16: 2246827-2246827 on Build GRCh38
rs910626189 Chr.16: 2246787-2246787 on Build GRCh38
rs534677999 Chr.16: 2246781-2246781 on Build GRCh38
rs201913960 Chr.16: 2246875-2246875 on Build GRCh38
rs112729404 Chr.16: 2246926-2246926 on Build GRCh38
rs764387323 Chr.16: 2246831-2246831 on Build GRCh38
rs140154367 Chr.16: 2246978-2246978 on Build GRCh38
rs763622996 Chr.16: 2247036-2247036 on Build GRCh38
rs972689132 Chr.16: 2246743-2246743 on Build GRCh38
rs773121638 Chr.16: 2246881-2246884 on Build GRCh38
rs747060221 Chr.16: 2246739-2246739 on Build GRCh38
rs776569985 Chr.16: 2246859-2246860 on Build GRCh38
rs781152332 Chr.16: 2246984-2246984 on Build GRCh38
rs559740681 Chr.16: 2246925-2246925 on Build GRCh38
rs900408381 Chr.16: 2246716-2246716 on Build GRCh38
rs773473142 Chr.16: 2246718-2246718 on Build GRCh38
rs116419478 Chr.16: 2246702-2246702 on Build GRCh38
rs745763135 Chr.16: 2246879-2246879 on Build GRCh38
rs933459132 Chr.16: 2246780-2246780 on Build GRCh38
rs745857313 Chr.16: 2246821-2246821 on Build GRCh38
rs765283725 Chr.16: 2246879-2246880 on Build GRCh38
rs565352095 Chr.16: 2246741-2246741 on Build GRCh38
rs372701605 Chr.16: 2246883-2246883 on Build GRCh38
rs962275162 Chr.16: 2246738-2246738 on Build GRCh38
rs770701669 Chr.16: 2246812-2246812 on Build GRCh38
rs750052090 Chr.16: 2246953-2246953 on Build GRCh38
rs978942017 Chr.16: 2246849-2246849 on Build GRCh38
rs369232279 Chr.16: 2246857-2246857 on Build GRCh38
rs972359296 Chr.16: 2246946-2246946 on Build GRCh38
rs759986682 Chr.16: 2247034-2247034 on Build GRCh38
rs551728983 Chr.16: 2246732-2246732 on Build GRCh38
rs752994056 Chr.16: 2247016-2247029 on Build GRCh38
rs768068993 Chr.16: 2246854-2246854 on Build GRCh38
rs773718798 Chr.16: 2246935-2246935 on Build GRCh38
rs140831866 Chr.16: 2246909-2246909 on Build GRCh38
rs755637402 Chr.16: 2246869-2246869 on Build GRCh38
rs762233537 Chr.16: 2246845-2246845 on Build GRCh38
rs3803695 Chr.16: 2246850-2246850 on Build GRCh38
rs780047225 Chr.16: 2246895-2246895 on Build GRCh38
rs768684246 Chr.16: 2246910-2246910 on Build GRCh38
rs771466917 Chr.16: 2247017-2247017 on Build GRCh38
rs542304564 Chr.16: 2247053-2247053 on Build GRCh38
rs758242540 Chr.16: 2246979-2246979 on Build GRCh38
rs989535894 Chr.16: 2247047-2247047 on Build GRCh38
rs945960448 Chr.16: 2246839-2246839 on Build GRCh38
rs760138466 Chr.16: 2246839-2246847 on Build GRCh38
rs886203258 Chr.16: 2246751-2246751 on Build GRCh38
rs774980164 Chr.16: 2247021-2247021 on Build GRCh38
rs770154838 Chr.16: 2246987-2246987 on Build GRCh38
rs760206331 Chr.16: 2246960-2246960 on Build GRCh38
rs529881535 Chr.16: 2247056-2247056 on Build GRCh38
rs148482178 Chr.16: 2246860-2246860 on Build GRCh38
rs202006480 Chr.16: 2246893-2246893 on Build GRCh38
rs772092995 Chr.16: 2246826-2246826 on Build GRCh38
rs761658196 Chr.16: 2246865-2246876 on Build GRCh38
rs752178469 Chr.16: 2246958-2246958 on Build GRCh38
rs763405963 Chr.16: 2247006-2247006 on Build GRCh38
rs1012347181 Chr.16: 2247044-2247044 on Build GRCh38
rs906126278 Chr.16: 2247049-2247049 on Build GRCh38

View Full Product Details