Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2251211-2251698 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECI1
Entrez Gene ID
1632
Gene Name
enoyl-CoA delta isomerase 1
Gene Aliases
DCI
UniGene
Hs.403436
Gene Chromosome Location
Chr.16: 2239872-2251601 on Build GRCh38
Transcripts
NM_001178029.1 NM_001919.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3948383 c.56C>T p.A19V Chr.16: 2251426-2251426 on Build GRCh38
4550365 c.49G>A p.A17T Chr.16: 2251518-2251518 on Build GRCh38
5627478 c.40C>G p.L14V Chr.16: 2251527-2251527 on Build GRCh38
5719120 c.53G>A p.G18E Chr.16: 2251429-2251429 on Build GRCh38
434911 c.48C>G p.R16R Chr.16: 2251519-2251519 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs940679333 Chr.16: 2251323-2251323 on Build GRCh38
rs386787815 Chr.16: 2251268-2251271 on Build GRCh38
rs112148283 Chr.16: 2251337-2251337 on Build GRCh38
rs772827843 Chr.16: 2251515-2251515 on Build GRCh38
rs764087034 Chr.16: 2251470-2251471 on Build GRCh38
rs984926166 Chr.16: 2251566-2251566 on Build GRCh38
rs746202495 Chr.16: 2251578-2251578 on Build GRCh38
rs754862311 Chr.16: 2251523-2251523 on Build GRCh38
rs550289429 Chr.16: 2251630-2251630 on Build GRCh38
rs890177950 Chr.16: 2251330-2251330 on Build GRCh38
rs463317 Chr.16: 2251438-2251438 on Build GRCh38
rs998685060 Chr.16: 2251306-2251306 on Build GRCh38
rs561290323 Chr.16: 2251604-2251604 on Build GRCh38
rs894404854 Chr.16: 2251235-2251235 on Build GRCh38
rs774613604 Chr.16: 2251475-2251475 on Build GRCh38
rs55650311 Chr.16: 2251268-2251268 on Build GRCh38
rs866855432 Chr.16: 2251353-2251353 on Build GRCh38
rs555879079 Chr.16: 2251433-2251433 on Build GRCh38
rs199982693 Chr.16: 2251598-2251598 on Build GRCh38
rs948822255 Chr.16: 2251668-2251668 on Build GRCh38
rs778024807 Chr.16: 2251538-2251538 on Build GRCh38
rs964268127 Chr.16: 2251347-2251347 on Build GRCh38
rs1029513797 Chr.16: 2251390-2251390 on Build GRCh38
rs777908966 Chr.16: 2251516-2251516 on Build GRCh38
rs778110537 Chr.16: 2251464-2251464 on Build GRCh38
rs998572890 Chr.16: 2251378-2251397 on Build GRCh38
rs754902204 Chr.16: 2251356-2251356 on Build GRCh38
rs988881339 Chr.16: 2251504-2251504 on Build GRCh38
rs915951596 Chr.16: 2251467-2251467 on Build GRCh38
rs772551198 Chr.16: 2251584-2251584 on Build GRCh38
rs1006665698 Chr.16: 2251474-2251474 on Build GRCh38
rs748164894 Chr.16: 2251417-2251417 on Build GRCh38
rs1007130560 Chr.16: 2251662-2251662 on Build GRCh38
rs950259459 Chr.16: 2251274-2251274 on Build GRCh38
rs927433603 Chr.16: 2251310-2251310 on Build GRCh38
rs761467772 Chr.16: 2251267-2251267 on Build GRCh38
rs757560123 Chr.16: 2251573-2251573 on Build GRCh38
rs182577651 Chr.16: 2251585-2251585 on Build GRCh38
rs751463939 Chr.16: 2251349-2251349 on Build GRCh38
rs779276270 Chr.16: 2251574-2251574 on Build GRCh38
rs761303776 Chr.16: 2251493-2251493 on Build GRCh38
rs756330053 Chr.16: 2251447-2251447 on Build GRCh38
rs746322253 Chr.16: 2251477-2251477 on Build GRCh38
rs937440091 Chr.16: 2251313-2251313 on Build GRCh38
rs776120071 Chr.16: 2251487-2251487 on Build GRCh38
rs933912653 Chr.16: 2251622-2251622 on Build GRCh38
rs998066549 Chr.16: 2251388-2251388 on Build GRCh38
rs186632369 Chr.16: 2251660-2251660 on Build GRCh38
rs11541763 Chr.16: 2251519-2251519 on Build GRCh38
rs992945500 Chr.16: 2251638-2251638 on Build GRCh38
rs953020929 Chr.16: 2251370-2251370 on Build GRCh38
rs563807767 Chr.16: 2251653-2251653 on Build GRCh38
rs1043393759 Chr.16: 2251500-2251500 on Build GRCh38
rs1017192129 Chr.16: 2251344-2251344 on Build GRCh38
rs771377106 Chr.16: 2251469-2251469 on Build GRCh38
rs1036989471 Chr.16: 2251339-2251339 on Build GRCh38
rs576283920 Chr.16: 2251669-2251669 on Build GRCh38
rs536340765 Chr.16: 2251318-2251318 on Build GRCh38
rs749574834 Chr.16: 2251465-2251465 on Build GRCh38
rs904387087 Chr.16: 2251287-2251287 on Build GRCh38
rs199944223 Chr.16: 2251526-2251526 on Build GRCh38
rs530086009 Chr.16: 2251620-2251620 on Build GRCh38
rs772468885 Chr.16: 2251483-2251483 on Build GRCh38
rs900211278 Chr.16: 2251560-2251560 on Build GRCh38
rs55985840 Chr.16: 2251271-2251271 on Build GRCh38
rs756205408 Chr.16: 2251413-2251425 on Build GRCh38
rs774121673 Chr.16: 2251612-2251612 on Build GRCh38
rs904419465 Chr.16: 2251517-2251517 on Build GRCh38
rs959113454 Chr.16: 2251554-2251554 on Build GRCh38
rs572664579 Chr.16: 2251591-2251591 on Build GRCh38
rs868120216 Chr.16: 2251582-2251582 on Build GRCh38
rs781278028 Chr.16: 2251363-2251363 on Build GRCh38
rs1015730243 Chr.16: 2251630-2251632 on Build GRCh38
rs981630859 Chr.16: 2251522-2251522 on Build GRCh38
rs464176 Chr.16: 2251406-2251406 on Build GRCh38
rs992829307 Chr.16: 2251466-2251466 on Build GRCh38
rs758158418 Chr.16: 2251291-2251291 on Build GRCh38
rs749484668 Chr.16: 2251545-2251545 on Build GRCh38
rs887811846 Chr.16: 2251322-2251322 on Build GRCh38
rs938715003 Chr.16: 2251394-2251394 on Build GRCh38
rs113641186 Chr.16: 2251511-2251511 on Build GRCh38
rs915982514 Chr.16: 2251657-2251657 on Build GRCh38
rs763873346 Chr.16: 2251255-2251255 on Build GRCh38
rs1004796054 Chr.16: 2251629-2251630 on Build GRCh38
rs1027231090 Chr.16: 2251364-2251364 on Build GRCh38
rs756244379 Chr.16: 2251527-2251527 on Build GRCh38
rs908632841 Chr.16: 2251393-2251393 on Build GRCh38
rs765936249 Chr.16: 2251607-2251607 on Build GRCh38
rs889653743 Chr.16: 2251652-2251652 on Build GRCh38

View Full Product Details