Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2275739-2276229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA3
Entrez Gene ID
21
Gene Name
ATP binding cassette subfamily A member 3
Gene Aliases
ABC-C, ABC3, EST111653, LBM180, SMDP3
UniGene
Hs.26630
Gene Chromosome Location
Chr.16: 2275878-2340746 on Build GRCh38
Transcripts
NM_001089.2

SNP Detail

SNP ID Chromosome Location
rs953246567 Chr.16: 2275852-2275852 on Build GRCh38
rs927411407 Chr.16: 2275944-2275944 on Build GRCh38
rs868380644 Chr.16: 2275823-2275823 on Build GRCh38
rs886051804 Chr.16: 2276053-2276053 on Build GRCh38
rs537157538 Chr.16: 2276079-2276079 on Build GRCh38
rs997294927 Chr.16: 2275837-2275837 on Build GRCh38
rs113314595 Chr.16: 2275858-2275858 on Build GRCh38
rs906964627 Chr.16: 2275905-2275905 on Build GRCh38
rs971857238 Chr.16: 2275892-2275892 on Build GRCh38
rs941363956 Chr.16: 2276080-2276080 on Build GRCh38
rs576879285 Chr.16: 2275798-2275798 on Build GRCh38
rs888067972 Chr.16: 2275799-2275799 on Build GRCh38
rs574578941 Chr.16: 2276144-2276144 on Build GRCh38
rs937553928 Chr.16: 2275780-2275780 on Build GRCh38
rs749749806 Chr.16: 2276067-2276067 on Build GRCh38
rs543592832 Chr.16: 2276153-2276153 on Build GRCh38
rs574589921 Chr.16: 2275774-2275774 on Build GRCh38
rs913257754 Chr.16: 2275873-2275873 on Build GRCh38
rs532849664 Chr.16: 2275911-2275911 on Build GRCh38
rs528439325 Chr.16: 2275981-2275981 on Build GRCh38
rs1038729989 Chr.16: 2275857-2275857 on Build GRCh38
rs558539302 Chr.16: 2276106-2276106 on Build GRCh38
rs1036414441 Chr.16: 2276028-2276028 on Build GRCh38
rs554616359 Chr.16: 2276138-2276138 on Build GRCh38
rs368709738 Chr.16: 2276062-2276062 on Build GRCh38
rs45471792 Chr.16: 2275918-2275918 on Build GRCh38
rs940894484 Chr.16: 2276152-2276152 on Build GRCh38
rs1004543025 Chr.16: 2275814-2275814 on Build GRCh38
rs538910072 Chr.16: 2276094-2276094 on Build GRCh38
rs1032165706 Chr.16: 2276068-2276068 on Build GRCh38
rs978268145 Chr.16: 2275781-2275781 on Build GRCh38
rs756870810 Chr.16: 2275791-2275791 on Build GRCh38
rs745540255 Chr.16: 2275851-2275851 on Build GRCh38
rs1050550947 Chr.16: 2276210-2276210 on Build GRCh38
rs183637156 Chr.16: 2275934-2275934 on Build GRCh38
rs1046293314 Chr.16: 2275760-2275760 on Build GRCh38
rs940944621 Chr.16: 2275982-2275982 on Build GRCh38
rs888904669 Chr.16: 2276052-2276052 on Build GRCh38
rs958805832 Chr.16: 2276139-2276139 on Build GRCh38
rs141507206 Chr.16: 2275969-2275969 on Build GRCh38
rs1000648903 Chr.16: 2276065-2276065 on Build GRCh38
rs373842568 Chr.16: 2275842-2275842 on Build GRCh38
rs565988498 Chr.16: 2276107-2276107 on Build GRCh38
rs997642336 Chr.16: 2276042-2276042 on Build GRCh38
rs1001317347 Chr.16: 2276110-2276110 on Build GRCh38
rs1020668703 Chr.16: 2276087-2276087 on Build GRCh38
rs534278448 Chr.16: 2276109-2276109 on Build GRCh38
rs774828715 Chr.16: 2276204-2276204 on Build GRCh38
rs45613643 Chr.16: 2275859-2275859 on Build GRCh38
rs1049292293 Chr.16: 2275970-2275970 on Build GRCh38
rs1000699551 Chr.16: 2275854-2275854 on Build GRCh38
rs569543243 Chr.16: 2276093-2276093 on Build GRCh38
rs1011196042 Chr.16: 2275957-2275957 on Build GRCh38
rs568167293 Chr.16: 2276049-2276049 on Build GRCh38
rs922890319 Chr.16: 2276178-2276178 on Build GRCh38
rs901571238 Chr.16: 2275831-2275831 on Build GRCh38
rs1045978327 Chr.16: 2276095-2276095 on Build GRCh38
rs1033985549 Chr.16: 2275919-2275919 on Build GRCh38
rs371221845 Chr.16: 2276146-2276146 on Build GRCh38
rs755535517 Chr.16: 2275856-2275856 on Build GRCh38
rs1045540169 Chr.16: 2275866-2275866 on Build GRCh38
rs574838476 Chr.16: 2275985-2275985 on Build GRCh38
rs548584647 Chr.16: 2276002-2276002 on Build GRCh38
rs901523515 Chr.16: 2276041-2276041 on Build GRCh38
rs911189454 Chr.16: 2275834-2275837 on Build GRCh38
rs188688704 Chr.16: 2276083-2276083 on Build GRCh38
rs978680776 Chr.16: 2276014-2276015 on Build GRCh38
rs112848120 Chr.16: 2275935-2275935 on Build GRCh38
rs780869300 Chr.16: 2275825-2275825 on Build GRCh38
rs934037064 Chr.16: 2275815-2275815 on Build GRCh38
rs909397724 Chr.16: 2276147-2276147 on Build GRCh38

View Full Product Details