Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2280920-2281420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA3
Entrez Gene ID
21
Gene Name
ATP binding cassette subfamily A member 3
Gene Aliases
ABC-C, ABC3, EST111653, LBM180, SMDP3
UniGene
Hs.26630
Gene Chromosome Location
Chr.16: 2275878-2340746 on Build GRCh38
Transcripts
NM_001089.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6589679 c.4261G>A p.G1421R Chr.16: 2281125-2281125 on Build GRCh38
6589711 c.4353C>T p.V1451V Chr.16: 2281033-2281033 on Build GRCh38
5606285 c.4319C>T p.A1440V Chr.16: 2281067-2281067 on Build GRCh38
252311 c.4354G>A p.G1452R Chr.16: 2281032-2281032 on Build GRCh38
702999 c.4164G>T p.K1388N Chr.16: 2281381-2281381 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754583762 Chr.16: 2281340-2281340 on Build GRCh38
rs757835350 Chr.16: 2281331-2281331 on Build GRCh38
rs764069673 Chr.16: 2281118-2281118 on Build GRCh38
rs773955508 Chr.16: 2281112-2281112 on Build GRCh38
rs778378721 Chr.16: 2281150-2281150 on Build GRCh38
rs767302159 Chr.16: 2281033-2281033 on Build GRCh38
rs1033438639 Chr.16: 2281337-2281337 on Build GRCh38
rs879019462 Chr.16: 2281324-2281324 on Build GRCh38
rs769129570 Chr.16: 2280995-2280995 on Build GRCh38
rs779442941 Chr.16: 2281333-2281333 on Build GRCh38
rs141811010 Chr.16: 2281265-2281265 on Build GRCh38
rs889654694 Chr.16: 2280975-2280975 on Build GRCh38
rs138340688 Chr.16: 2281171-2281171 on Build GRCh38
rs751140771 Chr.16: 2281334-2281334 on Build GRCh38
rs376918831 Chr.16: 2281231-2281231 on Build GRCh38
rs770589758 Chr.16: 2281025-2281025 on Build GRCh38
rs199852776 Chr.16: 2281081-2281081 on Build GRCh38
rs756695650 Chr.16: 2281149-2281149 on Build GRCh38
rs775345113 Chr.16: 2281044-2281044 on Build GRCh38
rs760426494 Chr.16: 2281117-2281117 on Build GRCh38
rs397518429 Chr.16: 2281064-2281064 on Build GRCh38
rs745530191 Chr.16: 2281113-2281113 on Build GRCh38
rs1033077802 Chr.16: 2281057-2281057 on Build GRCh38
rs772585636 Chr.16: 2280984-2280984 on Build GRCh38
rs369224394 Chr.16: 2281180-2281180 on Build GRCh38
rs771766048 Chr.16: 2281114-2281114 on Build GRCh38
rs368151842 Chr.16: 2281245-2281245 on Build GRCh38
rs764001106 Chr.16: 2281059-2281059 on Build GRCh38
rs765414485 Chr.16: 2281127-2281127 on Build GRCh38
rs760644085 Chr.16: 2281055-2281055 on Build GRCh38
rs781690684 Chr.16: 2281139-2281139 on Build GRCh38
rs751926114 Chr.16: 2281134-2281134 on Build GRCh38
rs148317800 Chr.16: 2281208-2281208 on Build GRCh38
rs769824378 Chr.16: 2281246-2281246 on Build GRCh38
rs376588751 Chr.16: 2280991-2280991 on Build GRCh38
rs747556947 Chr.16: 2280992-2280992 on Build GRCh38
rs114420154 Chr.16: 2281235-2281235 on Build GRCh38
rs773422546 Chr.16: 2281183-2281183 on Build GRCh38
rs147036502 Chr.16: 2281133-2281133 on Build GRCh38
rs569840321 Chr.16: 2281295-2281295 on Build GRCh38
rs1045444223 Chr.16: 2281294-2281294 on Build GRCh38
rs759908162 Chr.16: 2281203-2281203 on Build GRCh38
rs763166660 Chr.16: 2281191-2281191 on Build GRCh38
rs773336518 Chr.16: 2281250-2281250 on Build GRCh38
rs569956903 Chr.16: 2280980-2280980 on Build GRCh38
rs747392070 Chr.16: 2281093-2281093 on Build GRCh38
rs780579884 Chr.16: 2281087-2281087 on Build GRCh38
rs765505615 Chr.16: 2281082-2281082 on Build GRCh38
rs200566505 Chr.16: 2281144-2281144 on Build GRCh38
rs896878101 Chr.16: 2281029-2281029 on Build GRCh38
rs750616312 Chr.16: 2281132-2281132 on Build GRCh38
rs1029584079 Chr.16: 2281015-2281015 on Build GRCh38
rs774287586 Chr.16: 2281389-2281389 on Build GRCh38
rs755504528 Chr.16: 2281095-2281095 on Build GRCh38
rs755998204 Chr.16: 2281373-2281373 on Build GRCh38
rs189737924 Chr.16: 2281096-2281096 on Build GRCh38
rs141811925 Chr.16: 2281104-2281104 on Build GRCh38
rs200204553 Chr.16: 2281174-2281174 on Build GRCh38
rs375783245 Chr.16: 2281032-2281032 on Build GRCh38
rs762696482 Chr.16: 2281387-2281387 on Build GRCh38
rs576334134 Chr.16: 2281011-2281011 on Build GRCh38
rs776453529 Chr.16: 2281125-2281125 on Build GRCh38
rs543588148 Chr.16: 2281276-2281276 on Build GRCh38
rs548311633 Chr.16: 2281253-2281253 on Build GRCh38
rs772158879 Chr.16: 2281396-2281396 on Build GRCh38
rs529812305 Chr.16: 2281264-2281264 on Build GRCh38
rs1001603955 Chr.16: 2281316-2281316 on Build GRCh38
rs780837626 Chr.16: 2281243-2281243 on Build GRCh38
rs759661714 Chr.16: 2281257-2281257 on Build GRCh38
rs763081102 Chr.16: 2281252-2281252 on Build GRCh38
rs750703859 Chr.16: 2281083-2281083 on Build GRCh38
rs375731527 Chr.16: 2281192-2281192 on Build GRCh38
rs768442294 Chr.16: 2281236-2281236 on Build GRCh38
rs373102317 Chr.16: 2281126-2281126 on Build GRCh38
rs753160597 Chr.16: 2281209-2281209 on Build GRCh38
rs1050037960 Chr.16: 2280963-2280963 on Build GRCh38
rs755350979 Chr.16: 2281135-2281135 on Build GRCh38
rs568231563 Chr.16: 2281255-2281255 on Build GRCh38
rs749244982 Chr.16: 2281384-2281384 on Build GRCh38
rs754373928 Chr.16: 2281272-2281272 on Build GRCh38
rs751755351 Chr.16: 2281201-2281201 on Build GRCh38
rs767845267 Chr.16: 2281258-2281258 on Build GRCh38
rs777817178 Chr.16: 2281377-2281377 on Build GRCh38
rs753024204 Chr.16: 2281260-2281260 on Build GRCh38
rs34902969 Chr.16: 2281095-2281096 on Build GRCh38
rs538210968 Chr.16: 2281060-2281060 on Build GRCh38
rs780938275 Chr.16: 2281341-2281341 on Build GRCh38
rs144301862 Chr.16: 2281173-2281173 on Build GRCh38
rs756605510 Chr.16: 2281214-2281214 on Build GRCh38
rs1006878314 Chr.16: 2281317-2281317 on Build GRCh38
rs538969220 Chr.16: 2281363-2281363 on Build GRCh38
rs369872536 Chr.16: 2281115-2281115 on Build GRCh38
rs149559041 Chr.16: 2281172-2281172 on Build GRCh38
rs896928717 Chr.16: 2281277-2281277 on Build GRCh38
rs372848304 Chr.16: 2281013-2281013 on Build GRCh38
rs141744909 Chr.16: 2281215-2281215 on Build GRCh38
rs138769732 Chr.16: 2281229-2281229 on Build GRCh38
rs772765938 Chr.16: 2280997-2280997 on Build GRCh38
rs558244985 Chr.16: 2281393-2281393 on Build GRCh38
rs71386663 Chr.16: 2281291-2281291 on Build GRCh38
rs933898218 Chr.16: 2280961-2280961 on Build GRCh38
rs532847219 Chr.16: 2281216-2281216 on Build GRCh38
rs572167727 Chr.16: 2281085-2281085 on Build GRCh38

View Full Product Details