Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2284188-2284680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA3
Entrez Gene ID
21
Gene Name
ATP binding cassette subfamily A member 3
Gene Aliases
ABC-C, ABC3, EST111653, LBM180, SMDP3
UniGene
Hs.26630
Gene Chromosome Location
Chr.16: 2275878-2340746 on Build GRCh38
Transcripts
NM_001089.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6589697 c.3721C>T p.L1241F Chr.16: 2284420-2284420 on Build GRCh38
4059372 c.3818A>G p.Y1273C Chr.16: 2284323-2284323 on Build GRCh38
4995641 c.3727A>C p.K1243Q Chr.16: 2284414-2284414 on Build GRCh38
4059373 c.3735G>T p.L1245L Chr.16: 2284406-2284406 on Build GRCh38
1166367 c.3741C>T p.H1247H Chr.16: 2284400-2284400 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745518190 Chr.16: 2284328-2284328 on Build GRCh38
rs776731355 Chr.16: 2284336-2284336 on Build GRCh38
rs758713237 Chr.16: 2284240-2284240 on Build GRCh38
rs745633178 Chr.16: 2284268-2284268 on Build GRCh38
rs779803150 Chr.16: 2284443-2284443 on Build GRCh38
rs771764375 Chr.16: 2284269-2284269 on Build GRCh38
rs936157181 Chr.16: 2284579-2284579 on Build GRCh38
rs559774168 Chr.16: 2284280-2284280 on Build GRCh38
rs141997453 Chr.16: 2284420-2284420 on Build GRCh38
rs140948738 Chr.16: 2284346-2284346 on Build GRCh38
rs544213510 Chr.16: 2284256-2284256 on Build GRCh38
rs369424856 Chr.16: 2284239-2284239 on Build GRCh38
rs147123460 Chr.16: 2284301-2284301 on Build GRCh38
rs557721019 Chr.16: 2284595-2284595 on Build GRCh38
rs775437521 Chr.16: 2284275-2284275 on Build GRCh38
rs776940610 Chr.16: 2284236-2284237 on Build GRCh38
rs896598359 Chr.16: 2284632-2284632 on Build GRCh38
rs575419471 Chr.16: 2284470-2284470 on Build GRCh38
rs371099401 Chr.16: 2284329-2284329 on Build GRCh38
rs1183064 Chr.16: 2284553-2284553 on Build GRCh38
rs750706025 Chr.16: 2284303-2284303 on Build GRCh38
rs534137725 Chr.16: 2284478-2284478 on Build GRCh38
rs113774992 Chr.16: 2284373-2284373 on Build GRCh38
rs769912251 Chr.16: 2284467-2284467 on Build GRCh38
rs985502095 Chr.16: 2284396-2284414 on Build GRCh38
rs200209913 Chr.16: 2284358-2284358 on Build GRCh38
rs752021996 Chr.16: 2284306-2284306 on Build GRCh38
rs773427768 Chr.16: 2284349-2284349 on Build GRCh38
rs1040023691 Chr.16: 2284388-2284388 on Build GRCh38
rs374901788 Chr.16: 2284241-2284241 on Build GRCh38
rs781697167 Chr.16: 2284310-2284310 on Build GRCh38
rs151110890 Chr.16: 2284326-2284326 on Build GRCh38
rs369254373 Chr.16: 2284255-2284255 on Build GRCh38
rs757955175 Chr.16: 2284442-2284442 on Build GRCh38
rs778589372 Chr.16: 2284265-2284265 on Build GRCh38
rs1054604354 Chr.16: 2284592-2284592 on Build GRCh38
rs1014144312 Chr.16: 2284647-2284647 on Build GRCh38
rs752013927 Chr.16: 2284251-2284251 on Build GRCh38
rs756174838 Chr.16: 2284211-2284211 on Build GRCh38
rs911815157 Chr.16: 2284246-2284246 on Build GRCh38
rs748112009 Chr.16: 2284466-2284466 on Build GRCh38
rs754851837 Chr.16: 2284460-2284460 on Build GRCh38
rs116919064 Chr.16: 2284653-2284653 on Build GRCh38
rs912350290 Chr.16: 2284565-2284565 on Build GRCh38
rs768520218 Chr.16: 2284281-2284281 on Build GRCh38
rs943919554 Chr.16: 2284360-2284360 on Build GRCh38
rs199720998 Chr.16: 2284261-2284261 on Build GRCh38
rs778393424 Chr.16: 2284345-2284345 on Build GRCh38
rs1026611412 Chr.16: 2284532-2284532 on Build GRCh38
rs771040655 Chr.16: 2284475-2284475 on Build GRCh38
rs577525334 Chr.16: 2284599-2284599 on Build GRCh38
rs775888441 Chr.16: 2284484-2284484 on Build GRCh38
rs779988290 Chr.16: 2284229-2284229 on Build GRCh38
rs528994130 Chr.16: 2284283-2284283 on Build GRCh38
rs551756639 Chr.16: 2284363-2284363 on Build GRCh38
rs746859560 Chr.16: 2284331-2284331 on Build GRCh38
rs1043757820 Chr.16: 2284441-2284441 on Build GRCh38
rs141745439 Chr.16: 2284300-2284300 on Build GRCh38
rs761164469 Chr.16: 2284487-2284487 on Build GRCh38
rs536393732 Chr.16: 2284530-2284530 on Build GRCh38
rs969680834 Chr.16: 2284507-2284507 on Build GRCh38
rs779707514 Chr.16: 2284330-2284330 on Build GRCh38
rs778466596 Chr.16: 2284327-2284327 on Build GRCh38
rs991911318 Chr.16: 2284541-2284541 on Build GRCh38
rs749428655 Chr.16: 2284473-2284473 on Build GRCh38
rs903848866 Chr.16: 2284616-2284616 on Build GRCh38
rs186401775 Chr.16: 2284399-2284399 on Build GRCh38
rs539844017 Chr.16: 2284262-2284262 on Build GRCh38
rs982487535 Chr.16: 2284559-2284559 on Build GRCh38
rs1010761307 Chr.16: 2284598-2284598 on Build GRCh38
rs138817727 Chr.16: 2284400-2284400 on Build GRCh38
rs767967118 Chr.16: 2284387-2284387 on Build GRCh38
rs1024296823 Chr.16: 2284497-2284497 on Build GRCh38
rs567752838 Chr.16: 2284519-2284519 on Build GRCh38
rs940407941 Chr.16: 2284515-2284515 on Build GRCh38
rs143095423 Chr.16: 2284575-2284575 on Build GRCh38
rs762120728 Chr.16: 2284237-2284237 on Build GRCh38
rs761746210 Chr.16: 2284257-2284258 on Build GRCh38
rs765595710 Chr.16: 2284238-2284238 on Build GRCh38
rs935360352 Chr.16: 2284629-2284629 on Build GRCh38
rs374326090 Chr.16: 2284455-2284455 on Build GRCh38
rs781209534 Chr.16: 2284465-2284465 on Build GRCh38
rs931960394 Chr.16: 2284408-2284408 on Build GRCh38
rs377568144 Chr.16: 2284304-2284304 on Build GRCh38
rs1052501164 Chr.16: 2284630-2284630 on Build GRCh38
rs71386664 Chr.16: 2284244-2284244 on Build GRCh38
rs1043706336 Chr.16: 2284604-2284604 on Build GRCh38
rs966053727 Chr.16: 2284456-2284456 on Build GRCh38
rs767879016 Chr.16: 2284479-2284479 on Build GRCh38
rs201342214 Chr.16: 2284451-2284451 on Build GRCh38
rs974124577 Chr.16: 2284516-2284516 on Build GRCh38
rs907651114 Chr.16: 2284509-2284509 on Build GRCh38
rs549548321 Chr.16: 2284309-2284309 on Build GRCh38
rs1031126877 Chr.16: 2284464-2284464 on Build GRCh38
rs753327062 Chr.16: 2284319-2284319 on Build GRCh38
rs921149699 Chr.16: 2284594-2284594 on Build GRCh38
rs35089233 Chr.16: 2284357-2284357 on Build GRCh38
rs768692618 Chr.16: 2284332-2284332 on Build GRCh38
rs547822173 Chr.16: 2284512-2284512 on Build GRCh38
rs759998259 Chr.16: 2284374-2284374 on Build GRCh38
rs575310415 Chr.16: 2284574-2284574 on Build GRCh38
rs763073484 Chr.16: 2284356-2284356 on Build GRCh38
rs753203795 Chr.16: 2284394-2284394 on Build GRCh38
rs564420619 Chr.16: 2284257-2284257 on Build GRCh38
rs754028122 Chr.16: 2284230-2284230 on Build GRCh38
rs377661972 Chr.16: 2284477-2284477 on Build GRCh38
rs1014947786 Chr.16: 2284320-2284320 on Build GRCh38

View Full Product Details