Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2437982-2438443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNF
Entrez Gene ID
899
Gene Name
cyclin F
Gene Aliases
FBX1, FBXO1
UniGene
Hs.1973
Gene Chromosome Location
Chr.16: 2425438-2458858 on Build GRCh38
Transcripts
NM_001323538.1 NM_001761.2 XM_017023822.1 XM_017023823.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6623007 c.571G>T p.G191C Chr.16: 2438100-2438100 on Build GRCh38
7157942 c.582G>C p.L194L Chr.16: 2438111-2438111 on Build GRCh38
107490 c.594+2T>G p.? Chr.16: 2438125-2438125 on Build GRCh38
4796775 c.594+1G>A p.? Chr.16: 2438124-2438124 on Build GRCh38
7177628 c.591C>G p.F197L Chr.16: 2438120-2438120 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146626160 Chr.16: 2438319-2438319 on Build GRCh38
rs757135222 Chr.16: 2438034-2438034 on Build GRCh38
rs375261111 Chr.16: 2438176-2438176 on Build GRCh38
rs1055788400 Chr.16: 2438059-2438059 on Build GRCh38
rs913733855 Chr.16: 2438211-2438211 on Build GRCh38
rs778360498 Chr.16: 2438077-2438077 on Build GRCh38
rs374324488 Chr.16: 2438144-2438144 on Build GRCh38
rs776957393 Chr.16: 2438091-2438091 on Build GRCh38
rs1052001047 Chr.16: 2438332-2438332 on Build GRCh38
rs772080724 Chr.16: 2438075-2438075 on Build GRCh38
rs753555251 Chr.16: 2438027-2438027 on Build GRCh38
rs12926008 Chr.16: 2438210-2438210 on Build GRCh38
rs534465527 Chr.16: 2438318-2438318 on Build GRCh38
rs375501461 Chr.16: 2438300-2438300 on Build GRCh38
rs749297488 Chr.16: 2438189-2438189 on Build GRCh38
rs957867337 Chr.16: 2438201-2438201 on Build GRCh38
rs928676876 Chr.16: 2438298-2438298 on Build GRCh38
rs182386024 Chr.16: 2438382-2438382 on Build GRCh38
rs973921504 Chr.16: 2438415-2438415 on Build GRCh38
rs996873785 Chr.16: 2438357-2438357 on Build GRCh38
rs929574145 Chr.16: 2438325-2438325 on Build GRCh38
rs371389975 Chr.16: 2438162-2438162 on Build GRCh38
rs778712309 Chr.16: 2438035-2438035 on Build GRCh38
rs1015373813 Chr.16: 2438394-2438394 on Build GRCh38
rs961186379 Chr.16: 2438413-2438413 on Build GRCh38
rs928650420 Chr.16: 2438274-2438274 on Build GRCh38
rs200106833 Chr.16: 2438143-2438143 on Build GRCh38
rs367823441 Chr.16: 2438161-2438161 on Build GRCh38
rs763581056 Chr.16: 2438136-2438136 on Build GRCh38
rs200658437 Chr.16: 2438021-2438021 on Build GRCh38
rs898030827 Chr.16: 2438135-2438135 on Build GRCh38
rs890142181 Chr.16: 2438334-2438334 on Build GRCh38
rs536237460 Chr.16: 2438403-2438403 on Build GRCh38
rs775708444 Chr.16: 2438384-2438384 on Build GRCh38
rs745821656 Chr.16: 2438071-2438071 on Build GRCh38
rs755865006 Chr.16: 2438020-2438020 on Build GRCh38
rs762067213 Chr.16: 2438120-2438120 on Build GRCh38
rs1042150895 Chr.16: 2438015-2438015 on Build GRCh38
rs771290658 Chr.16: 2438299-2438299 on Build GRCh38
rs544042257 Chr.16: 2438371-2438371 on Build GRCh38
rs897983645 Chr.16: 2438323-2438323 on Build GRCh38
rs578112211 Chr.16: 2438359-2438359 on Build GRCh38
rs538085033 Chr.16: 2438205-2438205 on Build GRCh38
rs767098403 Chr.16: 2438141-2438141 on Build GRCh38
rs777158941 Chr.16: 2438340-2438340 on Build GRCh38
rs768832394 Chr.16: 2438086-2438086 on Build GRCh38
rs938661467 Chr.16: 2438293-2438293 on Build GRCh38
rs982037727 Chr.16: 2438249-2438249 on Build GRCh38
rs1028465068 Chr.16: 2438015-2438016 on Build GRCh38
rs563915133 Chr.16: 2438375-2438375 on Build GRCh38
rs750278031 Chr.16: 2438170-2438170 on Build GRCh38
rs996537554 Chr.16: 2438005-2438005 on Build GRCh38
rs370903607 Chr.16: 2438134-2438134 on Build GRCh38
rs142675674 Chr.16: 2438125-2438125 on Build GRCh38
rs556264911 Chr.16: 2438414-2438414 on Build GRCh38
rs747480031 Chr.16: 2438236-2438236 on Build GRCh38
rs893600671 Chr.16: 2438379-2438379 on Build GRCh38
rs770256445 Chr.16: 2438133-2438133 on Build GRCh38
rs571862675 Chr.16: 2438281-2438281 on Build GRCh38
rs973967083 Chr.16: 2438182-2438182 on Build GRCh38
rs191041595 Chr.16: 2438155-2438155 on Build GRCh38
rs776106564 Chr.16: 2438177-2438178 on Build GRCh38

View Full Product Details