Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2463927-2464423 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf59
Entrez Gene ID
80178
Gene Name
chromosome 16 open reading frame 59
Gene Aliases
-
UniGene
Hs.534491
Gene Chromosome Location
Chr.16: 2460111-2464963 on Build GRCh38
Transcripts
NM_025108.2 XM_011522667.1 NR_106826.1

Gene Symbol
MIR6768
Entrez Gene ID
102465460
Gene Name
microRNA 6768
Gene Aliases
hsa-mir-6768
UniGene
Gene Chromosome Location
Chr.16: 2463967-2464038 on Build GRCh38
Transcripts
NM_025108.2 XM_011522667.1 NR_106826.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3508060 c.1084C>T p.Q362* Chr.16: 2464158-2464158 on Build GRCh38
4666988 c.972G>A p.A324A Chr.16: 2464046-2464046 on Build GRCh38
3957380 c.1118G>A p.R373Q Chr.16: 2464192-2464192 on Build GRCh38
3508059 c.1011C>T p.P337P Chr.16: 2464085-2464085 on Build GRCh38
6390421 c.1034G>A p.G345D Chr.16: 2464108-2464108 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779877147 Chr.16: 2464219-2464219 on Build GRCh38
rs769550084 Chr.16: 2464381-2464381 on Build GRCh38
rs920963889 Chr.16: 2464297-2464297 on Build GRCh38
rs757077282 Chr.16: 2464142-2464142 on Build GRCh38
rs775186499 Chr.16: 2463952-2463952 on Build GRCh38
rs770921850 Chr.16: 2464034-2464034 on Build GRCh38
rs776221543 Chr.16: 2463964-2463964 on Build GRCh38
rs775488831 Chr.16: 2464101-2464101 on Build GRCh38
rs199781969 Chr.16: 2464208-2464208 on Build GRCh38
rs760852706 Chr.16: 2464040-2464040 on Build GRCh38
rs766801800 Chr.16: 2464191-2464191 on Build GRCh38
rs199551012 Chr.16: 2464201-2464201 on Build GRCh38
rs985663295 Chr.16: 2464287-2464287 on Build GRCh38
rs944347968 Chr.16: 2464164-2464164 on Build GRCh38
rs540503679 Chr.16: 2464145-2464145 on Build GRCh38
rs375499936 Chr.16: 2464013-2464013 on Build GRCh38
rs373663102 Chr.16: 2464187-2464187 on Build GRCh38
rs749037338 Chr.16: 2464138-2464138 on Build GRCh38
rs967198374 Chr.16: 2464104-2464104 on Build GRCh38
rs773041368 Chr.16: 2464030-2464030 on Build GRCh38
rs918931937 Chr.16: 2464348-2464349 on Build GRCh38
rs777420161 Chr.16: 2464136-2464136 on Build GRCh38
rs777024227 Chr.16: 2464172-2464172 on Build GRCh38
rs945556740 Chr.16: 2463950-2463950 on Build GRCh38
rs541593454 Chr.16: 2464039-2464039 on Build GRCh38
rs553048593 Chr.16: 2463999-2463999 on Build GRCh38
rs774756619 Chr.16: 2464270-2464270 on Build GRCh38
rs200370097 Chr.16: 2464094-2464094 on Build GRCh38
rs780606469 Chr.16: 2464076-2464076 on Build GRCh38
rs777541604 Chr.16: 2464085-2464085 on Build GRCh38
rs757553652 Chr.16: 2464261-2464261 on Build GRCh38
rs770210042 Chr.16: 2464263-2464263 on Build GRCh38
rs768950227 Chr.16: 2464158-2464158 on Build GRCh38
rs147924387 Chr.16: 2464093-2464093 on Build GRCh38
rs751696933 Chr.16: 2464000-2464000 on Build GRCh38
rs774351716 Chr.16: 2464036-2464036 on Build GRCh38
rs919992826 Chr.16: 2464242-2464242 on Build GRCh38
rs200960307 Chr.16: 2464080-2464080 on Build GRCh38
rs539770191 Chr.16: 2464284-2464284 on Build GRCh38
rs200856167 Chr.16: 2464178-2464178 on Build GRCh38
rs566488415 Chr.16: 2464313-2464313 on Build GRCh38
rs765034153 Chr.16: 2463986-2463986 on Build GRCh38
rs759629651 Chr.16: 2464037-2464037 on Build GRCh38
rs757893316 Chr.16: 2464017-2464017 on Build GRCh38
rs180697062 Chr.16: 2464151-2464151 on Build GRCh38
rs761385636 Chr.16: 2464279-2464279 on Build GRCh38
rs895632794 Chr.16: 2464354-2464354 on Build GRCh38
rs375357120 Chr.16: 2464115-2464115 on Build GRCh38
rs746522005 Chr.16: 2464024-2464024 on Build GRCh38
rs549233198 Chr.16: 2464184-2464184 on Build GRCh38
rs370948477 Chr.16: 2464119-2464119 on Build GRCh38
rs754384928 Chr.16: 2464079-2464080 on Build GRCh38
rs745651098 Chr.16: 2464213-2464213 on Build GRCh38
rs899587432 Chr.16: 2464150-2464150 on Build GRCh38
rs756958614 Chr.16: 2464205-2464205 on Build GRCh38
rs762290516 Chr.16: 2464109-2464109 on Build GRCh38
rs769601549 Chr.16: 2464028-2464028 on Build GRCh38
rs111815304 Chr.16: 2464324-2464324 on Build GRCh38
rs912784904 Chr.16: 2464112-2464112 on Build GRCh38
rs1011321949 Chr.16: 2464161-2464161 on Build GRCh38
rs572852254 Chr.16: 2464027-2464027 on Build GRCh38
rs887598950 Chr.16: 2464197-2464197 on Build GRCh38
rs1023284501 Chr.16: 2464379-2464379 on Build GRCh38
rs34948268 Chr.16: 2464111-2464111 on Build GRCh38
rs1012873485 Chr.16: 2464367-2464367 on Build GRCh38
rs912786275 Chr.16: 2464318-2464318 on Build GRCh38
rs748369654 Chr.16: 2464241-2464241 on Build GRCh38
rs150487803 Chr.16: 2464114-2464114 on Build GRCh38
rs755605092 Chr.16: 2464194-2464194 on Build GRCh38
rs372852875 Chr.16: 2464026-2464026 on Build GRCh38
rs369632827 Chr.16: 2464280-2464280 on Build GRCh38
rs768134371 Chr.16: 2464025-2464025 on Build GRCh38
rs772167368 Chr.16: 2464098-2464098 on Build GRCh38
rs761680512 Chr.16: 2463971-2463971 on Build GRCh38
rs755022463 Chr.16: 2464312-2464312 on Build GRCh38
rs747069248 Chr.16: 2464156-2464156 on Build GRCh38
rs767787914 Chr.16: 2464009-2464009 on Build GRCh38
rs768486599 Chr.16: 2463960-2463960 on Build GRCh38
rs112773540 Chr.16: 2464363-2464363 on Build GRCh38
rs765797107 Chr.16: 2464110-2464110 on Build GRCh38
rs969466004 Chr.16: 2464096-2464096 on Build GRCh38
rs993673287 Chr.16: 2464179-2464179 on Build GRCh38
rs779576847 Chr.16: 2464053-2464053 on Build GRCh38
rs929015920 Chr.16: 2464356-2464356 on Build GRCh38
rs763575316 Chr.16: 2464273-2464273 on Build GRCh38
rs762865317 Chr.16: 2464033-2464033 on Build GRCh38
rs775568437 Chr.16: 2464154-2464154 on Build GRCh38
rs905220304 Chr.16: 2464392-2464392 on Build GRCh38
rs764502281 Chr.16: 2464043-2464043 on Build GRCh38
rs773560313 Chr.16: 2464268-2464268 on Build GRCh38
rs564633721 Chr.16: 2464105-2464105 on Build GRCh38
rs149139826 Chr.16: 2464285-2464285 on Build GRCh38
rs768823594 Chr.16: 2464221-2464221 on Build GRCh38
rs764463737 Chr.16: 2464080-2464080 on Build GRCh38
rs374013660 Chr.16: 2464149-2464149 on Build GRCh38
rs770801282 Chr.16: 2464086-2464086 on Build GRCh38
rs62621760 Chr.16: 2464269-2464269 on Build GRCh38
rs755726556 Chr.16: 2464131-2464131 on Build GRCh38
rs960618535 Chr.16: 2464225-2464225 on Build GRCh38
rs377495646 Chr.16: 2464045-2464045 on Build GRCh38
rs759750271 Chr.16: 2464006-2464006 on Build GRCh38
rs570654235 Chr.16: 2464274-2464274 on Build GRCh38
rs371052068 Chr.16: 2464097-2464097 on Build GRCh38
rs11639943 Chr.16: 2464177-2464177 on Build GRCh38
rs745801135 Chr.16: 2464147-2464147 on Build GRCh38
rs1039331471 Chr.16: 2464129-2464129 on Build GRCh38
rs902919576 Chr.16: 2464322-2464322 on Build GRCh38
rs758294844 Chr.16: 2464216-2464216 on Build GRCh38
rs752195413 Chr.16: 2464192-2464192 on Build GRCh38
rs770083401 Chr.16: 2464320-2464320 on Build GRCh38
rs149358021 Chr.16: 2464199-2464200 on Build GRCh38
rs779939753 Chr.16: 2464309-2464309 on Build GRCh38
rs760370968 Chr.16: 2464338-2464338 on Build GRCh38
rs771675401 Chr.16: 2463951-2463951 on Build GRCh38
rs777077298 Chr.16: 2464107-2464107 on Build GRCh38
rs544744371 Chr.16: 2464102-2464102 on Build GRCh38
rs762920414 Chr.16: 2463993-2463993 on Build GRCh38
rs760843395 Chr.16: 2463951-2463952 on Build GRCh38
rs535399853 Chr.16: 2464319-2464319 on Build GRCh38
rs746642759 Chr.16: 2463959-2463959 on Build GRCh38
rs368866253 Chr.16: 2464046-2464046 on Build GRCh38
rs746988943 Chr.16: 2464200-2464200 on Build GRCh38
rs767043538 Chr.16: 2464117-2464117 on Build GRCh38
rs773230889 Chr.16: 2463991-2463991 on Build GRCh38
rs754545922 Chr.16: 2464064-2464064 on Build GRCh38
rs372529217 Chr.16: 2464011-2464011 on Build GRCh38
rs372258787 Chr.16: 2464220-2464220 on Build GRCh38
rs751427717 Chr.16: 2464310-2464310 on Build GRCh38
rs769391483 Chr.16: 2464081-2464081 on Build GRCh38
rs751064259 Chr.16: 2464060-2464060 on Build GRCh38
rs995475528 Chr.16: 2464393-2464393 on Build GRCh38
rs777154758 Chr.16: 2464339-2464339 on Build GRCh38
rs369572179 Chr.16: 2464022-2464022 on Build GRCh38
rs867048850 Chr.16: 2464353-2464353 on Build GRCh38
rs976068073 Chr.16: 2464291-2464291 on Build GRCh38
rs776899205 Chr.16: 2464229-2464229 on Build GRCh38
rs760079117 Chr.16: 2464272-2464272 on Build GRCh38
rs754371523 Chr.16: 2464016-2464016 on Build GRCh38

View Full Product Details