Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2519129-2519618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0C
Entrez Gene ID
527
Gene Name
ATPase H+ transporting V0 subunit c
Gene Aliases
ATP6C, ATP6L, ATPL, VATL, VPPC, Vma3
UniGene
Hs.389107
Gene Chromosome Location
Chr.16: 2513726-2520223 on Build GRCh38
Transcripts
NM_001198569.1 NM_001694.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5748501 c.203A>G p.Y68C Chr.16: 2519341-2519341 on Build GRCh38
5701492 c.316_317insGCCGGCTTT p.A106>GRLS Chr.16: 2519593-2519594 on Build GRCh38
5620338 c.197C>T p.A66V Chr.16: 2519335-2519335 on Build GRCh38
557336 c.148G>A p.E50K Chr.16: 2519286-2519286 on Build GRCh38
4627388 c.100A>G p.T34A Chr.16: 2519238-2519238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764908287 Chr.16: 2519566-2519566 on Build GRCh38
rs781117999 Chr.16: 2519589-2519589 on Build GRCh38
rs1018587043 Chr.16: 2519453-2519453 on Build GRCh38
rs999717849 Chr.16: 2519151-2519151 on Build GRCh38
rs199740723 Chr.16: 2519492-2519492 on Build GRCh38
rs977944190 Chr.16: 2519485-2519485 on Build GRCh38
rs1008570191 Chr.16: 2519440-2519440 on Build GRCh38
rs769866122 Chr.16: 2519526-2519526 on Build GRCh38
rs556217954 Chr.16: 2519559-2519559 on Build GRCh38
rs763478288 Chr.16: 2519502-2519502 on Build GRCh38
rs139354370 Chr.16: 2519249-2519249 on Build GRCh38
rs762019874 Chr.16: 2519204-2519204 on Build GRCh38
rs894023005 Chr.16: 2519481-2519481 on Build GRCh38
rs745772863 Chr.16: 2519294-2519294 on Build GRCh38
rs774705512 Chr.16: 2519496-2519496 on Build GRCh38
rs914979555 Chr.16: 2519478-2519478 on Build GRCh38
rs375160701 Chr.16: 2519342-2519342 on Build GRCh38
rs766845800 Chr.16: 2519450-2519450 on Build GRCh38
rs780072027 Chr.16: 2519443-2519443 on Build GRCh38
rs145396449 Chr.16: 2519381-2519381 on Build GRCh38
rs139030105 Chr.16: 2519479-2519479 on Build GRCh38
rs1012923215 Chr.16: 2519272-2519272 on Build GRCh38
rs775877366 Chr.16: 2519561-2519561 on Build GRCh38
rs774422091 Chr.16: 2519197-2519197 on Build GRCh38
rs369403915 Chr.16: 2519450-2519450 on Build GRCh38
rs370448357 Chr.16: 2519408-2519408 on Build GRCh38
rs371853850 Chr.16: 2519264-2519264 on Build GRCh38
rs768787745 Chr.16: 2519333-2519333 on Build GRCh38
rs745611504 Chr.16: 2519432-2519432 on Build GRCh38
rs753407052 Chr.16: 2519427-2519427 on Build GRCh38
rs773434350 Chr.16: 2519451-2519451 on Build GRCh38
rs946673994 Chr.16: 2519157-2519157 on Build GRCh38
rs760843165 Chr.16: 2519202-2519202 on Build GRCh38
rs201431472 Chr.16: 2519580-2519580 on Build GRCh38
rs1044901385 Chr.16: 2519586-2519586 on Build GRCh38
rs769289165 Chr.16: 2519181-2519181 on Build GRCh38
rs761344058 Chr.16: 2519562-2519562 on Build GRCh38
rs150070629 Chr.16: 2519285-2519285 on Build GRCh38
rs776821753 Chr.16: 2519447-2519447 on Build GRCh38
rs369475799 Chr.16: 2519366-2519366 on Build GRCh38
rs756977675 Chr.16: 2519429-2519429 on Build GRCh38
rs746899524 Chr.16: 2519520-2519520 on Build GRCh38
rs748200806 Chr.16: 2519523-2519523 on Build GRCh38
rs774746256 Chr.16: 2519452-2519453 on Build GRCh38
rs749034390 Chr.16: 2519276-2519276 on Build GRCh38
rs768053545 Chr.16: 2519165-2519165 on Build GRCh38
rs755572416 Chr.16: 2519423-2519423 on Build GRCh38
rs775593639 Chr.16: 2519303-2519303 on Build GRCh38
rs749567124 Chr.16: 2519532-2519532 on Build GRCh38
rs765565281 Chr.16: 2519371-2519371 on Build GRCh38
rs780055661 Chr.16: 2519519-2519519 on Build GRCh38
rs770613842 Chr.16: 2519280-2519280 on Build GRCh38
rs758265268 Chr.16: 2519518-2519518 on Build GRCh38
rs941952776 Chr.16: 2519331-2519331 on Build GRCh38
rs760742093 Chr.16: 2519268-2519268 on Build GRCh38
rs568971056 Chr.16: 2519154-2519154 on Build GRCh38
rs750917212 Chr.16: 2519225-2519225 on Build GRCh38
rs766645348 Chr.16: 2519363-2519363 on Build GRCh38
rs8063977 Chr.16: 2519456-2519456 on Build GRCh38
rs142995519 Chr.16: 2519200-2519200 on Build GRCh38
rs538440664 Chr.16: 2519444-2519444 on Build GRCh38
rs537621593 Chr.16: 2519195-2519195 on Build GRCh38
rs771258996 Chr.16: 2519533-2519533 on Build GRCh38
rs752697033 Chr.16: 2519595-2519595 on Build GRCh38
rs773480602 Chr.16: 2519220-2519220 on Build GRCh38
rs536381963 Chr.16: 2519534-2519534 on Build GRCh38
rs758283897 Chr.16: 2519439-2519439 on Build GRCh38
rs750107413 Chr.16: 2519514-2519514 on Build GRCh38
rs757038421 Chr.16: 2519585-2519585 on Build GRCh38
rs764928515 Chr.16: 2519507-2519507 on Build GRCh38
rs759853764 Chr.16: 2519553-2519553 on Build GRCh38
rs751318726 Chr.16: 2519583-2519583 on Build GRCh38
rs763328263 Chr.16: 2519393-2519393 on Build GRCh38
rs1027015366 Chr.16: 2519385-2519385 on Build GRCh38
rs760134812 Chr.16: 2519500-2519500 on Build GRCh38
rs766793344 Chr.16: 2519495-2519495 on Build GRCh38
rs747767376 Chr.16: 2519172-2519172 on Build GRCh38
rs766811949 Chr.16: 2519414-2519414 on Build GRCh38
rs376971724 Chr.16: 2519505-2519505 on Build GRCh38
rs915029938 Chr.16: 2519156-2519156 on Build GRCh38
rs781013495 Chr.16: 2519522-2519522 on Build GRCh38
rs758943055 Chr.16: 2519243-2519243 on Build GRCh38
rs773415809 Chr.16: 2519527-2519527 on Build GRCh38
rs753352515 Chr.16: 2519503-2519503 on Build GRCh38
rs754740949 Chr.16: 2519584-2519584 on Build GRCh38
rs755088900 Chr.16: 2519457-2519457 on Build GRCh38
rs772098705 Chr.16: 2519300-2519300 on Build GRCh38
rs549080837 Chr.16: 2519255-2519255 on Build GRCh38
rs534204765 Chr.16: 2519480-2519480 on Build GRCh38
rs748974505 Chr.16: 2519185-2519185 on Build GRCh38
rs750086896 Chr.16: 2519571-2519571 on Build GRCh38
rs752107920 Chr.16: 2519416-2519416 on Build GRCh38
rs765701130 Chr.16: 2519210-2519210 on Build GRCh38
rs1011608789 Chr.16: 2519491-2519492 on Build GRCh38
rs11546346 Chr.16: 2519306-2519306 on Build GRCh38
rs763510837 Chr.16: 2519449-2519450 on Build GRCh38
rs368293163 Chr.16: 2519449-2519449 on Build GRCh38
rs755661331 Chr.16: 2519263-2519263 on Build GRCh38
rs778743976 Chr.16: 2519431-2519431 on Build GRCh38
rs760616550 Chr.16: 2519315-2519315 on Build GRCh38
rs4786290 Chr.16: 2519203-2519203 on Build GRCh38
rs146121658 Chr.16: 2519426-2519426 on Build GRCh38

View Full Product Details