Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2530658-2531150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMDHD2
Entrez Gene ID
51005
Gene Name
amidohydrolase domain containing 2
Gene Aliases
CGI-14
UniGene
Hs.740430
Gene Chromosome Location
Chr.16: 2520351-2531414 on Build GRCh38
Transcripts
NM_001145815.1 XM_017023262.1 XM_017023263.1 XM_017023264.1 XM_017023265.1 XM_017023266.1 XM_017023267.1 NM_001048212.3

Gene Symbol
CEMP1
Entrez Gene ID
752014
Gene Name
cementum protein 1
Gene Aliases
CP-23, CP23
UniGene
Hs.433499
Gene Chromosome Location
Chr.16: 2530035-2531408 on Build GRCh38
Transcripts
NM_001145815.1 XM_017023262.1 XM_017023263.1 XM_017023264.1 XM_017023265.1 XM_017023266.1 XM_017023267.1 NM_001048212.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4059600 c.181A>G p.K61E Chr.16: 2530893-2530893 on Build GRCh38
3794723 c.19G>A p.D7N Chr.16: 2531055-2531055 on Build GRCh38
434982 c.264T>G p.P88P Chr.16: 2530810-2530810 on Build GRCh38
434983 c.242G>C p.R81P Chr.16: 2530832-2530832 on Build GRCh38
4862733 c.1768G>T p.G590W Chr.16: 2530742-2530742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752392343 Chr.16: 2530924-2530924 on Build GRCh38
rs764900825 Chr.16: 2531058-2531058 on Build GRCh38
rs747749604 Chr.16: 2530780-2530781 on Build GRCh38
rs1022928433 Chr.16: 2531005-2531005 on Build GRCh38
rs933853277 Chr.16: 2530921-2530921 on Build GRCh38
rs766873140 Chr.16: 2531032-2531032 on Build GRCh38
rs202039571 Chr.16: 2531008-2531008 on Build GRCh38
rs774286312 Chr.16: 2530943-2530943 on Build GRCh38
rs766257150 Chr.16: 2530883-2530883 on Build GRCh38
rs757714540 Chr.16: 2530901-2530901 on Build GRCh38
rs374505516 Chr.16: 2530934-2530934 on Build GRCh38
rs759261830 Chr.16: 2530969-2530969 on Build GRCh38
rs758022186 Chr.16: 2530790-2530790 on Build GRCh38
rs71386675 Chr.16: 2531080-2531080 on Build GRCh38
rs778787903 Chr.16: 2530930-2530930 on Build GRCh38
rs368755052 Chr.16: 2531093-2531093 on Build GRCh38
rs755435699 Chr.16: 2530691-2530691 on Build GRCh38
rs13331643 Chr.16: 2530911-2530911 on Build GRCh38
rs371750105 Chr.16: 2531126-2531126 on Build GRCh38
rs767272799 Chr.16: 2530903-2530903 on Build GRCh38
rs893784153 Chr.16: 2530808-2530808 on Build GRCh38
rs765539558 Chr.16: 2531117-2531118 on Build GRCh38
rs370312887 Chr.16: 2530796-2530796 on Build GRCh38
rs779553383 Chr.16: 2530793-2530793 on Build GRCh38
rs751844810 Chr.16: 2530753-2530753 on Build GRCh38
rs200977329 Chr.16: 2530832-2530832 on Build GRCh38
rs934812625 Chr.16: 2531058-2531059 on Build GRCh38
rs962656802 Chr.16: 2530985-2530985 on Build GRCh38
rs200546798 Chr.16: 2530684-2530684 on Build GRCh38
rs757728422 Chr.16: 2530851-2530851 on Build GRCh38
rs762140660 Chr.16: 2530972-2530972 on Build GRCh38
rs766387177 Chr.16: 2531122-2531122 on Build GRCh38
rs145505172 Chr.16: 2530881-2530881 on Build GRCh38
rs771840862 Chr.16: 2530707-2530707 on Build GRCh38
rs572300294 Chr.16: 2530873-2530873 on Build GRCh38
rs76722821 Chr.16: 2530754-2530754 on Build GRCh38
rs769543901 Chr.16: 2530804-2530815 on Build GRCh38
rs779577686 Chr.16: 2530852-2530852 on Build GRCh38
rs759596504 Chr.16: 2530885-2530885 on Build GRCh38
rs747325493 Chr.16: 2530742-2530770 on Build GRCh38
rs745368534 Chr.16: 2530776-2530776 on Build GRCh38
rs745387136 Chr.16: 2530703-2530703 on Build GRCh38
rs200972319 Chr.16: 2530882-2530882 on Build GRCh38
rs200449794 Chr.16: 2530992-2530992 on Build GRCh38
rs749714051 Chr.16: 2531087-2531087 on Build GRCh38
rs760358783 Chr.16: 2530727-2530727 on Build GRCh38
rs757402600 Chr.16: 2531104-2531138 on Build GRCh38
rs762205270 Chr.16: 2531020-2531020 on Build GRCh38
rs780217414 Chr.16: 2531009-2531009 on Build GRCh38
rs774882607 Chr.16: 2531089-2531089 on Build GRCh38
rs781515519 Chr.16: 2530693-2530693 on Build GRCh38
rs941126343 Chr.16: 2530751-2530751 on Build GRCh38
rs753486876 Chr.16: 2531045-2531045 on Build GRCh38
rs770459186 Chr.16: 2530699-2530699 on Build GRCh38
rs758547354 Chr.16: 2530685-2530685 on Build GRCh38
rs150766477 Chr.16: 2530750-2530750 on Build GRCh38
rs909514396 Chr.16: 2530741-2530741 on Build GRCh38
rs377440466 Chr.16: 2530959-2530959 on Build GRCh38
rs924783241 Chr.16: 2530826-2530826 on Build GRCh38
rs538285962 Chr.16: 2531048-2531048 on Build GRCh38
rs772671578 Chr.16: 2530713-2530713 on Build GRCh38
rs776279453 Chr.16: 2530763-2530774 on Build GRCh38
rs551843908 Chr.16: 2531106-2531106 on Build GRCh38
rs1011005781 Chr.16: 2530810-2530810 on Build GRCh38
rs368671860 Chr.16: 2530929-2530929 on Build GRCh38
rs370865287 Chr.16: 2530980-2530980 on Build GRCh38
rs757046078 Chr.16: 2530997-2530997 on Build GRCh38
rs756616489 Chr.16: 2530836-2530836 on Build GRCh38
rs752598306 Chr.16: 2531101-2531102 on Build GRCh38
rs778486371 Chr.16: 2530771-2530771 on Build GRCh38
rs986630093 Chr.16: 2530935-2530935 on Build GRCh38
rs773380131 Chr.16: 2531082-2531082 on Build GRCh38
rs866748331 Chr.16: 2530815-2530815 on Build GRCh38
rs778504764 Chr.16: 2530700-2530700 on Build GRCh38
rs780787745 Chr.16: 2530916-2530916 on Build GRCh38
rs200124887 Chr.16: 2530716-2530716 on Build GRCh38
rs865999241 Chr.16: 2531022-2531022 on Build GRCh38
rs780266564 Chr.16: 2530686-2530686 on Build GRCh38
rs751265253 Chr.16: 2530735-2530735 on Build GRCh38
rs546298668 Chr.16: 2531011-2531011 on Build GRCh38
rs754445253 Chr.16: 2530849-2530849 on Build GRCh38
rs750238685 Chr.16: 2531060-2531060 on Build GRCh38
rs775896222 Chr.16: 2530895-2530895 on Build GRCh38
rs547680075 Chr.16: 2531042-2531042 on Build GRCh38
rs769705863 Chr.16: 2530806-2530806 on Build GRCh38
rs765162148 Chr.16: 2530735-2530735 on Build GRCh38
rs199798578 Chr.16: 2531021-2531021 on Build GRCh38
rs201463378 Chr.16: 2531018-2531018 on Build GRCh38
rs748734289 Chr.16: 2530698-2530698 on Build GRCh38
rs766163532 Chr.16: 2531053-2531053 on Build GRCh38
rs139815200 Chr.16: 2531110-2531110 on Build GRCh38
rs372355463 Chr.16: 2530855-2530855 on Build GRCh38
rs535834483 Chr.16: 2530767-2530767 on Build GRCh38
rs574778073 Chr.16: 2530925-2530925 on Build GRCh38
rs768546415 Chr.16: 2530729-2530729 on Build GRCh38
rs565124426 Chr.16: 2531017-2531017 on Build GRCh38
rs754598974 Chr.16: 2530853-2530853 on Build GRCh38
rs773644356 Chr.16: 2531029-2531029 on Build GRCh38
rs766526734 Chr.16: 2530827-2530827 on Build GRCh38
rs778888632 Chr.16: 2531000-2531000 on Build GRCh38
rs780196931 Chr.16: 2531074-2531074 on Build GRCh38
rs774314270 Chr.16: 2530831-2530831 on Build GRCh38
rs777077113 Chr.16: 2530961-2530961 on Build GRCh38
rs761098943 Chr.16: 2530896-2530896 on Build GRCh38
rs774433891 Chr.16: 2530884-2530884 on Build GRCh38
rs770781256 Chr.16: 2530875-2530875 on Build GRCh38
rs146509706 Chr.16: 2531088-2531088 on Build GRCh38
rs368408361 Chr.16: 2530843-2530843 on Build GRCh38
rs772574715 Chr.16: 2530851-2530853 on Build GRCh38
rs75801560 Chr.16: 2531079-2531079 on Build GRCh38
rs765769029 Chr.16: 2530977-2530977 on Build GRCh38
rs1054748014 Chr.16: 2531004-2531004 on Build GRCh38
rs768222357 Chr.16: 2530797-2530797 on Build GRCh38
rs563820819 Chr.16: 2530994-2530994 on Build GRCh38
rs368199152 Chr.16: 2531121-2531121 on Build GRCh38
rs990123562 Chr.16: 2531127-2531127 on Build GRCh38
rs551569221 Chr.16: 2530913-2530913 on Build GRCh38
rs767649302 Chr.16: 2530833-2530833 on Build GRCh38
rs373411042 Chr.16: 2530902-2530902 on Build GRCh38
rs766503541 Chr.16: 2530752-2530752 on Build GRCh38
rs762985689 Chr.16: 2530825-2530825 on Build GRCh38
rs762481591 Chr.16: 2530857-2530858 on Build GRCh38
rs751853332 Chr.16: 2530687-2530687 on Build GRCh38
rs777500987 Chr.16: 2530927-2530927 on Build GRCh38
rs534857231 Chr.16: 2531111-2531111 on Build GRCh38
rs773196779 Chr.16: 2530813-2530813 on Build GRCh38
rs752193917 Chr.16: 2531037-2531037 on Build GRCh38
rs750930349 Chr.16: 2530979-2530979 on Build GRCh38
rs367937724 Chr.16: 2530755-2530755 on Build GRCh38
rs745997670 Chr.16: 2530683-2530684 on Build GRCh38
rs776419367 Chr.16: 2530800-2530800 on Build GRCh38
rs761698203 Chr.16: 2530733-2530733 on Build GRCh38
rs773108906 Chr.16: 2530870-2530870 on Build GRCh38
rs776236799 Chr.16: 2531105-2531105 on Build GRCh38
rs181853336 Chr.16: 2530900-2530900 on Build GRCh38
rs79379654 Chr.16: 2530835-2530835 on Build GRCh38
rs200698850 Chr.16: 2531051-2531051 on Build GRCh38
rs747929730 Chr.16: 2530862-2530862 on Build GRCh38
rs556228583 Chr.16: 2530774-2530774 on Build GRCh38
rs375338784 Chr.16: 2530897-2530897 on Build GRCh38
rs766962908 Chr.16: 2530981-2530981 on Build GRCh38
rs747949867 Chr.16: 2530802-2530802 on Build GRCh38
rs767779561 Chr.16: 2530891-2530891 on Build GRCh38
rs370735504 Chr.16: 2531030-2531030 on Build GRCh38
rs201162958 Chr.16: 2530732-2530732 on Build GRCh38
rs547303573 Chr.16: 2531068-2531068 on Build GRCh38
rs767214081 Chr.16: 2531101-2531101 on Build GRCh38
rs149961868 Chr.16: 2530889-2530889 on Build GRCh38
rs769612287 Chr.16: 2530863-2530863 on Build GRCh38
rs202046694 Chr.16: 2530736-2530736 on Build GRCh38
rs772776692 Chr.16: 2531040-2531040 on Build GRCh38
rs750169543 Chr.16: 2531119-2531119 on Build GRCh38
rs760941091 Chr.16: 2530938-2530938 on Build GRCh38
rs201417710 Chr.16: 2530995-2530995 on Build GRCh38
rs768912903 Chr.16: 2530760-2530760 on Build GRCh38
rs558752304 Chr.16: 2530861-2530861 on Build GRCh38

View Full Product Details