Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2929710-2930217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLYWCH1
Entrez Gene ID
84256
Gene Name
FLYWCH-type zinc finger 1
Gene Aliases
-
UniGene
Hs.655321
Gene Chromosome Location
Chr.16: 2911873-2951208 on Build GRCh38
Transcripts
NM_001308068.1 NM_020912.1 NM_032296.2 XM_006720962.3 XM_011522701.2 XM_011522702.2 XM_011522703.2 XM_011522704.2 XM_011522705.2 XM_011522707.2 XM_017023772.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6555364 c.266G>T p.G89V Chr.16: 2929951-2929951 on Build GRCh38
5460871 c.48C>A p.A16A Chr.16: 2929733-2929733 on Build GRCh38
5806192 c.69G>T p.K23N Chr.16: 2929754-2929754 on Build GRCh38
6665405 c.240G>A p.L80L Chr.16: 2929925-2929925 on Build GRCh38
7368488 c.67A>C p.K23Q Chr.16: 2929752-2929752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767579587 Chr.16: 2929976-2929977 on Build GRCh38
rs748804461 Chr.16: 2929877-2929877 on Build GRCh38
rs976586872 Chr.16: 2929863-2929863 on Build GRCh38
rs565406772 Chr.16: 2929961-2929961 on Build GRCh38
rs112444415 Chr.16: 2929878-2929878 on Build GRCh38
rs377357449 Chr.16: 2929732-2929732 on Build GRCh38
rs200866840 Chr.16: 2929769-2929769 on Build GRCh38
rs772778422 Chr.16: 2929974-2929974 on Build GRCh38
rs766769638 Chr.16: 2929920-2929920 on Build GRCh38
rs534034938 Chr.16: 2930088-2930088 on Build GRCh38
rs371083797 Chr.16: 2929914-2929914 on Build GRCh38
rs774065180 Chr.16: 2929767-2929767 on Build GRCh38
rs373220766 Chr.16: 2929762-2929762 on Build GRCh38
rs747760720 Chr.16: 2929758-2929758 on Build GRCh38
rs374491028 Chr.16: 2929926-2929926 on Build GRCh38
rs751202284 Chr.16: 2930047-2930047 on Build GRCh38
rs376062192 Chr.16: 2929905-2929905 on Build GRCh38
rs199887593 Chr.16: 2929832-2929832 on Build GRCh38
rs764743533 Chr.16: 2930028-2930028 on Build GRCh38
rs571525573 Chr.16: 2929848-2929848 on Build GRCh38
rs748975053 Chr.16: 2929828-2929828 on Build GRCh38
rs773592183 Chr.16: 2929912-2929912 on Build GRCh38
rs970537895 Chr.16: 2929822-2929822 on Build GRCh38
rs754753340 Chr.16: 2930051-2930051 on Build GRCh38
rs756132703 Chr.16: 2929731-2929731 on Build GRCh38
rs766601918 Chr.16: 2929871-2929871 on Build GRCh38
rs756055415 Chr.16: 2930060-2930060 on Build GRCh38
rs759854034 Chr.16: 2930008-2930008 on Build GRCh38
rs889431884 Chr.16: 2930096-2930096 on Build GRCh38
rs1002762075 Chr.16: 2930124-2930124 on Build GRCh38
rs201548170 Chr.16: 2929873-2929873 on Build GRCh38
rs757667264 Chr.16: 2929734-2929734 on Build GRCh38
rs563808709 Chr.16: 2930063-2930063 on Build GRCh38
rs1019510763 Chr.16: 2930112-2930112 on Build GRCh38
rs779614685 Chr.16: 2930040-2930040 on Build GRCh38
rs376488136 Chr.16: 2929763-2929763 on Build GRCh38
rs369521432 Chr.16: 2929780-2929780 on Build GRCh38
rs762024599 Chr.16: 2929910-2929910 on Build GRCh38
rs910565957 Chr.16: 2929901-2929901 on Build GRCh38
rs199909529 Chr.16: 2930035-2930035 on Build GRCh38
rs745495254 Chr.16: 2929886-2929886 on Build GRCh38
rs771888603 Chr.16: 2929887-2929887 on Build GRCh38
rs768466983 Chr.16: 2929979-2929979 on Build GRCh38
rs772650861 Chr.16: 2929735-2929735 on Build GRCh38
rs367577065 Chr.16: 2929821-2929821 on Build GRCh38
rs765622993 Chr.16: 2929855-2929855 on Build GRCh38
rs1015235056 Chr.16: 2930152-2930152 on Build GRCh38
rs752713222 Chr.16: 2929730-2929730 on Build GRCh38
rs778827755 Chr.16: 2929779-2929779 on Build GRCh38
rs931384720 Chr.16: 2929927-2929927 on Build GRCh38
rs758830170 Chr.16: 2929803-2929803 on Build GRCh38
rs567457162 Chr.16: 2930097-2930097 on Build GRCh38
rs368822385 Chr.16: 2929733-2929733 on Build GRCh38
rs758884268 Chr.16: 2929862-2929862 on Build GRCh38
rs183328490 Chr.16: 2929913-2929913 on Build GRCh38
rs779650067 Chr.16: 2929898-2929898 on Build GRCh38
rs370160424 Chr.16: 2929829-2929829 on Build GRCh38
rs922490000 Chr.16: 2930041-2930041 on Build GRCh38
rs937616405 Chr.16: 2929959-2929959 on Build GRCh38
rs1047075248 Chr.16: 2930109-2930109 on Build GRCh38
rs200664932 Chr.16: 2929944-2929944 on Build GRCh38
rs1057291391 Chr.16: 2929970-2929970 on Build GRCh38
rs373096010 Chr.16: 2929904-2929904 on Build GRCh38
rs754123279 Chr.16: 2929786-2929786 on Build GRCh38
rs756807311 Chr.16: 2929951-2929951 on Build GRCh38
rs965212167 Chr.16: 2930129-2930129 on Build GRCh38
rs370886934 Chr.16: 2929774-2929774 on Build GRCh38
rs747452816 Chr.16: 2929815-2929815 on Build GRCh38
rs575622306 Chr.16: 2929812-2929812 on Build GRCh38
rs775543736 Chr.16: 2929833-2929833 on Build GRCh38
rs530271363 Chr.16: 2930027-2930027 on Build GRCh38
rs906678284 Chr.16: 2929978-2929978 on Build GRCh38
rs906153681 Chr.16: 2929751-2929751 on Build GRCh38
rs761971688 Chr.16: 2929853-2929853 on Build GRCh38
rs376471191 Chr.16: 2930190-2930190 on Build GRCh38
rs748843365 Chr.16: 2929766-2929766 on Build GRCh38
rs1020673667 Chr.16: 2929810-2929810 on Build GRCh38
rs146665306 Chr.16: 2930005-2930005 on Build GRCh38
rs750875928 Chr.16: 2929802-2929802 on Build GRCh38
rs117969741 Chr.16: 2929983-2929983 on Build GRCh38
rs777355294 Chr.16: 2929876-2929876 on Build GRCh38
rs753222141 Chr.16: 2930025-2930025 on Build GRCh38
rs780695966 Chr.16: 2929754-2929754 on Build GRCh38
rs371336378 Chr.16: 2930032-2930032 on Build GRCh38
rs984018350 Chr.16: 2929847-2929847 on Build GRCh38
rs375491935 Chr.16: 2929885-2929885 on Build GRCh38
rs778342131 Chr.16: 2929956-2929956 on Build GRCh38
rs541355643 Chr.16: 2930004-2930004 on Build GRCh38
rs781096530 Chr.16: 2930058-2930058 on Build GRCh38
rs116353313 Chr.16: 2930074-2930074 on Build GRCh38
rs910601687 Chr.16: 2930062-2930062 on Build GRCh38
rs779747613 Chr.16: 2929963-2929963 on Build GRCh38
rs532360490 Chr.16: 2930067-2930067 on Build GRCh38
rs571423420 Chr.16: 2930077-2930077 on Build GRCh38
rs906716982 Chr.16: 2930119-2930119 on Build GRCh38
rs776627100 Chr.16: 2929907-2929907 on Build GRCh38
rs1025889602 Chr.16: 2930147-2930147 on Build GRCh38
rs760491940 Chr.16: 2929836-2929836 on Build GRCh38
rs781042164 Chr.16: 2929980-2929980 on Build GRCh38
rs555937349 Chr.16: 2930144-2930144 on Build GRCh38
rs745699739 Chr.16: 2929889-2929889 on Build GRCh38
rs772951532 Chr.16: 2929772-2929772 on Build GRCh38
rs373096823 Chr.16: 2929793-2929793 on Build GRCh38
rs994055505 Chr.16: 2930145-2930145 on Build GRCh38
rs368066668 Chr.16: 2930194-2930194 on Build GRCh38
rs544999887 Chr.16: 2929938-2929938 on Build GRCh38
rs961014015 Chr.16: 2930162-2930162 on Build GRCh38
rs945636995 Chr.16: 2930093-2930093 on Build GRCh38
rs764034113 Chr.16: 2929784-2929784 on Build GRCh38
rs977055314 Chr.16: 2930038-2930038 on Build GRCh38
rs762831104 Chr.16: 2929941-2929943 on Build GRCh38
rs534057517 Chr.16: 2930084-2930084 on Build GRCh38
rs549896842 Chr.16: 2929775-2929775 on Build GRCh38
rs769973889 Chr.16: 2929911-2929911 on Build GRCh38
rs953973030 Chr.16: 2929879-2929879 on Build GRCh38
rs536496583 Chr.16: 2930114-2930114 on Build GRCh38
rs757925057 Chr.16: 2930037-2930037 on Build GRCh38
rs773975965 Chr.16: 2929830-2929830 on Build GRCh38
rs764242615 Chr.16: 2929846-2929846 on Build GRCh38
rs773357182 Chr.16: 2930000-2930000 on Build GRCh38
rs755671309 Chr.16: 2929819-2929819 on Build GRCh38
rs1005255230 Chr.16: 2930151-2930151 on Build GRCh38
rs757932671 Chr.16: 2929962-2929962 on Build GRCh38
rs554030327 Chr.16: 2929875-2929875 on Build GRCh38
rs377001546 Chr.16: 2929999-2929999 on Build GRCh38
rs370723003 Chr.16: 2929997-2929997 on Build GRCh38
rs377382334 Chr.16: 2929795-2929795 on Build GRCh38
rs1046601344 Chr.16: 2929964-2929964 on Build GRCh38
rs953983426 Chr.16: 2930046-2930046 on Build GRCh38
rs1053821317 Chr.16: 2930076-2930076 on Build GRCh38
rs371606043 Chr.16: 2930177-2930177 on Build GRCh38
rs748099860 Chr.16: 2930059-2930059 on Build GRCh38
rs763282065 Chr.16: 2930001-2930001 on Build GRCh38
rs1054888290 Chr.16: 2930125-2930125 on Build GRCh38
rs538035178 Chr.16: 2929858-2929858 on Build GRCh38
rs1001144978 Chr.16: 2929782-2929782 on Build GRCh38
rs746846697 Chr.16: 2929971-2929971 on Build GRCh38

View Full Product Details