Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3012639-3013137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN9
Entrez Gene ID
9080
Gene Name
claudin 9
Gene Aliases
-
UniGene
Hs.296949
Gene Chromosome Location
Chr.16: 3012456-3014505 on Build GRCh38
Transcripts
NM_020982.3

SNP Detail

SNP ID Chromosome Location
rs952914608 Chr.16: 3012763-3012763 on Build GRCh38
rs550317435 Chr.16: 3012709-3012709 on Build GRCh38
rs954823279 Chr.16: 3012857-3012857 on Build GRCh38
rs578145851 Chr.16: 3012925-3012925 on Build GRCh38
rs1029414406 Chr.16: 3012967-3012967 on Build GRCh38
rs904382962 Chr.16: 3012702-3012702 on Build GRCh38
rs925854149 Chr.16: 3012851-3012851 on Build GRCh38
rs895179083 Chr.16: 3012767-3012767 on Build GRCh38
rs973181711 Chr.16: 3012876-3012876 on Build GRCh38
rs976497526 Chr.16: 3012812-3012812 on Build GRCh38
rs970725477 Chr.16: 3012728-3012728 on Build GRCh38
rs1048029138 Chr.16: 3012740-3012741 on Build GRCh38
rs932318897 Chr.16: 3012828-3012828 on Build GRCh38
rs932572287 Chr.16: 3012680-3012680 on Build GRCh38
rs371122842 Chr.16: 3013009-3013009 on Build GRCh38
rs964363050 Chr.16: 3012705-3012705 on Build GRCh38
rs891612027 Chr.16: 3012875-3012875 on Build GRCh38
rs1018415393 Chr.16: 3012945-3012945 on Build GRCh38
rs933214065 Chr.16: 3012723-3012723 on Build GRCh38
rs916680930 Chr.16: 3012882-3012882 on Build GRCh38
rs948097601 Chr.16: 3012887-3012887 on Build GRCh38
rs1005733214 Chr.16: 3012938-3012938 on Build GRCh38
rs1033890644 Chr.16: 3012749-3012749 on Build GRCh38
rs1030451039 Chr.16: 3012968-3012968 on Build GRCh38
rs954383913 Chr.16: 3012992-3012994 on Build GRCh38
rs906779670 Chr.16: 3012794-3012794 on Build GRCh38
rs1015725706 Chr.16: 3012940-3012940 on Build GRCh38
rs907736332 Chr.16: 3012860-3012860 on Build GRCh38
rs535395375 Chr.16: 3012780-3012780 on Build GRCh38
rs1002891854 Chr.16: 3012741-3012741 on Build GRCh38
rs1046368274 Chr.16: 3012786-3012786 on Build GRCh38
rs945466782 Chr.16: 3012902-3012902 on Build GRCh38
rs146688835 Chr.16: 3012710-3012710 on Build GRCh38
rs922288399 Chr.16: 3012818-3012818 on Build GRCh38
rs553059271 Chr.16: 3012739-3012739 on Build GRCh38
rs1002201661 Chr.16: 3012913-3012913 on Build GRCh38
rs1046457010 Chr.16: 3012903-3012903 on Build GRCh38
rs530715174 Chr.16: 3012698-3012698 on Build GRCh38
rs1052843128 Chr.16: 3013117-3013117 on Build GRCh38
rs1037731681 Chr.16: 3012755-3012755 on Build GRCh38
rs900260634 Chr.16: 3012951-3012951 on Build GRCh38
rs906625509 Chr.16: 3013091-3013091 on Build GRCh38
rs1052943613 Chr.16: 3012854-3012854 on Build GRCh38
rs1031619735 Chr.16: 3012811-3012811 on Build GRCh38
rs985822947 Chr.16: 3012842-3012842 on Build GRCh38
rs953880324 Chr.16: 3012974-3012974 on Build GRCh38
rs999746523 Chr.16: 3012810-3012810 on Build GRCh38
rs935921895 Chr.16: 3013010-3013010 on Build GRCh38
rs544962118 Chr.16: 3012908-3012908 on Build GRCh38
rs976934486 Chr.16: 3012962-3012962 on Build GRCh38
rs1005785558 Chr.16: 3013112-3013112 on Build GRCh38
rs1011099315 Chr.16: 3012926-3012926 on Build GRCh38
rs961128766 Chr.16: 3012806-3012806 on Build GRCh38
rs879182100 Chr.16: 3012803-3012803 on Build GRCh38
rs907692703 Chr.16: 3012744-3012744 on Build GRCh38
rs1049655234 Chr.16: 3012691-3012691 on Build GRCh38
rs1038648785 Chr.16: 3012898-3012898 on Build GRCh38
rs765505498 Chr.16: 3013110-3013110 on Build GRCh38
rs777032423 Chr.16: 3012708-3012708 on Build GRCh38
rs989898607 Chr.16: 3012695-3012695 on Build GRCh38
rs916378795 Chr.16: 3013097-3013098 on Build GRCh38
rs767539002 Chr.16: 3012878-3012879 on Build GRCh38
rs949910843 Chr.16: 3013026-3013026 on Build GRCh38
rs1052691013 Chr.16: 3012920-3012920 on Build GRCh38
rs555424519 Chr.16: 3012825-3012825 on Build GRCh38
rs985152966 Chr.16: 3012975-3012975 on Build GRCh38
rs377654796 Chr.16: 3012852-3012852 on Build GRCh38
rs1015778177 Chr.16: 3013115-3013115 on Build GRCh38
rs983801365 Chr.16: 3013006-3013006 on Build GRCh38
rs868387572 Chr.16: 3013050-3013050 on Build GRCh38
rs988699753 Chr.16: 3013013-3013013 on Build GRCh38
rs180735534 Chr.16: 3013103-3013103 on Build GRCh38
rs189534646 Chr.16: 3012921-3012921 on Build GRCh38
rs567988791 Chr.16: 3012670-3012670 on Build GRCh38
rs983384561 Chr.16: 3012858-3012858 on Build GRCh38
rs892562119 Chr.16: 3012711-3012711 on Build GRCh38
rs938054559 Chr.16: 3013093-3013093 on Build GRCh38
rs961552744 Chr.16: 3012950-3012950 on Build GRCh38
rs539155039 Chr.16: 3012724-3012724 on Build GRCh38
rs984260751 Chr.16: 3012784-3012784 on Build GRCh38
rs540358205 Chr.16: 3013017-3013017 on Build GRCh38
rs566327171 Chr.16: 3012740-3012740 on Build GRCh38
rs922266144 Chr.16: 3012679-3012679 on Build GRCh38
rs1025414880 Chr.16: 3012725-3012725 on Build GRCh38
rs1012215078 Chr.16: 3012783-3012783 on Build GRCh38
rs1046949859 Chr.16: 3013035-3013035 on Build GRCh38
rs865935440 Chr.16: 3012931-3012931 on Build GRCh38

View Full Product Details