Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3013027-3013530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN9
Entrez Gene ID
9080
Gene Name
claudin 9
Gene Aliases
-
UniGene
Hs.296949
Gene Chromosome Location
Chr.16: 3012456-3014505 on Build GRCh38
Transcripts
NM_020982.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
703272 c.65T>C p.L22P Chr.16: 3013427-3013427 on Build GRCh38
3112542 c.116G>A p.S39N Chr.16: 3013478-3013478 on Build GRCh38
4060166 c.15C>A p.G5G Chr.16: 3013377-3013377 on Build GRCh38
392220 c.38_39insG p.A14fs*141 Chr.16: 3013400-3013401 on Build GRCh38
358975 c.136G>T p.V46L Chr.16: 3013498-3013498 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750111126 Chr.16: 3013435-3013435 on Build GRCh38
rs749542431 Chr.16: 3013467-3013467 on Build GRCh38
rs1037040024 Chr.16: 3013428-3013428 on Build GRCh38
rs759968771 Chr.16: 3013327-3013327 on Build GRCh38
rs1037281300 Chr.16: 3013257-3013257 on Build GRCh38
rs773818198 Chr.16: 3013453-3013453 on Build GRCh38
rs532499831 Chr.16: 3013155-3013155 on Build GRCh38
rs368045321 Chr.16: 3013437-3013437 on Build GRCh38
rs145882247 Chr.16: 3013483-3013483 on Build GRCh38
rs897357774 Chr.16: 3013121-3013121 on Build GRCh38
rs775828414 Chr.16: 3013413-3013413 on Build GRCh38
rs927094153 Chr.16: 3013192-3013192 on Build GRCh38
rs138697130 Chr.16: 3013506-3013506 on Build GRCh38
rs759442841 Chr.16: 3013499-3013499 on Build GRCh38
rs747724516 Chr.16: 3013374-3013374 on Build GRCh38
rs754640455 Chr.16: 3013307-3013307 on Build GRCh38
rs564102297 Chr.16: 3013248-3013248 on Build GRCh38
rs890172716 Chr.16: 3013211-3013211 on Build GRCh38
rs2227269 Chr.16: 3013263-3013263 on Build GRCh38
rs764635498 Chr.16: 3013431-3013431 on Build GRCh38
rs750310555 Chr.16: 3013220-3013220 on Build GRCh38
rs1007062296 Chr.16: 3013465-3013465 on Build GRCh38
rs906625509 Chr.16: 3013091-3013091 on Build GRCh38
rs900000322 Chr.16: 3013174-3013174 on Build GRCh38
rs772812332 Chr.16: 3013486-3013486 on Build GRCh38
rs60420127 Chr.16: 3013273-3013273 on Build GRCh38
rs769348025 Chr.16: 3013485-3013485 on Build GRCh38
rs750035233 Chr.16: 3013339-3013339 on Build GRCh38
rs994704085 Chr.16: 3013244-3013244 on Build GRCh38
rs958512345 Chr.16: 3013409-3013409 on Build GRCh38
rs532767367 Chr.16: 3013227-3013228 on Build GRCh38
rs928091723 Chr.16: 3013269-3013269 on Build GRCh38
rs758341223 Chr.16: 3013126-3013126 on Build GRCh38
rs764977348 Chr.16: 3013337-3013337 on Build GRCh38
rs552662977 Chr.16: 3013259-3013259 on Build GRCh38
rs761206429 Chr.16: 3013335-3013335 on Build GRCh38
rs971687123 Chr.16: 3013173-3013173 on Build GRCh38
rs916378795 Chr.16: 3013097-3013098 on Build GRCh38
rs749037254 Chr.16: 3013306-3013306 on Build GRCh38
rs374429657 Chr.16: 3013395-3013395 on Build GRCh38
rs200336281 Chr.16: 3013470-3013470 on Build GRCh38
rs754710403 Chr.16: 3013440-3013440 on Build GRCh38
rs374414759 Chr.16: 3013321-3013321 on Build GRCh38
rs779933456 Chr.16: 3013353-3013353 on Build GRCh38
rs778472998 Chr.16: 3013341-3013341 on Build GRCh38
rs145786872 Chr.16: 3013482-3013482 on Build GRCh38
rs868387572 Chr.16: 3013050-3013050 on Build GRCh38
rs140410599 Chr.16: 3013391-3013391 on Build GRCh38
rs180735534 Chr.16: 3013103-3013103 on Build GRCh38
rs767203719 Chr.16: 3013503-3013503 on Build GRCh38
rs767532136 Chr.16: 3013454-3013454 on Build GRCh38
rs749501404 Chr.16: 3013385-3013385 on Build GRCh38
rs938054559 Chr.16: 3013093-3013093 on Build GRCh38
rs778124251 Chr.16: 3013377-3013377 on Build GRCh38
rs1032745333 Chr.16: 3013294-3013294 on Build GRCh38
rs2231607 Chr.16: 3013144-3013144 on Build GRCh38
rs941364995 Chr.16: 3013250-3013250 on Build GRCh38
rs2231608 Chr.16: 3013148-3013148 on Build GRCh38
rs772684061 Chr.16: 3013412-3013412 on Build GRCh38
rs953824965 Chr.16: 3013138-3013138 on Build GRCh38
rs1009913106 Chr.16: 3013319-3013319 on Build GRCh38
rs144730563 Chr.16: 3013370-3013370 on Build GRCh38
rs771283325 Chr.16: 3013313-3013313 on Build GRCh38
rs762719346 Chr.16: 3013488-3013488 on Build GRCh38
rs755940060 Chr.16: 3013510-3013510 on Build GRCh38
rs1055120277 Chr.16: 3013219-3013219 on Build GRCh38
rs199830559 Chr.16: 3013329-3013329 on Build GRCh38
rs550506031 Chr.16: 3013212-3013212 on Build GRCh38
rs757420145 Chr.16: 3013469-3013469 on Build GRCh38
rs751354671 Chr.16: 3013438-3013438 on Build GRCh38
rs114794799 Chr.16: 3013422-3013422 on Build GRCh38
rs529393711 Chr.16: 3013137-3013137 on Build GRCh38
rs538519186 Chr.16: 3013375-3013375 on Build GRCh38
rs779085754 Chr.16: 3013471-3013471 on Build GRCh38
rs751550677 Chr.16: 3013365-3013365 on Build GRCh38
rs996352134 Chr.16: 3013175-3013175 on Build GRCh38
rs752836370 Chr.16: 3013125-3013125 on Build GRCh38
rs772316258 Chr.16: 3013473-3013473 on Build GRCh38
rs763541879 Chr.16: 3013328-3013328 on Build GRCh38
rs1052843128 Chr.16: 3013117-3013117 on Build GRCh38
rs371752851 Chr.16: 3013262-3013262 on Build GRCh38
rs566533435 Chr.16: 3013288-3013288 on Build GRCh38
rs988765815 Chr.16: 3013156-3013156 on Build GRCh38
rs61743246 Chr.16: 3013464-3013464 on Build GRCh38
rs535547771 Chr.16: 3013310-3013310 on Build GRCh38
rs1005785558 Chr.16: 3013112-3013112 on Build GRCh38
rs758324404 Chr.16: 3013352-3013352 on Build GRCh38
rs756277481 Chr.16: 3013462-3013462 on Build GRCh38
rs777041708 Chr.16: 3013356-3013356 on Build GRCh38
rs151195275 Chr.16: 3013371-3013371 on Build GRCh38
rs765505498 Chr.16: 3013110-3013110 on Build GRCh38
rs938120136 Chr.16: 3013210-3013210 on Build GRCh38
rs1039931093 Chr.16: 3013157-3013157 on Build GRCh38
rs1015778177 Chr.16: 3013115-3013115 on Build GRCh38
rs913138545 Chr.16: 3013158-3013158 on Build GRCh38
rs746271447 Chr.16: 3013472-3013472 on Build GRCh38
rs752573201 Chr.16: 3013461-3013461 on Build GRCh38
rs766188648 Chr.16: 3013497-3013497 on Build GRCh38
rs963270400 Chr.16: 3013239-3013239 on Build GRCh38
rs774657538 Chr.16: 3013394-3013394 on Build GRCh38
rs889922850 Chr.16: 3013287-3013287 on Build GRCh38
rs773774141 Chr.16: 3013418-3013418 on Build GRCh38
rs957014964 Chr.16: 3013151-3013151 on Build GRCh38
rs915237113 Chr.16: 3013305-3013305 on Build GRCh38
rs774119241 Chr.16: 3013498-3013498 on Build GRCh38
rs909963370 Chr.16: 3013421-3013421 on Build GRCh38
rs938035574 Chr.16: 3013162-3013162 on Build GRCh38
rs530704025 Chr.16: 3013185-3013185 on Build GRCh38

View Full Product Details