Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3014940-3015439 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN6
Entrez Gene ID
9074
Gene Name
claudin 6
Gene Aliases
-
UniGene
Hs.533779
Gene Chromosome Location
Chr.16: 3014712-3018187 on Build GRCh38
Transcripts
NM_021195.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4672698 c.627G>A p.R209R Chr.16: 3015395-3015395 on Build GRCh38
4060173 c.651G>T p.K217N Chr.16: 3015371-3015371 on Build GRCh38
1201317 c.658G>A p.V220I Chr.16: 3015364-3015364 on Build GRCh38
3817802 c.651G>C p.K217N Chr.16: 3015371-3015371 on Build GRCh38
3370395 c.596G>A p.R199H Chr.16: 3015426-3015426 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748972591 Chr.16: 3015326-3015326 on Build GRCh38
rs752498258 Chr.16: 3014973-3014973 on Build GRCh38
rs761856546 Chr.16: 3015416-3015416 on Build GRCh38
rs1057435585 Chr.16: 3015013-3015013 on Build GRCh38
rs757369834 Chr.16: 3015198-3015198 on Build GRCh38
rs540117566 Chr.16: 3015082-3015082 on Build GRCh38
rs946273629 Chr.16: 3015042-3015042 on Build GRCh38
rs750643600 Chr.16: 3015361-3015361 on Build GRCh38
rs200931668 Chr.16: 3015327-3015327 on Build GRCh38
rs143075646 Chr.16: 3014964-3014965 on Build GRCh38
rs777380085 Chr.16: 3015325-3015325 on Build GRCh38
rs758784600 Chr.16: 3015205-3015205 on Build GRCh38
rs917253166 Chr.16: 3015324-3015324 on Build GRCh38
rs572129261 Chr.16: 3015200-3015200 on Build GRCh38
rs781199940 Chr.16: 3015392-3015393 on Build GRCh38
rs372194239 Chr.16: 3015336-3015336 on Build GRCh38
rs373048665 Chr.16: 3015035-3015035 on Build GRCh38
rs201318784 Chr.16: 3015360-3015360 on Build GRCh38
rs904673832 Chr.16: 3015078-3015078 on Build GRCh38
rs190340097 Chr.16: 3015030-3015030 on Build GRCh38
rs959003062 Chr.16: 3015289-3015289 on Build GRCh38
rs1006562858 Chr.16: 3015094-3015094 on Build GRCh38
rs1053556226 Chr.16: 3015090-3015090 on Build GRCh38
rs184067359 Chr.16: 3015310-3015310 on Build GRCh38
rs149605777 Chr.16: 3015396-3015396 on Build GRCh38
rs138284432 Chr.16: 3015389-3015389 on Build GRCh38
rs545074448 Chr.16: 3015173-3015173 on Build GRCh38
rs994072441 Chr.16: 3015399-3015399 on Build GRCh38
rs764177909 Chr.16: 3015340-3015340 on Build GRCh38
rs918410578 Chr.16: 3014967-3014967 on Build GRCh38
rs927418259 Chr.16: 3015281-3015281 on Build GRCh38
rs745469516 Chr.16: 3015393-3015393 on Build GRCh38
rs760563507 Chr.16: 3015337-3015337 on Build GRCh38
rs532562507 Chr.16: 3015365-3015365 on Build GRCh38
rs1025453189 Chr.16: 3015207-3015207 on Build GRCh38
rs751035245 Chr.16: 3015125-3015125 on Build GRCh38
rs552757538 Chr.16: 3015377-3015377 on Build GRCh38
rs376502802 Chr.16: 3015224-3015224 on Build GRCh38
rs113135801 Chr.16: 3015358-3015358 on Build GRCh38
rs200821404 Chr.16: 3015390-3015390 on Build GRCh38
rs764091813 Chr.16: 3015144-3015144 on Build GRCh38
rs768615835 Chr.16: 3015405-3015405 on Build GRCh38
rs984149421 Chr.16: 3014980-3014980 on Build GRCh38
rs753962881 Chr.16: 3015349-3015349 on Build GRCh38
rs746780451 Chr.16: 3015401-3015401 on Build GRCh38
rs758008110 Chr.16: 3015061-3015061 on Build GRCh38
rs535126754 Chr.16: 3015397-3015397 on Build GRCh38
rs780482113 Chr.16: 3015228-3015228 on Build GRCh38
rs576187833 Chr.16: 3015135-3015135 on Build GRCh38
rs756822589 Chr.16: 3015331-3015331 on Build GRCh38
rs55730488 Chr.16: 3015031-3015031 on Build GRCh38
rs375517434 Chr.16: 3015351-3015351 on Build GRCh38
rs926164295 Chr.16: 3015357-3015357 on Build GRCh38
rs769434896 Chr.16: 3015319-3015319 on Build GRCh38
rs889511808 Chr.16: 3015092-3015092 on Build GRCh38
rs927314748 Chr.16: 3015001-3015001 on Build GRCh38
rs564796765 Chr.16: 3015199-3015199 on Build GRCh38
rs141263027 Chr.16: 3015362-3015362 on Build GRCh38
rs937371839 Chr.16: 3015007-3015007 on Build GRCh38
rs984332782 Chr.16: 3015251-3015251 on Build GRCh38
rs778491532 Chr.16: 3015391-3015391 on Build GRCh38
rs774027751 Chr.16: 3015330-3015330 on Build GRCh38
rs150318164 Chr.16: 3015364-3015364 on Build GRCh38
rs952532337 Chr.16: 3015211-3015211 on Build GRCh38
rs755090347 Chr.16: 3015194-3015195 on Build GRCh38
rs776544375 Chr.16: 3015410-3015410 on Build GRCh38

View Full Product Details