Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3029082-3029620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BICDL2
Entrez Gene ID
146439
Gene Name
BICD family like cargo adaptor 2
Gene Aliases
BICDR-2, BICDR2, CCDC64B
UniGene
Hs.513285
Gene Chromosome Location
Chr.16: 3027710-3036985 on Build GRCh38
Transcripts
NM_001103175.1 XM_005255135.4 XM_011522391.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3361733 c.1094A>T p.K365M Chr.16: 3029293-3029293 on Build GRCh38
970013 c.919G>A p.D307N Chr.16: 3029583-3029583 on Build GRCh38
970012 c.932G>C p.G311A Chr.16: 3029570-3029570 on Build GRCh38
4637344 c.905G>A p.S302N Chr.16: 3029597-3029597 on Build GRCh38
3113209 c.1032delC p.K345fs*67 Chr.16: 3029355-3029355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371973819 Chr.16: 3029169-3029169 on Build GRCh38
rs774472557 Chr.16: 3029330-3029330 on Build GRCh38
rs372183892 Chr.16: 3029422-3029422 on Build GRCh38
rs888288302 Chr.16: 3029111-3029111 on Build GRCh38
rs888339332 Chr.16: 3029492-3029492 on Build GRCh38
rs1026103453 Chr.16: 3029234-3029234 on Build GRCh38
rs561586542 Chr.16: 3029502-3029502 on Build GRCh38
rs550592901 Chr.16: 3029506-3029506 on Build GRCh38
rs897951228 Chr.16: 3029536-3029536 on Build GRCh38
rs533652080 Chr.16: 3029527-3029527 on Build GRCh38
rs779585975 Chr.16: 3029287-3029287 on Build GRCh38
rs374832856 Chr.16: 3029417-3029417 on Build GRCh38
rs750413160 Chr.16: 3029546-3029546 on Build GRCh38
rs545723438 Chr.16: 3029347-3029347 on Build GRCh38
rs766653565 Chr.16: 3029232-3029232 on Build GRCh38
rs756012274 Chr.16: 3029381-3029381 on Build GRCh38
rs1019850671 Chr.16: 3029154-3029154 on Build GRCh38
rs757038997 Chr.16: 3029539-3029539 on Build GRCh38
rs763422024 Chr.16: 3029469-3029470 on Build GRCh38
rs889753567 Chr.16: 3029144-3029144 on Build GRCh38
rs950512040 Chr.16: 3029270-3029270 on Build GRCh38
rs754344293 Chr.16: 3029363-3029363 on Build GRCh38
rs749239864 Chr.16: 3029391-3029391 on Build GRCh38
rs749215024 Chr.16: 3029328-3029328 on Build GRCh38
rs1005770207 Chr.16: 3029151-3029151 on Build GRCh38
rs530662460 Chr.16: 3029503-3029503 on Build GRCh38
rs753599003 Chr.16: 3029495-3029495 on Build GRCh38
rs372064266 Chr.16: 3029405-3029405 on Build GRCh38
rs201951347 Chr.16: 3029379-3029379 on Build GRCh38
rs770962990 Chr.16: 3029329-3029329 on Build GRCh38
rs760858298 Chr.16: 3029408-3029408 on Build GRCh38
rs763530764 Chr.16: 3029519-3029519 on Build GRCh38
rs889805828 Chr.16: 3029466-3029466 on Build GRCh38
rs759075114 Chr.16: 3029432-3029432 on Build GRCh38
rs773390928 Chr.16: 3029355-3029355 on Build GRCh38
rs769749885 Chr.16: 3029354-3029355 on Build GRCh38
rs764619628 Chr.16: 3029362-3029362 on Build GRCh38
rs538924928 Chr.16: 3029224-3029224 on Build GRCh38
rs745893983 Chr.16: 3029402-3029402 on Build GRCh38
rs757158464 Chr.16: 3029500-3029500 on Build GRCh38
rs747027191 Chr.16: 3029584-3029584 on Build GRCh38
rs753052474 Chr.16: 3029250-3029250 on Build GRCh38
rs770892329 Chr.16: 3029394-3029394 on Build GRCh38
rs778864088 Chr.16: 3029501-3029501 on Build GRCh38
rs767923966 Chr.16: 3029249-3029249 on Build GRCh38
rs576703580 Chr.16: 3029203-3029203 on Build GRCh38
rs762186927 Chr.16: 3029511-3029511 on Build GRCh38
rs746691204 Chr.16: 3029303-3029303 on Build GRCh38
rs749268919 Chr.16: 3029530-3029531 on Build GRCh38
rs954720018 Chr.16: 3029569-3029569 on Build GRCh38
rs780175282 Chr.16: 3029577-3029577 on Build GRCh38
rs747289191 Chr.16: 3029505-3029505 on Build GRCh38
rs748285436 Chr.16: 3029597-3029597 on Build GRCh38
rs372529213 Chr.16: 3029368-3029368 on Build GRCh38
rs759817577 Chr.16: 3029336-3029336 on Build GRCh38
rs371864506 Chr.16: 3029315-3029315 on Build GRCh38
rs1030335565 Chr.16: 3029557-3029557 on Build GRCh38
rs781434469 Chr.16: 3029592-3029592 on Build GRCh38
rs770233287 Chr.16: 3029513-3029513 on Build GRCh38
rs748472235 Chr.16: 3029313-3029315 on Build GRCh38
rs753600031 Chr.16: 3029535-3029535 on Build GRCh38
rs763777633 Chr.16: 3029590-3029590 on Build GRCh38
rs763741621 Chr.16: 3029532-3029532 on Build GRCh38
rs764531674 Chr.16: 3029280-3029280 on Build GRCh38
rs564100878 Chr.16: 3029515-3029515 on Build GRCh38
rs910758581 Chr.16: 3029338-3029338 on Build GRCh38
rs375164652 Chr.16: 3029370-3029370 on Build GRCh38
rs747893032 Chr.16: 3029316-3029316 on Build GRCh38
rs1033968834 Chr.16: 3029108-3029108 on Build GRCh38
rs542011017 Chr.16: 3029489-3029489 on Build GRCh38
rs1007943148 Chr.16: 3029571-3029571 on Build GRCh38
rs760980874 Chr.16: 3029349-3029349 on Build GRCh38
rs755811374 Chr.16: 3029446-3029446 on Build GRCh38
rs757809971 Chr.16: 3029285-3029285 on Build GRCh38
rs777493357 Chr.16: 3029473-3029473 on Build GRCh38
rs923791212 Chr.16: 3029586-3029586 on Build GRCh38
rs752245890 Chr.16: 3029524-3029524 on Build GRCh38
rs778859674 Chr.16: 3029399-3029399 on Build GRCh38
rs113579453 Chr.16: 3029403-3029403 on Build GRCh38
rs759779427 Chr.16: 3029245-3029245 on Build GRCh38
rs557774072 Chr.16: 3029156-3029156 on Build GRCh38
rs765499461 Chr.16: 3029352-3029352 on Build GRCh38
rs778544976 Chr.16: 3029545-3029545 on Build GRCh38
rs775653345 Chr.16: 3029406-3029406 on Build GRCh38
rs1007168773 Chr.16: 3029148-3029151 on Build GRCh38
rs965604129 Chr.16: 3029193-3029193 on Build GRCh38
rs962084287 Chr.16: 3029300-3029300 on Build GRCh38
rs767190427 Chr.16: 3029523-3029523 on Build GRCh38
rs767104550 Chr.16: 3029437-3029437 on Build GRCh38
rs756621960 Chr.16: 3029264-3029264 on Build GRCh38
rs937620335 Chr.16: 3029454-3029454 on Build GRCh38
rs777599764 Chr.16: 3029387-3029387 on Build GRCh38
rs367690206 Chr.16: 3029123-3029123 on Build GRCh38
rs546945000 Chr.16: 3029549-3029549 on Build GRCh38
rs747985543 Chr.16: 3029380-3029380 on Build GRCh38
rs752443847 Chr.16: 3029438-3029438 on Build GRCh38
rs781275864 Chr.16: 3029310-3029312 on Build GRCh38
rs769873879 Chr.16: 3029321-3029321 on Build GRCh38
rs749974622 Chr.16: 3029284-3029284 on Build GRCh38
rs772981200 Chr.16: 3029322-3029322 on Build GRCh38
rs774474231 Chr.16: 3029239-3029239 on Build GRCh38
rs936462750 Chr.16: 3029595-3029595 on Build GRCh38
rs536415472 Chr.16: 3029304-3029304 on Build GRCh38
rs751051666 Chr.16: 3029430-3029430 on Build GRCh38
rs528244396 Chr.16: 3029488-3029488 on Build GRCh38
rs1041344444 Chr.16: 3029485-3029485 on Build GRCh38
rs368055371 Chr.16: 3029288-3029288 on Build GRCh38
rs993542570 Chr.16: 3029544-3029544 on Build GRCh38
rs754690316 Chr.16: 3029377-3029377 on Build GRCh38
rs751077412 Chr.16: 3029372-3029372 on Build GRCh38

View Full Product Details