Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3964863-3965369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY9
Entrez Gene ID
115
Gene Name
adenylate cyclase 9
Gene Aliases
AC9, ACIX
UniGene
Hs.391860
Gene Chromosome Location
Chr.16: 3953387-4116185 on Build GRCh38
Transcripts
NM_001116.3 XM_005255079.3 XM_011522353.2

SNP Detail

SNP ID Chromosome Location
rs927484318 Chr.16: 3965001-3965002 on Build GRCh38
rs994116655 Chr.16: 3965091-3965091 on Build GRCh38
rs901727387 Chr.16: 3965294-3965294 on Build GRCh38
rs746534307 Chr.16: 3964911-3964911 on Build GRCh38
rs974952425 Chr.16: 3964883-3964883 on Build GRCh38
rs895580683 Chr.16: 3964948-3964948 on Build GRCh38
rs532462231 Chr.16: 3964978-3964978 on Build GRCh38
rs945727438 Chr.16: 3965015-3965015 on Build GRCh38
rs910457106 Chr.16: 3965268-3965268 on Build GRCh38
rs571757606 Chr.16: 3965057-3965057 on Build GRCh38
rs886371919 Chr.16: 3965110-3965110 on Build GRCh38
rs191287403 Chr.16: 3965095-3965095 on Build GRCh38
rs1040640456 Chr.16: 3965160-3965160 on Build GRCh38
rs552550652 Chr.16: 3965190-3965190 on Build GRCh38
rs567635211 Chr.16: 3965300-3965300 on Build GRCh38
rs949861867 Chr.16: 3965082-3965082 on Build GRCh38
rs554809144 Chr.16: 3965308-3965308 on Build GRCh38
rs1044532848 Chr.16: 3965085-3965085 on Build GRCh38
rs999063710 Chr.16: 3965319-3965319 on Build GRCh38
rs776888112 Chr.16: 3965302-3965302 on Build GRCh38
rs560685322 Chr.16: 3965101-3965101 on Build GRCh38
rs574986413 Chr.16: 3965050-3965050 on Build GRCh38
rs916897986 Chr.16: 3965081-3965081 on Build GRCh38
rs768592885 Chr.16: 3965145-3965145 on Build GRCh38
rs368586638 Chr.16: 3965143-3965143 on Build GRCh38
rs907696907 Chr.16: 3965267-3965267 on Build GRCh38
rs922070982 Chr.16: 3964905-3964905 on Build GRCh38
rs2241074 Chr.16: 3964981-3964981 on Build GRCh38
rs556738201 Chr.16: 3965044-3965044 on Build GRCh38
rs146875372 Chr.16: 3964883-3964886 on Build GRCh38
rs547787930 Chr.16: 3965282-3965282 on Build GRCh38
rs986234583 Chr.16: 3965264-3965264 on Build GRCh38
rs934317041 Chr.16: 3964925-3964925 on Build GRCh38
rs992395897 Chr.16: 3965162-3965162 on Build GRCh38
rs1045475 Chr.16: 3965315-3965315 on Build GRCh38
rs886320856 Chr.16: 3964999-3964999 on Build GRCh38
rs1040377320 Chr.16: 3965125-3965125 on Build GRCh38
rs186443342 Chr.16: 3965006-3965006 on Build GRCh38
rs1052735555 Chr.16: 3964939-3964939 on Build GRCh38
rs878915934 Chr.16: 3965114-3965114 on Build GRCh38
rs539326823 Chr.16: 3965048-3965048 on Build GRCh38
rs529878977 Chr.16: 3964962-3964962 on Build GRCh38
rs147134502 Chr.16: 3965310-3965310 on Build GRCh38
rs749776530 Chr.16: 3965328-3965331 on Build GRCh38
rs536472988 Chr.16: 3965020-3965020 on Build GRCh38
rs887247 Chr.16: 3964977-3964977 on Build GRCh38
rs76046840 Chr.16: 3964921-3964921 on Build GRCh38
rs558780812 Chr.16: 3965052-3965052 on Build GRCh38
rs1000020630 Chr.16: 3964916-3964916 on Build GRCh38
rs944548633 Chr.16: 3965159-3965159 on Build GRCh38
rs968124851 Chr.16: 3965146-3965146 on Build GRCh38
rs182828449 Chr.16: 3965151-3965151 on Build GRCh38
rs994232348 Chr.16: 3965169-3965169 on Build GRCh38
rs143392571 Chr.16: 3965281-3965281 on Build GRCh38
rs988492037 Chr.16: 3965077-3965077 on Build GRCh38
rs1004733181 Chr.16: 3965122-3965122 on Build GRCh38
rs935871643 Chr.16: 3965150-3965150 on Build GRCh38
rs996314998 Chr.16: 3965128-3965128 on Build GRCh38
rs984237488 Chr.16: 3964994-3964994 on Build GRCh38
rs956964465 Chr.16: 3965161-3965161 on Build GRCh38
rs967542706 Chr.16: 3965317-3965317 on Build GRCh38
rs888567429 Chr.16: 3965307-3965307 on Build GRCh38
rs957026488 Chr.16: 3965254-3965254 on Build GRCh38
rs889845948 Chr.16: 3964938-3964938 on Build GRCh38
rs186942576 Chr.16: 3965303-3965303 on Build GRCh38
rs940642204 Chr.16: 3965275-3965275 on Build GRCh38
rs773225512 Chr.16: 3965152-3965152 on Build GRCh38
rs1048899503 Chr.16: 3965231-3965231 on Build GRCh38
rs1027983970 Chr.16: 3964985-3964985 on Build GRCh38
rs1045476 Chr.16: 3965312-3965312 on Build GRCh38
rs540615828 Chr.16: 3965093-3965093 on Build GRCh38
rs1031151016 Chr.16: 3965153-3965153 on Build GRCh38
rs959245374 Chr.16: 3965195-3965195 on Build GRCh38
rs927189312 Chr.16: 3965092-3965092 on Build GRCh38
rs710893 Chr.16: 3964963-3964963 on Build GRCh38
rs182829004 Chr.16: 3965002-3965002 on Build GRCh38
rs958904946 Chr.16: 3965126-3965126 on Build GRCh38
rs112776696 Chr.16: 3965170-3965170 on Build GRCh38
rs980084005 Chr.16: 3964912-3964912 on Build GRCh38
rs894708229 Chr.16: 3965311-3965311 on Build GRCh38
rs1005719848 Chr.16: 3965187-3965187 on Build GRCh38
rs547949685 Chr.16: 3965008-3965008 on Build GRCh38
rs55636902 Chr.16: 3964900-3964903 on Build GRCh38
rs138031058 Chr.16: 3965230-3965230 on Build GRCh38
rs927044824 Chr.16: 3965228-3965228 on Build GRCh38
rs567744516 Chr.16: 3965010-3965010 on Build GRCh38
rs146947190 Chr.16: 3965001-3965001 on Build GRCh38
rs901288281 Chr.16: 3965083-3965083 on Build GRCh38
rs879619 Chr.16: 3965045-3965045 on Build GRCh38
rs1029195929 Chr.16: 3965029-3965029 on Build GRCh38
rs771113627 Chr.16: 3965290-3965290 on Build GRCh38
rs532170098 Chr.16: 3965175-3965175 on Build GRCh38
rs386787951 Chr.16: 3965310-3965312 on Build GRCh38
rs1014409342 Chr.16: 3965216-3965216 on Build GRCh38
rs574454565 Chr.16: 3965127-3965127 on Build GRCh38

View Full Product Details