Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3966027-3966533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY9
Entrez Gene ID
115
Gene Name
adenylate cyclase 9
Gene Aliases
AC9, ACIX
UniGene
Hs.391860
Gene Chromosome Location
Chr.16: 3953387-4116185 on Build GRCh38
Transcripts
NM_001116.3 XM_005255079.3 XM_011522353.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
69560 c.3589A>G p.M1197V Chr.16: 3966248-3966248 on Build GRCh38
4544213 c.3520G>A p.A1174T Chr.16: 3966317-3966317 on Build GRCh38
5468503 c.3369G>A p.A1123A Chr.16: 3966468-3966468 on Build GRCh38
1182051 c.3550T>C p.Y1184H Chr.16: 3966287-3966287 on Build GRCh38
2920116 c.3351G>A p.A1117A Chr.16: 3966486-3966486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370606217 Chr.16: 3966456-3966456 on Build GRCh38
rs759501985 Chr.16: 3966285-3966285 on Build GRCh38
rs763810376 Chr.16: 3966364-3966364 on Build GRCh38
rs758359636 Chr.16: 3966387-3966387 on Build GRCh38
rs749968135 Chr.16: 3966071-3966071 on Build GRCh38
rs971860689 Chr.16: 3966175-3966175 on Build GRCh38
rs773926075 Chr.16: 3966414-3966414 on Build GRCh38
rs759986887 Chr.16: 3966153-3966153 on Build GRCh38
rs771534758 Chr.16: 3966420-3966420 on Build GRCh38
rs774577020 Chr.16: 3966056-3966056 on Build GRCh38
rs141452008 Chr.16: 3966152-3966152 on Build GRCh38
rs758404652 Chr.16: 3966073-3966073 on Build GRCh38
rs750569843 Chr.16: 3966454-3966454 on Build GRCh38
rs201893525 Chr.16: 3966178-3966178 on Build GRCh38
rs201993529 Chr.16: 3966444-3966444 on Build GRCh38
rs112769957 Chr.16: 3966390-3966390 on Build GRCh38
rs200863972 Chr.16: 3966196-3966196 on Build GRCh38
rs769101516 Chr.16: 3966330-3966330 on Build GRCh38
rs748299013 Chr.16: 3966107-3966107 on Build GRCh38
rs763389710 Chr.16: 3966050-3966050 on Build GRCh38
rs148534347 Chr.16: 3966432-3966432 on Build GRCh38
rs146365616 Chr.16: 3966306-3966306 on Build GRCh38
rs148547580 Chr.16: 3966462-3966462 on Build GRCh38
rs747992457 Chr.16: 3966218-3966218 on Build GRCh38
rs770583603 Chr.16: 3966413-3966413 on Build GRCh38
rs749440226 Chr.16: 3966138-3966138 on Build GRCh38
rs765047478 Chr.16: 3966193-3966193 on Build GRCh38
rs997201258 Chr.16: 3966368-3966368 on Build GRCh38
rs753481552 Chr.16: 3966064-3966064 on Build GRCh38
rs151282873 Chr.16: 3966047-3966047 on Build GRCh38
rs749352306 Chr.16: 3966224-3966224 on Build GRCh38
rs559533290 Chr.16: 3966149-3966149 on Build GRCh38
rs778993166 Chr.16: 3966312-3966312 on Build GRCh38
rs148224574 Chr.16: 3966240-3966240 on Build GRCh38
rs767418285 Chr.16: 3966293-3966293 on Build GRCh38
rs765844506 Chr.16: 3966270-3966270 on Build GRCh38
rs537741401 Chr.16: 3966459-3966459 on Build GRCh38
rs553001520 Chr.16: 3966117-3966117 on Build GRCh38
rs781677508 Chr.16: 3966400-3966400 on Build GRCh38
rs745449501 Chr.16: 3966469-3966469 on Build GRCh38
rs139727267 Chr.16: 3966327-3966327 on Build GRCh38
rs757425450 Chr.16: 3966311-3966311 on Build GRCh38
rs113410021 Chr.16: 3966498-3966498 on Build GRCh38
rs776481614 Chr.16: 3966442-3966442 on Build GRCh38
rs957965225 Chr.16: 3966171-3966171 on Build GRCh38
rs940194040 Chr.16: 3966466-3966466 on Build GRCh38
rs750492924 Chr.16: 3966386-3966386 on Build GRCh38
rs751041166 Chr.16: 3966272-3966272 on Build GRCh38
rs777570578 Chr.16: 3966305-3966305 on Build GRCh38
rs61731445 Chr.16: 3966376-3966376 on Build GRCh38
rs981533550 Chr.16: 3966180-3966180 on Build GRCh38
rs770998158 Chr.16: 3966226-3966226 on Build GRCh38
rs778234726 Chr.16: 3966468-3966468 on Build GRCh38
rs751743531 Chr.16: 3966460-3966460 on Build GRCh38
rs745559844 Chr.16: 3966419-3966419 on Build GRCh38
rs780981066 Chr.16: 3966216-3966216 on Build GRCh38
rs772950125 Chr.16: 3966259-3966259 on Build GRCh38
rs774968448 Chr.16: 3966471-3966471 on Build GRCh38
rs376101747 Chr.16: 3966396-3966396 on Build GRCh38
rs200368797 Chr.16: 3966315-3966315 on Build GRCh38
rs200746030 Chr.16: 3966318-3966318 on Build GRCh38
rs775005712 Chr.16: 3966296-3966296 on Build GRCh38
rs752461702 Chr.16: 3966300-3966300 on Build GRCh38
rs145997273 Chr.16: 3966051-3966051 on Build GRCh38
rs780077845 Chr.16: 3966082-3966082 on Build GRCh38
rs372820312 Chr.16: 3966345-3966345 on Build GRCh38
rs906369306 Chr.16: 3966371-3966373 on Build GRCh38
rs752586436 Chr.16: 3966357-3966357 on Build GRCh38
rs762725307 Chr.16: 3966267-3966267 on Build GRCh38
rs1007408384 Chr.16: 3966089-3966089 on Build GRCh38
rs145693515 Chr.16: 3966397-3966397 on Build GRCh38
rs776446870 Chr.16: 3966457-3966457 on Build GRCh38
rs770013848 Chr.16: 3966111-3966111 on Build GRCh38
rs746758399 Chr.16: 3966091-3966091 on Build GRCh38
rs770544319 Chr.16: 3966342-3966342 on Build GRCh38
rs755552694 Chr.16: 3966398-3966398 on Build GRCh38
rs144864117 Chr.16: 3966344-3966344 on Build GRCh38
rs201367484 Chr.16: 3966445-3966445 on Build GRCh38
rs764077367 Chr.16: 3966441-3966441 on Build GRCh38
rs770070508 Chr.16: 3966209-3966209 on Build GRCh38
rs202245497 Chr.16: 3966078-3966078 on Build GRCh38
rs754752263 Chr.16: 3966099-3966099 on Build GRCh38
rs767884304 Chr.16: 3966166-3966166 on Build GRCh38
rs762603873 Chr.16: 3966336-3966336 on Build GRCh38
rs114568080 Chr.16: 3966507-3966507 on Build GRCh38
rs771725261 Chr.16: 3966470-3966470 on Build GRCh38
rs748524658 Chr.16: 3966404-3966404 on Build GRCh38
rs778892072 Chr.16: 3966233-3966233 on Build GRCh38
rs551293106 Chr.16: 3966434-3966434 on Build GRCh38
rs267604541 Chr.16: 3966235-3966235 on Build GRCh38
rs754871847 Chr.16: 3966210-3966210 on Build GRCh38
rs780311928 Chr.16: 3966389-3966389 on Build GRCh38
rs369920383 Chr.16: 3966194-3966194 on Build GRCh38
rs149465612 Chr.16: 3966236-3966236 on Build GRCh38
rs780753391 Chr.16: 3966326-3966326 on Build GRCh38
rs11543317 Chr.16: 3966048-3966048 on Build GRCh38
rs756327580 Chr.16: 3966222-3966222 on Build GRCh38
rs143019825 Chr.16: 3966237-3966237 on Build GRCh38
rs201165281 Chr.16: 3966489-3966489 on Build GRCh38
rs141188630 Chr.16: 3966303-3966303 on Build GRCh38
rs940309439 Chr.16: 3966331-3966331 on Build GRCh38
rs756388353 Chr.16: 3966467-3966467 on Build GRCh38
rs376191720 Chr.16: 3966168-3966168 on Build GRCh38
rs769546776 Chr.16: 3966257-3966257 on Build GRCh38
rs544468394 Chr.16: 3966464-3966464 on Build GRCh38
rs139967862 Chr.16: 3966372-3966372 on Build GRCh38
rs759989865 Chr.16: 3966057-3966057 on Build GRCh38
rs576787672 Chr.16: 3966475-3966475 on Build GRCh38
rs774989006 Chr.16: 3966425-3966425 on Build GRCh38
rs757002362 Chr.16: 3966069-3966069 on Build GRCh38
rs371159341 Chr.16: 3966486-3966486 on Build GRCh38
rs773073326 Chr.16: 3966490-3966490 on Build GRCh38
rs764775400 Chr.16: 3966070-3966070 on Build GRCh38
rs746320046 Chr.16: 3966234-3966234 on Build GRCh38
rs761501337 Chr.16: 3966181-3966181 on Build GRCh38
rs778012664 Chr.16: 3966223-3966223 on Build GRCh38
rs74003478 Chr.16: 3966354-3966354 on Build GRCh38
rs374305565 Chr.16: 3966463-3966463 on Build GRCh38
rs773282474 Chr.16: 3966049-3966049 on Build GRCh38
rs766932381 Chr.16: 3966055-3966055 on Build GRCh38
rs61731447 Chr.16: 3966195-3966195 on Build GRCh38
rs777136439 Chr.16: 3966334-3966334 on Build GRCh38
rs757489326 Chr.16: 3966374-3966374 on Build GRCh38
rs200726858 Chr.16: 3966474-3966474 on Build GRCh38
rs914794233 Chr.16: 3966199-3966199 on Build GRCh38
rs147811130 Chr.16: 3966100-3966100 on Build GRCh38
rs545899558 Chr.16: 3966501-3966501 on Build GRCh38
rs376614156 Chr.16: 3966061-3966061 on Build GRCh38
rs773264079 Chr.16: 3966128-3966128 on Build GRCh38

View Full Product Details