Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3977215-3977721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCY9
Entrez Gene ID
115
Gene Name
adenylate cyclase 9
Gene Aliases
AC9, ACIX
UniGene
Hs.391860
Gene Chromosome Location
Chr.16: 3953387-4116185 on Build GRCh38
Transcripts
NM_001116.3 XM_005255079.3 XM_011522353.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
970631 c.2792C>T p.P931L Chr.16: 3977518-3977518 on Build GRCh38
970630 c.2797C>T p.L933F Chr.16: 3977513-3977513 on Build GRCh38
50420 c.2699C>T p.S900L Chr.16: 3977611-3977611 on Build GRCh38
4954167 c.2700G>A p.S900S Chr.16: 3977610-3977610 on Build GRCh38
4129059 c.2706G>A p.A902A Chr.16: 3977604-3977604 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375855314 Chr.16: 3977574-3977574 on Build GRCh38
rs960124920 Chr.16: 3977269-3977269 on Build GRCh38
rs1055171131 Chr.16: 3977279-3977279 on Build GRCh38
rs752983999 Chr.16: 3977567-3977567 on Build GRCh38
rs370804007 Chr.16: 3977523-3977523 on Build GRCh38
rs562312155 Chr.16: 3977499-3977499 on Build GRCh38
rs57040218 Chr.16: 3977595-3977595 on Build GRCh38
rs751190594 Chr.16: 3977640-3977640 on Build GRCh38
rs371184915 Chr.16: 3977431-3977431 on Build GRCh38
rs569435156 Chr.16: 3977674-3977674 on Build GRCh38
rs369126593 Chr.16: 3977439-3977439 on Build GRCh38
rs748833027 Chr.16: 3977467-3977467 on Build GRCh38
rs2072346 Chr.16: 3977422-3977422 on Build GRCh38
rs759954267 Chr.16: 3977551-3977551 on Build GRCh38
rs2230737 Chr.16: 3977516-3977516 on Build GRCh38
rs979016483 Chr.16: 3977322-3977322 on Build GRCh38
rs765175060 Chr.16: 3977438-3977438 on Build GRCh38
rs754261607 Chr.16: 3977625-3977625 on Build GRCh38
rs1045137938 Chr.16: 3977582-3977582 on Build GRCh38
rs543503895 Chr.16: 3977637-3977637 on Build GRCh38
rs142991024 Chr.16: 3977517-3977517 on Build GRCh38
rs771344306 Chr.16: 3977611-3977611 on Build GRCh38
rs966526069 Chr.16: 3977419-3977419 on Build GRCh38
rs779529291 Chr.16: 3977603-3977603 on Build GRCh38
rs754871190 Chr.16: 3977430-3977430 on Build GRCh38
rs776026141 Chr.16: 3977618-3977618 on Build GRCh38
rs1051940251 Chr.16: 3977364-3977364 on Build GRCh38
rs77550523 Chr.16: 3977433-3977433 on Build GRCh38
rs947411197 Chr.16: 3977292-3977292 on Build GRCh38
rs2240735 Chr.16: 3977604-3977604 on Build GRCh38
rs776953320 Chr.16: 3977682-3977682 on Build GRCh38
rs200478184 Chr.16: 3977475-3977475 on Build GRCh38
rs149752018 Chr.16: 3977478-3977478 on Build GRCh38
rs774265293 Chr.16: 3977616-3977616 on Build GRCh38
rs749110853 Chr.16: 3977642-3977642 on Build GRCh38
rs536043373 Chr.16: 3977350-3977350 on Build GRCh38
rs116699664 Chr.16: 3977409-3977409 on Build GRCh38
rs879613529 Chr.16: 3977308-3977308 on Build GRCh38
rs190578831 Chr.16: 3977692-3977692 on Build GRCh38
rs989376090 Chr.16: 3977408-3977408 on Build GRCh38
rs745542592 Chr.16: 3977529-3977529 on Build GRCh38
rs751717694 Chr.16: 3977543-3977543 on Build GRCh38
rs764388635 Chr.16: 3977624-3977624 on Build GRCh38
rs930130747 Chr.16: 3977464-3977464 on Build GRCh38
rs377690683 Chr.16: 3977511-3977511 on Build GRCh38
rs2531990 Chr.16: 3977687-3977687 on Build GRCh38
rs756453051 Chr.16: 3977591-3977591 on Build GRCh38
rs374209133 Chr.16: 3977501-3977501 on Build GRCh38
rs181898213 Chr.16: 3977641-3977641 on Build GRCh38
rs368437489 Chr.16: 3977667-3977667 on Build GRCh38
rs1038528149 Chr.16: 3977333-3977333 on Build GRCh38
rs749133341 Chr.16: 3977652-3977652 on Build GRCh38
rs905073122 Chr.16: 3977584-3977584 on Build GRCh38
rs567101028 Chr.16: 3977323-3977323 on Build GRCh38
rs374631917 Chr.16: 3977519-3977519 on Build GRCh38
rs746866743 Chr.16: 3977532-3977532 on Build GRCh38
rs550558850 Chr.16: 3977623-3977623 on Build GRCh38
rs746616451 Chr.16: 3977665-3977666 on Build GRCh38
rs763072978 Chr.16: 3977505-3977505 on Build GRCh38
rs928035957 Chr.16: 3977427-3977427 on Build GRCh38
rs144319192 Chr.16: 3977525-3977525 on Build GRCh38
rs768273214 Chr.16: 3977494-3977494 on Build GRCh38
rs779548082 Chr.16: 3977531-3977531 on Build GRCh38
rs993216643 Chr.16: 3977375-3977375 on Build GRCh38
rs773524719 Chr.16: 3977538-3977538 on Build GRCh38
rs1038646010 Chr.16: 3977442-3977442 on Build GRCh38
rs939351835 Chr.16: 3977274-3977274 on Build GRCh38
rs746108935 Chr.16: 3977664-3977664 on Build GRCh38
rs941623549 Chr.16: 3977298-3977298 on Build GRCh38
rs1039087539 Chr.16: 3977488-3977488 on Build GRCh38
rs372533992 Chr.16: 3977562-3977562 on Build GRCh38
rs150826321 Chr.16: 3977594-3977594 on Build GRCh38
rs113072194 Chr.16: 3977267-3977267 on Build GRCh38
rs764654411 Chr.16: 3977677-3977677 on Build GRCh38
rs750505834 Chr.16: 3977503-3977503 on Build GRCh38
rs1044966166 Chr.16: 3977423-3977423 on Build GRCh38
rs755257131 Chr.16: 3977451-3977451 on Build GRCh38
rs756178677 Chr.16: 3977646-3977646 on Build GRCh38
rs753174574 Chr.16: 3977521-3977521 on Build GRCh38
rs755725221 Chr.16: 3977388-3977388 on Build GRCh38
rs912752455 Chr.16: 3977605-3977605 on Build GRCh38
rs374557206 Chr.16: 3977607-3977607 on Build GRCh38
rs760701703 Chr.16: 3977493-3977493 on Build GRCh38
rs775450035 Chr.16: 3977670-3977670 on Build GRCh38
rs890192216 Chr.16: 3977477-3977477 on Build GRCh38
rs749507415 Chr.16: 3977610-3977610 on Build GRCh38
rs539831915 Chr.16: 3977695-3977695 on Build GRCh38
rs960583841 Chr.16: 3977675-3977675 on Build GRCh38
rs777704656 Chr.16: 3977649-3977649 on Build GRCh38
rs765593436 Chr.16: 3977502-3977502 on Build GRCh38
rs201954088 Chr.16: 3977476-3977476 on Build GRCh38
rs974060738 Chr.16: 3977696-3977696 on Build GRCh38
rs897346006 Chr.16: 3977353-3977353 on Build GRCh38
rs1023725538 Chr.16: 3977691-3977691 on Build GRCh38
rs185673232 Chr.16: 3977660-3977660 on Build GRCh38
rs759712460 Chr.16: 3977617-3977617 on Build GRCh38
rs374307728 Chr.16: 3977666-3977666 on Build GRCh38
rs1026491304 Chr.16: 3977382-3977382 on Build GRCh38
rs932139100 Chr.16: 3977353-3977353 on Build GRCh38
rs370600219 Chr.16: 3977658-3977658 on Build GRCh38
rs79048557 Chr.16: 3977290-3977290 on Build GRCh38
rs147280539 Chr.16: 3977606-3977606 on Build GRCh38
rs116089419 Chr.16: 3977445-3977445 on Build GRCh38
rs776256089 Chr.16: 3977534-3977534 on Build GRCh38
rs772124621 Chr.16: 3977669-3977669 on Build GRCh38
rs541835290 Chr.16: 3977387-3977387 on Build GRCh38
rs898817710 Chr.16: 3977417-3977417 on Build GRCh38
rs74003485 Chr.16: 3977535-3977535 on Build GRCh38
rs999474870 Chr.16: 3977673-3977673 on Build GRCh38
rs1017302825 Chr.16: 3977397-3977397 on Build GRCh38
rs375357908 Chr.16: 3977459-3977459 on Build GRCh38
rs73506099 Chr.16: 3977398-3977398 on Build GRCh38
rs373556046 Chr.16: 3977592-3977592 on Build GRCh38
rs758044219 Chr.16: 3977628-3977628 on Build GRCh38
rs543707645 Chr.16: 3977460-3977460 on Build GRCh38
rs141549186 Chr.16: 3977541-3977541 on Build GRCh38
rs577594589 Chr.16: 3977694-3977694 on Build GRCh38

View Full Product Details