Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4434351-4434864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJA3
Entrez Gene ID
9093
Gene Name
DnaJ heat shock protein family (Hsp40) member A3
Gene Aliases
HCA57, TID1, hTID-1
UniGene
Hs.459779
Gene Chromosome Location
Chr.16: 4425805-4456775 on Build GRCh38
Transcripts
NM_001135110.2 NM_001286516.1 NM_005147.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7365041 c.283T>C p.Y95H Chr.16: 4434455-4434455 on Build GRCh38
4678633 c.305G>A p.R102Q Chr.16: 4434477-4434477 on Build GRCh38
4437440 c.231C>G p.F77L Chr.16: 4434403-4434403 on Build GRCh38
7132378 c.223A>T p.N75Y Chr.16: 4434395-4434395 on Build GRCh38
7127200 c.288G>C p.Q96H Chr.16: 4434460-4434460 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760612520 Chr.16: 4434397-4434397 on Build GRCh38
rs559299405 Chr.16: 4434724-4434724 on Build GRCh38
rs996672072 Chr.16: 4434808-4434808 on Build GRCh38
rs763564390 Chr.16: 4434558-4434558 on Build GRCh38
rs532144603 Chr.16: 4434688-4434688 on Build GRCh38
rs777227683 Chr.16: 4434444-4434446 on Build GRCh38
rs141763240 Chr.16: 4434411-4434411 on Build GRCh38
rs963833062 Chr.16: 4434689-4434689 on Build GRCh38
rs895343440 Chr.16: 4434747-4434747 on Build GRCh38
rs570295898 Chr.16: 4434622-4434622 on Build GRCh38
rs929660256 Chr.16: 4434711-4434711 on Build GRCh38
rs548062244 Chr.16: 4434777-4434777 on Build GRCh38
rs752265170 Chr.16: 4434426-4434426 on Build GRCh38
rs747250647 Chr.16: 4434497-4434497 on Build GRCh38
rs549880420 Chr.16: 4434836-4434836 on Build GRCh38
rs146234364 Chr.16: 4434457-4434457 on Build GRCh38
rs778933963 Chr.16: 4434567-4434567 on Build GRCh38
rs575064576 Chr.16: 4434636-4434636 on Build GRCh38
rs559211238 Chr.16: 4434596-4434596 on Build GRCh38
rs763246928 Chr.16: 4434539-4434539 on Build GRCh38
rs989578725 Chr.16: 4434678-4434678 on Build GRCh38
rs61263579 Chr.16: 4434812-4434812 on Build GRCh38
rs901886590 Chr.16: 4434805-4434805 on Build GRCh38
rs976650709 Chr.16: 4434709-4434710 on Build GRCh38
rs757280127 Chr.16: 4434566-4434566 on Build GRCh38
rs1005356450 Chr.16: 4434464-4434464 on Build GRCh38
rs770247411 Chr.16: 4434513-4434513 on Build GRCh38
rs757241270 Chr.16: 4434469-4434469 on Build GRCh38
rs1012400478 Chr.16: 4434770-4434770 on Build GRCh38
rs746143433 Chr.16: 4434392-4434392 on Build GRCh38
rs181511914 Chr.16: 4434413-4434413 on Build GRCh38
rs765636804 Chr.16: 4434412-4434412 on Build GRCh38
rs917039979 Chr.16: 4434614-4434614 on Build GRCh38
rs775505220 Chr.16: 4434488-4434488 on Build GRCh38
rs141642995 Chr.16: 4434393-4434393 on Build GRCh38
rs1031093669 Chr.16: 4434671-4434671 on Build GRCh38
rs183024665 Chr.16: 4434712-4434712 on Build GRCh38
rs373910237 Chr.16: 4434763-4434763 on Build GRCh38
rs562062802 Chr.16: 4434541-4434541 on Build GRCh38
rs572603821 Chr.16: 4434601-4434601 on Build GRCh38
rs764667473 Chr.16: 4434404-4434404 on Build GRCh38
rs989685752 Chr.16: 4434431-4434431 on Build GRCh38
rs528412987 Chr.16: 4434740-4434740 on Build GRCh38
rs375801040 Chr.16: 4434727-4434727 on Build GRCh38
rs768675796 Chr.16: 4434505-4434505 on Build GRCh38
rs777337152 Chr.16: 4434441-4434441 on Build GRCh38
rs371319724 Chr.16: 4434794-4434794 on Build GRCh38
rs17137026 Chr.16: 4434677-4434677 on Build GRCh38
rs745607149 Chr.16: 4434477-4434477 on Build GRCh38
rs202008839 Chr.16: 4434795-4434795 on Build GRCh38
rs978217090 Chr.16: 4434403-4434403 on Build GRCh38
rs1139653 Chr.16: 4434395-4434395 on Build GRCh38
rs561505736 Chr.16: 4434630-4434630 on Build GRCh38
rs927454654 Chr.16: 4434655-4434655 on Build GRCh38
rs978662041 Chr.16: 4434593-4434593 on Build GRCh38
rs4785964 Chr.16: 4434612-4434612 on Build GRCh38
rs760365954 Chr.16: 4434546-4434546 on Build GRCh38
rs138128625 Chr.16: 4434472-4434472 on Build GRCh38
rs953929439 Chr.16: 4434549-4434549 on Build GRCh38
rs922301209 Chr.16: 4434494-4434494 on Build GRCh38
rs149525378 Chr.16: 4434508-4434508 on Build GRCh38
rs1012525158 Chr.16: 4434615-4434615 on Build GRCh38
rs888153583 Chr.16: 4434589-4434589 on Build GRCh38
rs543564602 Chr.16: 4434643-4434643 on Build GRCh38
rs61737690 Chr.16: 4434394-4434394 on Build GRCh38
rs1055531873 Chr.16: 4434746-4434747 on Build GRCh38
rs950263101 Chr.16: 4434619-4434619 on Build GRCh38
rs955756314 Chr.16: 4434597-4434597 on Build GRCh38
rs922499124 Chr.16: 4434709-4434709 on Build GRCh38
rs34689956 Chr.16: 4434381-4434381 on Build GRCh38
rs771027872 Chr.16: 4434666-4434666 on Build GRCh38
rs186046750 Chr.16: 4434434-4434434 on Build GRCh38
rs999896539 Chr.16: 4434669-4434669 on Build GRCh38
rs189922134 Chr.16: 4434571-4434571 on Build GRCh38
rs767031971 Chr.16: 4434417-4434417 on Build GRCh38
rs909541028 Chr.16: 4434720-4434720 on Build GRCh38
rs779974169 Chr.16: 4434542-4434542 on Build GRCh38
rs773589914 Chr.16: 4434524-4434524 on Build GRCh38

View Full Product Details