Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4700881-4701382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKS3
Entrez Gene ID
124401
Gene Name
ankyrin repeat and sterile alpha motif domain containing 3
Gene Aliases
-
UniGene
Hs.654859
Gene Chromosome Location
Chr.16: 4696510-4734377 on Build GRCh38
Transcripts
NM_001242929.1 NM_001308089.1 NM_001324129.1 NM_001324130.1 NM_133450.3 NR_040252.1 XM_005255119.3 XM_005255121.3 XM_011522372.1 XM_011522373.1 XM_011522374.1 XM_011522378.1 XM_017022935.1 XM_017022936.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4864506 c.628A>G p.S210G Chr.16: 4701008-4701008 on Build GRCh38
6600938 c.1265C>A p.A422D Chr.16: 4700989-4700989 on Build GRCh38
5953299 c.1208G>A p.R403H Chr.16: 4701046-4701046 on Build GRCh38
6600940 c.1236C>T p.L412L Chr.16: 4701018-4701018 on Build GRCh38
2921197 c.1198T>C p.W400R Chr.16: 4701056-4701056 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949456177 Chr.16: 4701355-4701355 on Build GRCh38
rs750485043 Chr.16: 4701095-4701095 on Build GRCh38
rs769007502 Chr.16: 4701061-4701061 on Build GRCh38
rs561559700 Chr.16: 4701152-4701152 on Build GRCh38
rs1001476050 Chr.16: 4701306-4701306 on Build GRCh38
rs61747670 Chr.16: 4701117-4701117 on Build GRCh38
rs747237679 Chr.16: 4700935-4700936 on Build GRCh38
rs766904769 Chr.16: 4701100-4701100 on Build GRCh38
rs779464781 Chr.16: 4700926-4700926 on Build GRCh38
rs780213832 Chr.16: 4700932-4700933 on Build GRCh38
rs770860603 Chr.16: 4701018-4701018 on Build GRCh38
rs201518736 Chr.16: 4701065-4701065 on Build GRCh38
rs779978721 Chr.16: 4701124-4701124 on Build GRCh38
rs919541136 Chr.16: 4701308-4701308 on Build GRCh38
rs756913301 Chr.16: 4701167-4701167 on Build GRCh38
rs956981480 Chr.16: 4701315-4701315 on Build GRCh38
rs35706305 Chr.16: 4700939-4700939 on Build GRCh38
rs766291824 Chr.16: 4700964-4700964 on Build GRCh38
rs796584761 Chr.16: 4701322-4701322 on Build GRCh38
rs767905967 Chr.16: 4701163-4701163 on Build GRCh38
rs780322777 Chr.16: 4701045-4701045 on Build GRCh38
rs377508134 Chr.16: 4700988-4700988 on Build GRCh38
rs759922184 Chr.16: 4701153-4701153 on Build GRCh38
rs777339698 Chr.16: 4701048-4701048 on Build GRCh38
rs753942598 Chr.16: 4700989-4700989 on Build GRCh38
rs748793746 Chr.16: 4701014-4701014 on Build GRCh38
rs373203318 Chr.16: 4701294-4701294 on Build GRCh38
rs534018882 Chr.16: 4701228-4701228 on Build GRCh38
rs924060977 Chr.16: 4701213-4701213 on Build GRCh38
rs376850764 Chr.16: 4701154-4701154 on Build GRCh38
rs953820300 Chr.16: 4701277-4701277 on Build GRCh38
rs755830480 Chr.16: 4701047-4701047 on Build GRCh38
rs773376148 Chr.16: 4700962-4700962 on Build GRCh38
rs184477066 Chr.16: 4701227-4701227 on Build GRCh38
rs764313516 Chr.16: 4700987-4700987 on Build GRCh38
rs61747669 Chr.16: 4701123-4701123 on Build GRCh38
rs556840530 Chr.16: 4701010-4701010 on Build GRCh38
rs1032190855 Chr.16: 4701242-4701242 on Build GRCh38
rs757869867 Chr.16: 4701177-4701177 on Build GRCh38
rs863980 Chr.16: 4701044-4701044 on Build GRCh38
rs767796055 Chr.16: 4700973-4700973 on Build GRCh38
rs917903292 Chr.16: 4701347-4701347 on Build GRCh38
rs140370704 Chr.16: 4701009-4701009 on Build GRCh38
rs763419237 Chr.16: 4701098-4701098 on Build GRCh38
rs745366095 Chr.16: 4701122-4701122 on Build GRCh38
rs919442296 Chr.16: 4701208-4701208 on Build GRCh38
rs560985943 Chr.16: 4701314-4701314 on Build GRCh38
rs147502184 Chr.16: 4701034-4701034 on Build GRCh38
rs374718308 Chr.16: 4701169-4701169 on Build GRCh38
rs752829122 Chr.16: 4700919-4700919 on Build GRCh38
rs909359848 Chr.16: 4701323-4701323 on Build GRCh38
rs749910201 Chr.16: 4701171-4701171 on Build GRCh38
rs200554062 Chr.16: 4701053-4701053 on Build GRCh38
rs149148220 Chr.16: 4701046-4701046 on Build GRCh38
rs769243319 Chr.16: 4700945-4700945 on Build GRCh38
rs777026174 Chr.16: 4701134-4701134 on Build GRCh38
rs750801351 Chr.16: 4701004-4701004 on Build GRCh38
rs759189093 Chr.16: 4700972-4700972 on Build GRCh38
rs895837795 Chr.16: 4701194-4701194 on Build GRCh38
rs765193230 Chr.16: 4701094-4701094 on Build GRCh38
rs765789292 Chr.16: 4701043-4701043 on Build GRCh38
rs552056904 Chr.16: 4701211-4701211 on Build GRCh38
rs373611584 Chr.16: 4700931-4700931 on Build GRCh38
rs116804869 Chr.16: 4701324-4701324 on Build GRCh38
rs200119184 Chr.16: 4701102-4701102 on Build GRCh38
rs144928887 Chr.16: 4701024-4701024 on Build GRCh38
rs956726102 Chr.16: 4701214-4701214 on Build GRCh38
rs376592108 Chr.16: 4701166-4701166 on Build GRCh38
rs142502357 Chr.16: 4700985-4700985 on Build GRCh38
rs747705530 Chr.16: 4700939-4700939 on Build GRCh38
rs534219168 Chr.16: 4700920-4700920 on Build GRCh38
rs992267348 Chr.16: 4701326-4701326 on Build GRCh38
rs904621593 Chr.16: 4701234-4701234 on Build GRCh38
rs545641938 Chr.16: 4701267-4701267 on Build GRCh38
rs896430929 Chr.16: 4701032-4701032 on Build GRCh38
rs778332227 Chr.16: 4701118-4701118 on Build GRCh38
rs1044318684 Chr.16: 4701237-4701237 on Build GRCh38
rs768524954 Chr.16: 4701131-4701131 on Build GRCh38
rs549661337 Chr.16: 4701125-4701125 on Build GRCh38
rs141742096 Chr.16: 4701025-4701025 on Build GRCh38
rs878897413 Chr.16: 4701200-4701200 on Build GRCh38
rs746202464 Chr.16: 4700927-4700927 on Build GRCh38
rs761920423 Chr.16: 4701038-4701038 on Build GRCh38
rs538380208 Chr.16: 4701183-4701183 on Build GRCh38
rs1029139432 Chr.16: 4701203-4701203 on Build GRCh38
rs748422660 Chr.16: 4701146-4701146 on Build GRCh38
rs968238092 Chr.16: 4701204-4701204 on Build GRCh38
rs950989057 Chr.16: 4701313-4701313 on Build GRCh38
rs979523062 Chr.16: 4701210-4701210 on Build GRCh38
rs762008368 Chr.16: 4701074-4701074 on Build GRCh38
rs558864636 Chr.16: 4701276-4701276 on Build GRCh38
rs559682456 Chr.16: 4701001-4701001 on Build GRCh38
rs188380762 Chr.16: 4701244-4701244 on Build GRCh38
rs930509565 Chr.16: 4701243-4701243 on Build GRCh38
rs770035706 Chr.16: 4701147-4701147 on Build GRCh38
rs554176368 Chr.16: 4701241-4701241 on Build GRCh38
rs192672796 Chr.16: 4701149-4701149 on Build GRCh38
rs142863773 Chr.16: 4701307-4701307 on Build GRCh38
rs1044183995 Chr.16: 4701144-4701144 on Build GRCh38
rs1001143593 Chr.16: 4701271-4701271 on Build GRCh38
rs1013515201 Chr.16: 4701256-4701256 on Build GRCh38
rs560707397 Chr.16: 4701049-4701049 on Build GRCh38
rs140099226 Chr.16: 4701261-4701261 on Build GRCh38
rs748808238 Chr.16: 4701050-4701050 on Build GRCh38
rs544466108 Chr.16: 4701057-4701057 on Build GRCh38
rs763756520 Chr.16: 4701106-4701106 on Build GRCh38
rs542978994 Chr.16: 4701346-4701346 on Build GRCh38
rs199714601 Chr.16: 4700948-4700948 on Build GRCh38
rs949940695 Chr.16: 4701229-4701229 on Build GRCh38
rs117007233 Chr.16: 4701003-4701003 on Build GRCh38
rs929423699 Chr.16: 4701219-4701220 on Build GRCh38
rs576778362 Chr.16: 4701266-4701266 on Build GRCh38
rs374857337 Chr.16: 4701211-4701212 on Build GRCh38
rs780701770 Chr.16: 4701270-4701270 on Build GRCh38
rs140571025 Chr.16: 4701088-4701088 on Build GRCh38
rs778937795 Chr.16: 4701052-4701052 on Build GRCh38
rs774270387 Chr.16: 4700965-4700965 on Build GRCh38
rs768419782 Chr.16: 4700929-4700929 on Build GRCh38
rs757811663 Chr.16: 4700925-4700925 on Build GRCh38
rs151280564 Chr.16: 4701060-4701060 on Build GRCh38
rs371478473 Chr.16: 4701168-4701168 on Build GRCh38
rs780164841 Chr.16: 4701179-4701179 on Build GRCh38
rs753445730 Chr.16: 4701109-4701109 on Build GRCh38
rs754042181 Chr.16: 4701037-4701037 on Build GRCh38
rs572141497 Chr.16: 4701292-4701292 on Build GRCh38
rs145685232 Chr.16: 4701104-4701104 on Build GRCh38
rs757893307 Chr.16: 4700994-4700994 on Build GRCh38
rs1006571926 Chr.16: 4701283-4701283 on Build GRCh38
rs1027293114 Chr.16: 4700961-4700961 on Build GRCh38
rs117674949 Chr.16: 4701245-4701245 on Build GRCh38
rs369241127 Chr.16: 4701022-4701022 on Build GRCh38
rs141062211 Chr.16: 4701012-4701012 on Build GRCh38

View Full Product Details