Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 5055731-5056232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf89
Entrez Gene ID
146556
Gene Name
chromosome 16 open reading frame 89
Gene Aliases
-
UniGene
Hs.11782
Gene Chromosome Location
Chr.16: 5042772-5066145 on Build GRCh38
Transcripts
NM_001098514.2 NM_152459.4 XM_005255143.3 XM_011522392.2 XM_017022972.1 XM_017022973.1 XM_017022974.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4630226 c.635G>A p.C212Y Chr.16: 5056181-5056181 on Build GRCh38
4630227 c.749G>A p.C250Y Chr.16: 5056181-5056181 on Build GRCh38
3510117 c.705C>T p.N235N Chr.16: 5056111-5056111 on Build GRCh38
3510118 c.819C>T p.N273N Chr.16: 5056111-5056111 on Build GRCh38
3510116 c.819C>T p.N273N Chr.16: 5056111-5056111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991965815 Chr.16: 5056146-5056146 on Build GRCh38
rs999572208 Chr.16: 5056176-5056176 on Build GRCh38
rs1022703899 Chr.16: 5055889-5055889 on Build GRCh38
rs370420720 Chr.16: 5056087-5056087 on Build GRCh38
rs538095556 Chr.16: 5056160-5056160 on Build GRCh38
rs869062887 Chr.16: 5055995-5056000 on Build GRCh38
rs1025674131 Chr.16: 5055881-5055881 on Build GRCh38
rs917404918 Chr.16: 5055971-5055971 on Build GRCh38
rs757406420 Chr.16: 5056125-5056125 on Build GRCh38
rs756025423 Chr.16: 5056148-5056148 on Build GRCh38
rs531204165 Chr.16: 5055754-5055754 on Build GRCh38
rs765445379 Chr.16: 5056131-5056131 on Build GRCh38
rs17853191 Chr.16: 5056172-5056172 on Build GRCh38
rs202196210 Chr.16: 5056147-5056147 on Build GRCh38
rs755407771 Chr.16: 5056005-5056005 on Build GRCh38
rs79730909 Chr.16: 5055781-5055781 on Build GRCh38
rs758832774 Chr.16: 5055905-5055906 on Build GRCh38
rs1038070427 Chr.16: 5056051-5056051 on Build GRCh38
rs780895009 Chr.16: 5056080-5056080 on Build GRCh38
rs751358584 Chr.16: 5055868-5055868 on Build GRCh38
rs765355947 Chr.16: 5056186-5056186 on Build GRCh38
rs367572318 Chr.16: 5056206-5056206 on Build GRCh38
rs771654013 Chr.16: 5056165-5056165 on Build GRCh38
rs555108239 Chr.16: 5056110-5056110 on Build GRCh38
rs756627552 Chr.16: 5056068-5056068 on Build GRCh38
rs77055786 Chr.16: 5056049-5056049 on Build GRCh38
rs751948302 Chr.16: 5056002-5056002 on Build GRCh38
rs1050616548 Chr.16: 5056004-5056004 on Build GRCh38
rs369429066 Chr.16: 5056027-5056027 on Build GRCh38
rs374969779 Chr.16: 5056026-5056026 on Build GRCh38
rs759228896 Chr.16: 5056098-5056098 on Build GRCh38
rs768631583 Chr.16: 5056037-5056037 on Build GRCh38
rs766685046 Chr.16: 5056001-5056001 on Build GRCh38
rs750558810 Chr.16: 5056194-5056194 on Build GRCh38
rs780995753 Chr.16: 5056144-5056144 on Build GRCh38
rs548928819 Chr.16: 5055997-5056000 on Build GRCh38
rs376423612 Chr.16: 5056023-5056023 on Build GRCh38
rs1018171147 Chr.16: 5055809-5055809 on Build GRCh38
rs749504039 Chr.16: 5056016-5056016 on Build GRCh38
rs751869294 Chr.16: 5056204-5056204 on Build GRCh38
rs748065832 Chr.16: 5056081-5056081 on Build GRCh38
rs554986494 Chr.16: 5056118-5056118 on Build GRCh38
rs890962743 Chr.16: 5055792-5055792 on Build GRCh38
rs371492300 Chr.16: 5056006-5056006 on Build GRCh38
rs1032065596 Chr.16: 5055793-5055793 on Build GRCh38
rs370738780 Chr.16: 5056142-5056142 on Build GRCh38
rs775787206 Chr.16: 5055761-5055761 on Build GRCh38
rs774928212 Chr.16: 5056073-5056073 on Build GRCh38
rs762565918 Chr.16: 5055824-5055824 on Build GRCh38
rs762456086 Chr.16: 5056184-5056184 on Build GRCh38
rs989472888 Chr.16: 5055865-5055865 on Build GRCh38
rs991679300 Chr.16: 5055939-5055939 on Build GRCh38
rs753215618 Chr.16: 5056065-5056065 on Build GRCh38
rs1022334245 Chr.16: 5056164-5056164 on Build GRCh38
rs189796530 Chr.16: 5055779-5055779 on Build GRCh38
rs762622122 Chr.16: 5056048-5056048 on Build GRCh38
rs531301214 Chr.16: 5055977-5056000 on Build GRCh38
rs760934059 Chr.16: 5056174-5056174 on Build GRCh38
rs777859745 Chr.16: 5056069-5056069 on Build GRCh38
rs866338204 Chr.16: 5055912-5055912 on Build GRCh38
rs768537438 Chr.16: 5056075-5056075 on Build GRCh38
rs770398633 Chr.16: 5056154-5056154 on Build GRCh38
rs1006815438 Chr.16: 5055808-5055808 on Build GRCh38
rs752315815 Chr.16: 5056005-5056005 on Build GRCh38
rs759878886 Chr.16: 5056061-5056061 on Build GRCh38
rs761177465 Chr.16: 5056117-5056117 on Build GRCh38
rs746814468 Chr.16: 5056031-5056031 on Build GRCh38
rs741164 Chr.16: 5056183-5056183 on Build GRCh38
rs375098791 Chr.16: 5056132-5056132 on Build GRCh38
rs372296873 Chr.16: 5056173-5056173 on Build GRCh38
rs767934427 Chr.16: 5056062-5056062 on Build GRCh38
rs539842654 Chr.16: 5056202-5056202 on Build GRCh38
rs1006727820 Chr.16: 5055813-5055813 on Build GRCh38
rs772623903 Chr.16: 5056047-5056047 on Build GRCh38
rs967072768 Chr.16: 5056171-5056171 on Build GRCh38
rs977071433 Chr.16: 5055879-5055879 on Build GRCh38
rs984091142 Chr.16: 5055907-5055907 on Build GRCh38
rs376024213 Chr.16: 5056071-5056071 on Build GRCh38
rs748422574 Chr.16: 5056109-5056109 on Build GRCh38
rs1018491143 Chr.16: 5055820-5055820 on Build GRCh38
rs373197393 Chr.16: 5056038-5056038 on Build GRCh38
rs577604693 Chr.16: 5056198-5056198 on Build GRCh38
rs893459334 Chr.16: 5056185-5056185 on Build GRCh38
rs754823631 Chr.16: 5056030-5056030 on Build GRCh38
rs965356907 Chr.16: 5055847-5055847 on Build GRCh38
rs535300927 Chr.16: 5056093-5056093 on Build GRCh38
rs1025577728 Chr.16: 5056082-5056082 on Build GRCh38
rs754425569 Chr.16: 5056123-5056123 on Build GRCh38
rs369867447 Chr.16: 5056060-5056060 on Build GRCh38
rs375608707 Chr.16: 5056158-5056158 on Build GRCh38
rs201720626 Chr.16: 5056070-5056070 on Build GRCh38
rs762467164 Chr.16: 5056092-5056092 on Build GRCh38
rs76078092 Chr.16: 5055758-5055758 on Build GRCh38
rs1046089445 Chr.16: 5055777-5055777 on Build GRCh38
rs550721990 Chr.16: 5055766-5055766 on Build GRCh38
rs777754698 Chr.16: 5056008-5056008 on Build GRCh38
rs746157593 Chr.16: 5056074-5056074 on Build GRCh38
rs777560889 Chr.16: 5056153-5056153 on Build GRCh38
rs937382123 Chr.16: 5055921-5055921 on Build GRCh38
rs769645480 Chr.16: 5056086-5056086 on Build GRCh38

View Full Product Details