Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 5085000-5085494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG1
Entrez Gene ID
56052
Gene Name
ALG1, chitobiosyldiphosphodolichol beta-mannosyltransferase
Gene Aliases
CDG1K, HMAT1, HMT-1, HMT1, MT-1, Mat-1, hMat-1
UniGene
Hs.592086
Gene Chromosome Location
Chr.16: 5071809-5087379 on Build GRCh38
Transcripts
NM_019109.4 XM_011522565.1 XM_017023457.1 XM_017023458.1 NM_001289029.1 NM_201400.3 NM_201598.3 XM_005255157.4 XM_005255158.4 XM_011522404.2

Gene Symbol
EEF2KMT
Entrez Gene ID
196483
Gene Name
eukaryotic elongation factor 2 lysine methyltransferase
Gene Aliases
FAM86A, SB153, eEF2-KMT
UniGene
Hs.406461
Gene Chromosome Location
Chr.16: 5084298-5097820 on Build GRCh38
Transcripts
NM_019109.4 XM_011522565.1 XM_017023457.1 XM_017023458.1 NM_001289029.1 NM_201400.3 NM_201598.3 XM_005255157.4 XM_005255158.4 XM_011522404.2

SNP Detail

SNP ID Chromosome Location
rs1063200 Chr.16: 5085059-5085059 on Build GRCh38
rs910823463 Chr.16: 5085046-5085046 on Build GRCh38
rs1047639 Chr.16: 5085049-5085049 on Build GRCh38
rs554392935 Chr.16: 5085400-5085400 on Build GRCh38
rs558389580 Chr.16: 5085288-5085288 on Build GRCh38
rs549652869 Chr.16: 5085257-5085257 on Build GRCh38
rs982829657 Chr.16: 5085472-5085472 on Build GRCh38
rs531718461 Chr.16: 5085393-5085393 on Build GRCh38
rs867352336 Chr.16: 5085331-5085331 on Build GRCh38
rs957568943 Chr.16: 5085040-5085040 on Build GRCh38
rs55848638 Chr.16: 5085156-5085156 on Build GRCh38
rs709257 Chr.16: 5085022-5085022 on Build GRCh38
rs990240413 Chr.16: 5085045-5085045 on Build GRCh38
rs569317828 Chr.16: 5085261-5085261 on Build GRCh38
rs145044749 Chr.16: 5085188-5085188 on Build GRCh38
rs1047566 Chr.16: 5085452-5085452 on Build GRCh38
rs760391565 Chr.16: 5085325-5085325 on Build GRCh38
rs867547136 Chr.16: 5085351-5085351 on Build GRCh38
rs999228998 Chr.16: 5085332-5085332 on Build GRCh38
rs547607392 Chr.16: 5085110-5085110 on Build GRCh38
rs907193961 Chr.16: 5085321-5085321 on Build GRCh38
rs534404562 Chr.16: 5085380-5085380 on Build GRCh38
rs1047576 Chr.16: 5085417-5085417 on Build GRCh38
rs975079270 Chr.16: 5085086-5085086 on Build GRCh38
rs1012821076 Chr.16: 5085289-5085289 on Build GRCh38
rs368205784 Chr.16: 5085095-5085097 on Build GRCh38
rs1047573 Chr.16: 5085423-5085423 on Build GRCh38
rs1038180261 Chr.16: 5085265-5085265 on Build GRCh38
rs543353647 Chr.16: 5085463-5085463 on Build GRCh38
rs943596760 Chr.16: 5085069-5085069 on Build GRCh38
rs1047744 Chr.16: 5085077-5085077 on Build GRCh38
rs567329568 Chr.16: 5085131-5085131 on Build GRCh38
rs976303888 Chr.16: 5085427-5085427 on Build GRCh38
rs6775 Chr.16: 5085379-5085379 on Build GRCh38
rs1031759797 Chr.16: 5085352-5085352 on Build GRCh38
rs16843033 Chr.16: 5085462-5085462 on Build GRCh38
rs908247968 Chr.16: 5085171-5085173 on Build GRCh38
rs574276334 Chr.16: 5085461-5085461 on Build GRCh38
rs1017716654 Chr.16: 5085399-5085399 on Build GRCh38
rs527512215 Chr.16: 5085048-5085048 on Build GRCh38
rs941220621 Chr.16: 5085197-5085197 on Build GRCh38
rs1045801011 Chr.16: 5085299-5085299 on Build GRCh38
rs1011715831 Chr.16: 5085369-5085369 on Build GRCh38
rs933776146 Chr.16: 5085114-5085114 on Build GRCh38
rs149110534 Chr.16: 5085267-5085267 on Build GRCh38

View Full Product Details