Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 10761272-10761767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUBP1
Entrez Gene ID
4682
Gene Name
nucleotide binding protein 1
Gene Aliases
NBP, NBP1, NBP35
UniGene
Hs.81469
Gene Chromosome Location
Chr.16: 10743164-10769351 on Build GRCh38
Transcripts
NM_001278506.1 NM_001323594.1 NM_001323595.1 NM_001323596.1 NM_001323597.1 NM_002484.3 XM_017023252.1 NR_134901.1 NR_134902.1 NR_134903.1 NR_134904.1 NR_134905.1 NR_134906.1

Gene Symbol
TVP23A
Entrez Gene ID
780776
Gene Name
trans-golgi network vesicle protein 23 homolog A
Gene Aliases
FAM18A, YDR084C
UniGene
Hs.371576
Gene Chromosome Location
Chr.16: 10758034-10818902 on Build GRCh38
Transcripts
NM_001278506.1 NM_001323594.1 NM_001323595.1 NM_001323596.1 NM_001323597.1 NM_002484.3 XM_017023252.1 NR_134901.1 NR_134902.1 NR_134903.1 NR_134904.1 NR_134905.1 NR_134906.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7216014 c.717+10C>T p.? Chr.16: 10761484-10761484 on Build GRCh38
6429782 c.650G>A p.R217H Chr.16: 10761407-10761407 on Build GRCh38
966934 c.725C>A p.S242Y Chr.16: 10761764-10761764 on Build GRCh38
7201979 c.607-2A>T p.? Chr.16: 10761362-10761362 on Build GRCh38
3701038 c.607-4G>T p.? Chr.16: 10761360-10761360 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975577113 Chr.16: 10761369-10761369 on Build GRCh38
rs755223093 Chr.16: 10761477-10761477 on Build GRCh38
rs199557719 Chr.16: 10761737-10761737 on Build GRCh38
rs765283235 Chr.16: 10761300-10761300 on Build GRCh38
rs200931059 Chr.16: 10761462-10761462 on Build GRCh38
rs934554474 Chr.16: 10761436-10761436 on Build GRCh38
rs368752350 Chr.16: 10761385-10761385 on Build GRCh38
rs990703632 Chr.16: 10761481-10761481 on Build GRCh38
rs36097495 Chr.16: 10761678-10761678 on Build GRCh38
rs754936367 Chr.16: 10761386-10761386 on Build GRCh38
rs745819659 Chr.16: 10761433-10761433 on Build GRCh38
rs146921748 Chr.16: 10761299-10761299 on Build GRCh38
rs778653318 Chr.16: 10761430-10761430 on Build GRCh38
rs79754840 Chr.16: 10761665-10761665 on Build GRCh38
rs760375953 Chr.16: 10761409-10761409 on Build GRCh38
rs780183700 Chr.16: 10761319-10761319 on Build GRCh38
rs771665228 Chr.16: 10761439-10761439 on Build GRCh38
rs756397997 Chr.16: 10761735-10761735 on Build GRCh38
rs554915911 Chr.16: 10761472-10761472 on Build GRCh38
rs2233539 Chr.16: 10761484-10761484 on Build GRCh38
rs770830312 Chr.16: 10761328-10761328 on Build GRCh38
rs778858546 Chr.16: 10761665-10761665 on Build GRCh38
rs372881425 Chr.16: 10761726-10761726 on Build GRCh38
rs765648223 Chr.16: 10761464-10761464 on Build GRCh38
rs765734724 Chr.16: 10761398-10761398 on Build GRCh38
rs759050463 Chr.16: 10761355-10761355 on Build GRCh38
rs369337742 Chr.16: 10761485-10761485 on Build GRCh38
rs887541640 Chr.16: 10761554-10761554 on Build GRCh38
rs371836842 Chr.16: 10761480-10761480 on Build GRCh38
rs763434290 Chr.16: 10761707-10761707 on Build GRCh38
rs767145379 Chr.16: 10761356-10761356 on Build GRCh38
rs763534337 Chr.16: 10761430-10761430 on Build GRCh38
rs551882281 Chr.16: 10761307-10761307 on Build GRCh38
rs909536976 Chr.16: 10761345-10761345 on Build GRCh38
rs752063069 Chr.16: 10761723-10761723 on Build GRCh38
rs543363072 Chr.16: 10761637-10761637 on Build GRCh38
rs761553476 Chr.16: 10761502-10761502 on Build GRCh38
rs569894248 Chr.16: 10761457-10761457 on Build GRCh38
rs748985080 Chr.16: 10761731-10761731 on Build GRCh38
rs1014508687 Chr.16: 10761422-10761422 on Build GRCh38
rs951036653 Chr.16: 10761535-10761535 on Build GRCh38
rs779700267 Chr.16: 10761494-10761494 on Build GRCh38
rs927528421 Chr.16: 10761479-10761479 on Build GRCh38
rs758416439 Chr.16: 10761402-10761402 on Build GRCh38
rs781115964 Chr.16: 10761449-10761449 on Build GRCh38
rs745996416 Chr.16: 10761313-10761313 on Build GRCh38
rs1028041909 Chr.16: 10761694-10761694 on Build GRCh38
rs770765119 Chr.16: 10761424-10761424 on Build GRCh38
rs752341242 Chr.16: 10761396-10761396 on Build GRCh38
rs755431529 Chr.16: 10761627-10761631 on Build GRCh38
rs758406967 Chr.16: 10761742-10761742 on Build GRCh38
rs758619843 Chr.16: 10761315-10761315 on Build GRCh38
rs751694744 Chr.16: 10761476-10761476 on Build GRCh38
rs775178236 Chr.16: 10761442-10761442 on Build GRCh38
rs528477000 Chr.16: 10761664-10761664 on Build GRCh38
rs975081844 Chr.16: 10761674-10761674 on Build GRCh38
rs565315341 Chr.16: 10761483-10761483 on Build GRCh38
rs760384677 Chr.16: 10761447-10761447 on Build GRCh38
rs377298156 Chr.16: 10761348-10761348 on Build GRCh38
rs879199312 Chr.16: 10761685-10761685 on Build GRCh38
rs543949006 Chr.16: 10761532-10761532 on Build GRCh38
rs749103884 Chr.16: 10761419-10761419 on Build GRCh38
rs189499982 Chr.16: 10761644-10761644 on Build GRCh38
rs746585413 Chr.16: 10761495-10761495 on Build GRCh38
rs564409724 Chr.16: 10761335-10761335 on Build GRCh38
rs768254675 Chr.16: 10761456-10761456 on Build GRCh38
rs557280291 Chr.16: 10761556-10761556 on Build GRCh38
rs62027821 Chr.16: 10761663-10761663 on Build GRCh38
rs545110621 Chr.16: 10761670-10761670 on Build GRCh38
rs996400733 Chr.16: 10761329-10761329 on Build GRCh38
rs755240224 Chr.16: 10761412-10761412 on Build GRCh38
rs1027872750 Chr.16: 10761542-10761542 on Build GRCh38
rs777596593 Chr.16: 10761531-10761531 on Build GRCh38
rs971070406 Chr.16: 10761577-10761577 on Build GRCh38
rs753391157 Chr.16: 10761716-10761717 on Build GRCh38
rs749225198 Chr.16: 10761326-10761326 on Build GRCh38
rs778827686 Chr.16: 10761736-10761736 on Build GRCh38
rs2233538 Chr.16: 10761323-10761323 on Build GRCh38
rs1016483356 Chr.16: 10761555-10761555 on Build GRCh38
rs772913665 Chr.16: 10761325-10761325 on Build GRCh38
rs570096517 Chr.16: 10761488-10761488 on Build GRCh38
rs757099185 Chr.16: 10761725-10761726 on Build GRCh38
rs760431991 Chr.16: 10761360-10761360 on Build GRCh38
rs983092534 Chr.16: 10761421-10761421 on Build GRCh38
rs533138388 Chr.16: 10761407-10761407 on Build GRCh38
rs574348170 Chr.16: 10761629-10761629 on Build GRCh38
rs565336258 Chr.16: 10761678-10761678 on Build GRCh38
rs866565194 Chr.16: 10761311-10761311 on Build GRCh38
rs766936204 Chr.16: 10761714-10761714 on Build GRCh38
rs1040345593 Chr.16: 10761506-10761506 on Build GRCh38
rs750501348 Chr.16: 10761306-10761306 on Build GRCh38
rs1014623556 Chr.16: 10761560-10761560 on Build GRCh38
rs200400312 Chr.16: 10761468-10761468 on Build GRCh38
rs775231355 Chr.16: 10761359-10761359 on Build GRCh38
rs924559922 Chr.16: 10761427-10761427 on Build GRCh38
rs981057885 Chr.16: 10761624-10761624 on Build GRCh38
rs138217163 Chr.16: 10761429-10761429 on Build GRCh38
rs1041745551 Chr.16: 10761498-10761498 on Build GRCh38
rs747246242 Chr.16: 10761322-10761322 on Build GRCh38
rs752978296 Chr.16: 10761482-10761482 on Build GRCh38
rs777610049 Chr.16: 10761414-10761414 on Build GRCh38
rs780130696 Chr.16: 10761405-10761405 on Build GRCh38
rs752822396 Chr.16: 10761733-10761733 on Build GRCh38
rs767909676 Chr.16: 10761356-10761358 on Build GRCh38
rs577710170 Chr.16: 10761500-10761500 on Build GRCh38
rs770018086 Chr.16: 10761514-10761514 on Build GRCh38
rs773609985 Chr.16: 10761520-10761520 on Build GRCh38
rs1006877203 Chr.16: 10761536-10761536 on Build GRCh38
rs768217453 Chr.16: 10761499-10761499 on Build GRCh38

View Full Product Details