Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 10771587-10772096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TVP23A
Entrez Gene ID
780776
Gene Name
trans-golgi network vesicle protein 23 homolog A
Gene Aliases
FAM18A, YDR084C
UniGene
Hs.371576
Gene Chromosome Location
Chr.16: 10758034-10818902 on Build GRCh38
Transcripts
NM_001079512.3 NM_001318873.1 NR_134901.1 NR_134902.1 NR_134903.1 NR_134904.1 NR_134905.1 NR_134906.1 XM_006720944.3 XM_011522654.2 XM_017023649.1 XM_017023650.1 XM_017023651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3957103 c.558C>G p.F186L Chr.16: 10771694-10771694 on Build GRCh38
5677171 c.485C>T p.A162V Chr.16: 10771767-10771767 on Build GRCh38
6333863 c.576C>A p.F192L Chr.16: 10771676-10771676 on Build GRCh38
4425608 c.512G>T p.C171F Chr.16: 10771740-10771740 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs35752179 Chr.16: 10772044-10772045 on Build GRCh38
rs139056128 Chr.16: 10771857-10771857 on Build GRCh38
rs899289811 Chr.16: 10771958-10771958 on Build GRCh38
rs759928215 Chr.16: 10771776-10771776 on Build GRCh38
rs386788940 Chr.16: 10771919-10771920 on Build GRCh38
rs962712256 Chr.16: 10771964-10771964 on Build GRCh38
rs763485294 Chr.16: 10771796-10771796 on Build GRCh38
rs1006992722 Chr.16: 10771946-10771946 on Build GRCh38
rs371286920 Chr.16: 10771842-10771842 on Build GRCh38
rs771172564 Chr.16: 10771645-10771645 on Build GRCh38
rs760027687 Chr.16: 10771829-10771829 on Build GRCh38
rs760408687 Chr.16: 10771660-10771660 on Build GRCh38
rs913020475 Chr.16: 10771823-10771823 on Build GRCh38
rs757805880 Chr.16: 10771740-10771740 on Build GRCh38
rs573784908 Chr.16: 10771850-10771850 on Build GRCh38
rs2697659 Chr.16: 10772011-10772011 on Build GRCh38
rs368223957 Chr.16: 10771751-10771751 on Build GRCh38
rs993138878 Chr.16: 10772069-10772069 on Build GRCh38
rs896015614 Chr.16: 10771617-10771617 on Build GRCh38
rs1057001658 Chr.16: 10771829-10771829 on Build GRCh38
rs536972636 Chr.16: 10771627-10771627 on Build GRCh38
rs374042646 Chr.16: 10771684-10771684 on Build GRCh38
rs1006557006 Chr.16: 10772009-10772009 on Build GRCh38
rs950644450 Chr.16: 10772022-10772022 on Build GRCh38
rs773665087 Chr.16: 10771795-10771795 on Build GRCh38
rs954343761 Chr.16: 10771945-10771945 on Build GRCh38
rs201138145 Chr.16: 10771696-10771696 on Build GRCh38
rs751697835 Chr.16: 10771693-10771693 on Build GRCh38
rs572870725 Chr.16: 10771905-10771905 on Build GRCh38
rs962678210 Chr.16: 10771698-10771698 on Build GRCh38
rs1000428908 Chr.16: 10771636-10771636 on Build GRCh38
rs768873957 Chr.16: 10771667-10771667 on Build GRCh38
rs919141393 Chr.16: 10771784-10771784 on Build GRCh38
rs753748763 Chr.16: 10771709-10771709 on Build GRCh38
rs1017000714 Chr.16: 10771954-10771954 on Build GRCh38
rs1029221336 Chr.16: 10771975-10771975 on Build GRCh38
rs550865251 Chr.16: 10771944-10771944 on Build GRCh38
rs567444021 Chr.16: 10772008-10772008 on Build GRCh38
rs368036929 Chr.16: 10771683-10771683 on Build GRCh38
rs371948396 Chr.16: 10771767-10771767 on Build GRCh38
rs953671910 Chr.16: 10771741-10771741 on Build GRCh38
rs771294224 Chr.16: 10771731-10771731 on Build GRCh38
rs948525735 Chr.16: 10771870-10771870 on Build GRCh38
rs867988876 Chr.16: 10771669-10771669 on Build GRCh38
rs559427410 Chr.16: 10771858-10771858 on Build GRCh38
rs766343600 Chr.16: 10771797-10771797 on Build GRCh38
rs538694589 Chr.16: 10771708-10771708 on Build GRCh38
rs371841589 Chr.16: 10772071-10772071 on Build GRCh38
rs369163539 Chr.16: 10771805-10771805 on Build GRCh38
rs34216148 Chr.16: 10772045-10772046 on Build GRCh38
rs1044521863 Chr.16: 10771901-10771901 on Build GRCh38
rs778560358 Chr.16: 10771634-10771634 on Build GRCh38
rs553416754 Chr.16: 10771670-10771670 on Build GRCh38
rs998333393 Chr.16: 10771652-10771652 on Build GRCh38
rs770159381 Chr.16: 10771783-10771783 on Build GRCh38
rs1043342924 Chr.16: 10771882-10771882 on Build GRCh38
rs386788941 Chr.16: 10771920-10771921 on Build GRCh38
rs913082847 Chr.16: 10772042-10772042 on Build GRCh38
rs767710338 Chr.16: 10771811-10771811 on Build GRCh38
rs774891008 Chr.16: 10771787-10771788 on Build GRCh38
rs771046015 Chr.16: 10771632-10771632 on Build GRCh38
rs778936268 Chr.16: 10771718-10771718 on Build GRCh38
rs2697658 Chr.16: 10771920-10771920 on Build GRCh38
rs765804368 Chr.16: 10771676-10771676 on Build GRCh38
rs749154951 Chr.16: 10771630-10771630 on Build GRCh38
rs762295494 Chr.16: 10771671-10771671 on Build GRCh38
rs775049145 Chr.16: 10771723-10771723 on Build GRCh38
rs1053311079 Chr.16: 10771912-10771912 on Build GRCh38
rs985221806 Chr.16: 10771771-10771771 on Build GRCh38
rs771718620 Chr.16: 10771656-10771656 on Build GRCh38
rs749407128 Chr.16: 10771642-10771642 on Build GRCh38
rs141087240 Chr.16: 10771921-10771921 on Build GRCh38
rs536819580 Chr.16: 10771801-10771801 on Build GRCh38
rs755819298 Chr.16: 10771623-10771623 on Build GRCh38
rs757122452 Chr.16: 10771713-10771713 on Build GRCh38
rs965163880 Chr.16: 10772013-10772013 on Build GRCh38
rs564688730 Chr.16: 10771724-10771724 on Build GRCh38
rs558580884 Chr.16: 10771750-10771750 on Build GRCh38
rs200073480 Chr.16: 10772042-10772043 on Build GRCh38
rs751822305 Chr.16: 10771622-10771622 on Build GRCh38
rs575346575 Chr.16: 10771773-10771773 on Build GRCh38
rs146145462 Chr.16: 10772043-10772044 on Build GRCh38
rs368782664 Chr.16: 10771821-10771821 on Build GRCh38
rs767985487 Chr.16: 10771826-10771826 on Build GRCh38
rs745822000 Chr.16: 10771722-10771722 on Build GRCh38
rs8051333 Chr.16: 10771919-10771919 on Build GRCh38
rs772411270 Chr.16: 10771841-10771842 on Build GRCh38
rs572649501 Chr.16: 10771646-10771646 on Build GRCh38
rs904217348 Chr.16: 10771904-10771904 on Build GRCh38
rs776856112 Chr.16: 10771769-10771769 on Build GRCh38

View Full Product Details