Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 10902416-10902957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIITA
Entrez Gene ID
4261
Gene Name
class II, major histocompatibility complex, transactivator
Gene Aliases
C2TA, CIITAIV, MHC2TA, NLRA
UniGene
Hs.592051
Gene Chromosome Location
Chr.16: 10866208-10941562 on Build GRCh38
Transcripts
NM_000246.3 NM_001286402.1 NM_001286403.1 NR_104444.1 XM_006720880.3 XM_011522484.2 XM_011522485.2 XM_011522486.2 XM_011522487.2 XM_011522488.2 XM_011522489.2 XM_011522490.2 XM_011522491.2 XM_011522492.2 XM_011522493.2 XM_011522494.2 XM_011522495.2 XM_011522496.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
702008 c.644C>G p.S215C Chr.16: 10902673-10902673 on Build GRCh38
2141919 c.754A>C p.S252R Chr.16: 10902783-10902783 on Build GRCh38
3505736 c.729C>T p.I243I Chr.16: 10902758-10902758 on Build GRCh38
702007 c.723G>A p.W241* Chr.16: 10902752-10902752 on Build GRCh38
3505735 c.674G>A p.G225E Chr.16: 10902703-10902703 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs960991583 Chr.16: 10902928-10902928 on Build GRCh38
rs147472574 Chr.16: 10902720-10902720 on Build GRCh38
rs745425345 Chr.16: 10902846-10902846 on Build GRCh38
rs8045125 Chr.16: 10902592-10902592 on Build GRCh38
rs1056990364 Chr.16: 10902590-10902590 on Build GRCh38
rs773897183 Chr.16: 10902678-10902678 on Build GRCh38
rs545529593 Chr.16: 10902594-10902594 on Build GRCh38
rs1022665349 Chr.16: 10902597-10902597 on Build GRCh38
rs565678751 Chr.16: 10902773-10902773 on Build GRCh38
rs777294112 Chr.16: 10902803-10902803 on Build GRCh38
rs149554407 Chr.16: 10902813-10902813 on Build GRCh38
rs35407222 Chr.16: 10902567-10902567 on Build GRCh38
rs745359262 Chr.16: 10902753-10902753 on Build GRCh38
rs755101452 Chr.16: 10902485-10902485 on Build GRCh38
rs775983190 Chr.16: 10902686-10902686 on Build GRCh38
rs895271987 Chr.16: 10902897-10902897 on Build GRCh38
rs374178689 Chr.16: 10902472-10902472 on Build GRCh38
rs757381623 Chr.16: 10902612-10902612 on Build GRCh38
rs371896170 Chr.16: 10902927-10902927 on Build GRCh38
rs954347047 Chr.16: 10902676-10902676 on Build GRCh38
rs1053789672 Chr.16: 10902851-10902851 on Build GRCh38
rs11640448 Chr.16: 10902572-10902572 on Build GRCh38
rs1031267583 Chr.16: 10902635-10902635 on Build GRCh38
rs750572482 Chr.16: 10902810-10902810 on Build GRCh38
rs757438915 Chr.16: 10902705-10902705 on Build GRCh38
rs771443378 Chr.16: 10902849-10902849 on Build GRCh38
rs751737619 Chr.16: 10902814-10902814 on Build GRCh38
rs985933995 Chr.16: 10902519-10902519 on Build GRCh38
rs372749477 Chr.16: 10902649-10902649 on Build GRCh38
rs144398594 Chr.16: 10902830-10902830 on Build GRCh38
rs573965744 Chr.16: 10902567-10902567 on Build GRCh38
rs758710934 Chr.16: 10902715-10902715 on Build GRCh38
rs972800956 Chr.16: 10902906-10902906 on Build GRCh38
rs35170497 Chr.16: 10902825-10902825 on Build GRCh38
rs200418706 Chr.16: 10902619-10902619 on Build GRCh38
rs369197968 Chr.16: 10902683-10902683 on Build GRCh38
rs138110525 Chr.16: 10902559-10902559 on Build GRCh38
rs564243852 Chr.16: 10902606-10902606 on Build GRCh38
rs540159423 Chr.16: 10902884-10902884 on Build GRCh38
rs943020267 Chr.16: 10902571-10902571 on Build GRCh38
rs902367424 Chr.16: 10902598-10902598 on Build GRCh38
rs1031884541 Chr.16: 10902461-10902461 on Build GRCh38
rs1046863572 Chr.16: 10902901-10902901 on Build GRCh38
rs766614231 Chr.16: 10902467-10902467 on Build GRCh38
rs267604407 Chr.16: 10902689-10902689 on Build GRCh38
rs369527269 Chr.16: 10902807-10902807 on Build GRCh38
rs4781016 Chr.16: 10902542-10902542 on Build GRCh38
rs765306236 Chr.16: 10902709-10902709 on Build GRCh38
rs962812917 Chr.16: 10902896-10902896 on Build GRCh38
rs909924114 Chr.16: 10902527-10902527 on Build GRCh38
rs906550015 Chr.16: 10902903-10902903 on Build GRCh38
rs372545934 Chr.16: 10902618-10902618 on Build GRCh38
rs150307400 Chr.16: 10902741-10902741 on Build GRCh38
rs528875833 Chr.16: 10902739-10902739 on Build GRCh38
rs750616786 Chr.16: 10902710-10902710 on Build GRCh38
rs754021708 Chr.16: 10902611-10902611 on Build GRCh38
rs369695610 Chr.16: 10902822-10902822 on Build GRCh38
rs932773828 Chr.16: 10902494-10902494 on Build GRCh38
rs904240303 Chr.16: 10902605-10902605 on Build GRCh38
rs374843831 Chr.16: 10902677-10902677 on Build GRCh38
rs773842701 Chr.16: 10902749-10902749 on Build GRCh38
rs780977067 Chr.16: 10902644-10902644 on Build GRCh38
rs557455283 Chr.16: 10902809-10902809 on Build GRCh38
rs533849173 Chr.16: 10902566-10902567 on Build GRCh38
rs1030330045 Chr.16: 10902826-10902826 on Build GRCh38
rs143152174 Chr.16: 10902674-10902674 on Build GRCh38
rs766452566 Chr.16: 10902810-10902810 on Build GRCh38
rs764509469 Chr.16: 10902797-10902797 on Build GRCh38
rs373996375 Chr.16: 10902824-10902824 on Build GRCh38
rs868112925 Chr.16: 10902752-10902752 on Build GRCh38
rs773108670 Chr.16: 10902666-10902666 on Build GRCh38
rs553052381 Chr.16: 10902873-10902873 on Build GRCh38
rs761152169 Chr.16: 10902787-10902787 on Build GRCh38
rs537284369 Chr.16: 10902819-10902819 on Build GRCh38
rs368422088 Chr.16: 10902634-10902634 on Build GRCh38
rs759168388 Chr.16: 10902680-10902680 on Build GRCh38
rs6498125 Chr.16: 10902475-10902475 on Build GRCh38
rs138147861 Chr.16: 10902805-10902805 on Build GRCh38
rs376221650 Chr.16: 10902806-10902806 on Build GRCh38
rs771807788 Chr.16: 10902767-10902767 on Build GRCh38
rs371653584 Chr.16: 10902703-10902703 on Build GRCh38
rs557888281 Chr.16: 10902451-10902451 on Build GRCh38
rs779875908 Chr.16: 10902850-10902850 on Build GRCh38
rs1017090777 Chr.16: 10902892-10902892 on Build GRCh38
rs564988266 Chr.16: 10902523-10902523 on Build GRCh38
rs956308767 Chr.16: 10902645-10902645 on Build GRCh38
rs761100720 Chr.16: 10902697-10902697 on Build GRCh38
rs754357271 Chr.16: 10902704-10902704 on Build GRCh38
rs543087884 Chr.16: 10902916-10902916 on Build GRCh38
rs918627823 Chr.16: 10902917-10902917 on Build GRCh38
rs1012228445 Chr.16: 10902898-10902898 on Build GRCh38
rs770360091 Chr.16: 10902747-10902747 on Build GRCh38
rs747982404 Chr.16: 10902727-10902727 on Build GRCh38
rs201764329 Chr.16: 10902660-10902660 on Build GRCh38
rs918575509 Chr.16: 10902721-10902721 on Build GRCh38
rs897759203 Chr.16: 10902568-10902568 on Build GRCh38

View Full Product Details