Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 10971043-10971547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC16A
Entrez Gene ID
23274
Gene Name
C-type lectin domain family 16 member A
Gene Aliases
Gop-1, KIAA0350
UniGene
Hs.35490
Gene Chromosome Location
Chr.16: 10944488-11193272 on Build GRCh38
Transcripts
NM_001243403.1 NM_015226.2 XM_005255210.1 XM_005255211.1 XM_005255213.1 XM_005255214.1 XM_005255215.3 XM_005255216.1 XM_006720870.3 XM_011522434.1 XM_011522435.1 XM_011522436.2 XM_011522437.2 XM_011522438.2 XM_011522439.2 XM_011522440.2 XM_017023089.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6630114 c.523G>A p.A175T Chr.16: 10971155-10971155 on Build GRCh38
6143553 c.597A>T p.K199N Chr.16: 10971229-10971229 on Build GRCh38
4640034 c.513G>A p.L171L Chr.16: 10971145-10971145 on Build GRCh38
4635933 c.552C>T p.S184S Chr.16: 10971184-10971184 on Build GRCh38
7232359 c.537C>G p.F179L Chr.16: 10971169-10971169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201695473 Chr.16: 10971244-10971244 on Build GRCh38
rs568057166 Chr.16: 10971249-10971249 on Build GRCh38
rs114035917 Chr.16: 10971299-10971299 on Build GRCh38
rs116481474 Chr.16: 10971304-10971304 on Build GRCh38
rs964084227 Chr.16: 10971163-10971163 on Build GRCh38
rs199890590 Chr.16: 10971258-10971258 on Build GRCh38
rs759604074 Chr.16: 10971172-10971172 on Build GRCh38
rs982982412 Chr.16: 10971455-10971455 on Build GRCh38
rs201211737 Chr.16: 10971259-10971259 on Build GRCh38
rs752669267 Chr.16: 10971483-10971483 on Build GRCh38
rs918191968 Chr.16: 10971290-10971290 on Build GRCh38
rs75388786 Chr.16: 10971510-10971510 on Build GRCh38
rs774299916 Chr.16: 10971171-10971171 on Build GRCh38
rs763222619 Chr.16: 10971231-10971232 on Build GRCh38
rs201761542 Chr.16: 10971275-10971275 on Build GRCh38
rs115906116 Chr.16: 10971349-10971349 on Build GRCh38
rs567150021 Chr.16: 10971257-10971257 on Build GRCh38
rs139651990 Chr.16: 10971433-10971433 on Build GRCh38
rs760996776 Chr.16: 10971279-10971279 on Build GRCh38
rs546276010 Chr.16: 10971301-10971301 on Build GRCh38
rs1018214855 Chr.16: 10971394-10971394 on Build GRCh38
rs867691682 Chr.16: 10971207-10971207 on Build GRCh38
rs747798193 Chr.16: 10971117-10971117 on Build GRCh38
rs772317085 Chr.16: 10971268-10971268 on Build GRCh38
rs559003745 Chr.16: 10971348-10971348 on Build GRCh38
rs763394459 Chr.16: 10971140-10971140 on Build GRCh38
rs753032767 Chr.16: 10971200-10971200 on Build GRCh38
rs868458091 Chr.16: 10971419-10971419 on Build GRCh38
rs759956126 Chr.16: 10971265-10971265 on Build GRCh38
rs192972852 Chr.16: 10971233-10971233 on Build GRCh38
rs1018285498 Chr.16: 10971139-10971139 on Build GRCh38
rs757175950 Chr.16: 10971076-10971076 on Build GRCh38
rs776327190 Chr.16: 10971109-10971109 on Build GRCh38
rs773546785 Chr.16: 10971136-10971136 on Build GRCh38
rs183861674 Chr.16: 10971361-10971361 on Build GRCh38
rs778844500 Chr.16: 10971082-10971082 on Build GRCh38
rs538774095 Chr.16: 10971454-10971454 on Build GRCh38
rs758331636 Chr.16: 10971087-10971087 on Build GRCh38
rs995543019 Chr.16: 10971442-10971442 on Build GRCh38
rs758294558 Chr.16: 10971234-10971234 on Build GRCh38
rs545152174 Chr.16: 10971519-10971519 on Build GRCh38
rs200404847 Chr.16: 10971094-10971094 on Build GRCh38
rs949041452 Chr.16: 10971293-10971293 on Build GRCh38
rs983398131 Chr.16: 10971274-10971274 on Build GRCh38
rs760947013 Chr.16: 10971211-10971211 on Build GRCh38
rs998996249 Chr.16: 10971446-10971446 on Build GRCh38
rs1040147348 Chr.16: 10971400-10971400 on Build GRCh38
rs936328780 Chr.16: 10971498-10971498 on Build GRCh38
rs1053567904 Chr.16: 10971336-10971336 on Build GRCh38
rs533833103 Chr.16: 10971303-10971303 on Build GRCh38
rs921266395 Chr.16: 10971403-10971403 on Build GRCh38
rs762362514 Chr.16: 10971291-10971291 on Build GRCh38
rs1007460252 Chr.16: 10971372-10971372 on Build GRCh38
rs199868443 Chr.16: 10971106-10971106 on Build GRCh38
rs542874615 Chr.16: 10971482-10971482 on Build GRCh38
rs369393972 Chr.16: 10971086-10971086 on Build GRCh38
rs969070327 Chr.16: 10971520-10971520 on Build GRCh38
rs554994812 Chr.16: 10971405-10971405 on Build GRCh38
rs373282181 Chr.16: 10971316-10971316 on Build GRCh38
rs72650684 Chr.16: 10971141-10971141 on Build GRCh38
rs200366711 Chr.16: 10971093-10971093 on Build GRCh38
rs779970758 Chr.16: 10971236-10971236 on Build GRCh38
rs934265264 Chr.16: 10971414-10971414 on Build GRCh38
rs16957849 Chr.16: 10971450-10971450 on Build GRCh38
rs553095365 Chr.16: 10971364-10971364 on Build GRCh38
rs989189092 Chr.16: 10971503-10971503 on Build GRCh38
rs76368782 Chr.16: 10971460-10971460 on Build GRCh38
rs771431541 Chr.16: 10971151-10971151 on Build GRCh38
rs530438782 Chr.16: 10971215-10971215 on Build GRCh38
rs981059755 Chr.16: 10971468-10971468 on Build GRCh38
rs1025590198 Chr.16: 10971458-10971458 on Build GRCh38
rs909902243 Chr.16: 10971143-10971143 on Build GRCh38
rs1010955983 Chr.16: 10971307-10971307 on Build GRCh38
rs998444598 Chr.16: 10971283-10971286 on Build GRCh38
rs1051335245 Chr.16: 10971431-10971431 on Build GRCh38
rs770186127 Chr.16: 10971123-10971123 on Build GRCh38
rs200906965 Chr.16: 10971243-10971243 on Build GRCh38
rs371603009 Chr.16: 10971261-10971261 on Build GRCh38
rs767691187 Chr.16: 10971176-10971176 on Build GRCh38
rs201388822 Chr.16: 10971105-10971105 on Build GRCh38

View Full Product Details