Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 11020064-11020555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC16A
Entrez Gene ID
23274
Gene Name
C-type lectin domain family 16 member A
Gene Aliases
Gop-1, KIAA0350
UniGene
Hs.35490
Gene Chromosome Location
Chr.16: 10944488-11193272 on Build GRCh38
Transcripts
NM_001243403.1 NM_015226.2 XM_005255210.1 XM_005255211.1 XM_005255213.1 XM_005255214.1 XM_005255215.3 XM_005255216.1 XM_006720870.3 XM_011522434.1 XM_011522435.1 XM_011522436.2 XM_011522437.2 XM_011522438.2 XM_011522439.2 XM_011522440.2 XM_017023089.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3690726 c.1402G>A p.A468T Chr.16: 11020291-11020291 on Build GRCh38
7320197 c.1404C>T p.A468A Chr.16: 11020293-11020293 on Build GRCh38
7320198 c.1350C>T p.A450A Chr.16: 11020293-11020293 on Build GRCh38
5042780 c.1273C>T p.R425C Chr.16: 11020216-11020216 on Build GRCh38
7193700 c.1314G>T p.Q438H Chr.16: 11020257-11020257 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201477336 Chr.16: 11020321-11020321 on Build GRCh38
rs372006524 Chr.16: 11020318-11020318 on Build GRCh38
rs977936610 Chr.16: 11020107-11020107 on Build GRCh38
rs375520553 Chr.16: 11020324-11020324 on Build GRCh38
rs201357891 Chr.16: 11020344-11020344 on Build GRCh38
rs1052089557 Chr.16: 11020116-11020116 on Build GRCh38
rs746938479 Chr.16: 11020220-11020220 on Build GRCh38
rs199574791 Chr.16: 11020213-11020213 on Build GRCh38
rs76611790 Chr.16: 11020272-11020272 on Build GRCh38
rs766735356 Chr.16: 11020191-11020191 on Build GRCh38
rs376798629 Chr.16: 11020085-11020085 on Build GRCh38
rs1032817251 Chr.16: 11020466-11020466 on Build GRCh38
rs753870343 Chr.16: 11020338-11020338 on Build GRCh38
rs749505638 Chr.16: 11020214-11020214 on Build GRCh38
rs879041315 Chr.16: 11020166-11020166 on Build GRCh38
rs916172447 Chr.16: 11020129-11020129 on Build GRCh38
rs552988990 Chr.16: 11020146-11020146 on Build GRCh38
rs765440972 Chr.16: 11020342-11020342 on Build GRCh38
rs778029439 Chr.16: 11020205-11020205 on Build GRCh38
rs762674209 Chr.16: 11020248-11020248 on Build GRCh38
rs61741309 Chr.16: 11020317-11020317 on Build GRCh38
rs979993768 Chr.16: 11020297-11020297 on Build GRCh38
rs781113229 Chr.16: 11020347-11020347 on Build GRCh38
rs752056185 Chr.16: 11020197-11020197 on Build GRCh38
rs201716099 Chr.16: 11020249-11020249 on Build GRCh38
rs201162849 Chr.16: 11020294-11020294 on Build GRCh38
rs1037578677 Chr.16: 11020238-11020238 on Build GRCh38
rs779744241 Chr.16: 11020288-11020288 on Build GRCh38
rs76668113 Chr.16: 11020430-11020430 on Build GRCh38
rs773723084 Chr.16: 11020179-11020179 on Build GRCh38
rs1023661350 Chr.16: 11020460-11020460 on Build GRCh38
rs200678636 Chr.16: 11020180-11020180 on Build GRCh38
rs752711166 Chr.16: 11020201-11020201 on Build GRCh38
rs915989217 Chr.16: 11020501-11020501 on Build GRCh38
rs778696765 Chr.16: 11020367-11020367 on Build GRCh38
rs888876517 Chr.16: 11020189-11020189 on Build GRCh38
rs1030578173 Chr.16: 11020524-11020524 on Build GRCh38
rs1037924438 Chr.16: 11020409-11020409 on Build GRCh38
rs200493825 Chr.16: 11020336-11020336 on Build GRCh38
rs1032860732 Chr.16: 11020496-11020496 on Build GRCh38
rs991594518 Chr.16: 11020493-11020493 on Build GRCh38
rs776285118 Chr.16: 11020222-11020222 on Build GRCh38
rs200123237 Chr.16: 11020275-11020275 on Build GRCh38
rs910290804 Chr.16: 11020360-11020360 on Build GRCh38
rs999103355 Chr.16: 11020457-11020457 on Build GRCh38
rs2286975 Chr.16: 11020149-11020149 on Build GRCh38
rs898025274 Chr.16: 11020159-11020159 on Build GRCh38
rs750359729 Chr.16: 11020511-11020512 on Build GRCh38
rs117039506 Chr.16: 11020429-11020429 on Build GRCh38
rs76222412 Chr.16: 11020122-11020122 on Build GRCh38
rs757448042 Chr.16: 11020278-11020278 on Build GRCh38
rs754883882 Chr.16: 11020291-11020291 on Build GRCh38
rs544361281 Chr.16: 11020517-11020517 on Build GRCh38
rs111796202 Chr.16: 11020120-11020120 on Build GRCh38
rs769479554 Chr.16: 11020311-11020311 on Build GRCh38
rs748778935 Chr.16: 11020364-11020364 on Build GRCh38
rs545012360 Chr.16: 11020505-11020505 on Build GRCh38
rs766763248 Chr.16: 11020400-11020400 on Build GRCh38
rs755380312 Chr.16: 11020534-11020534 on Build GRCh38
rs771884767 Chr.16: 11020108-11020108 on Build GRCh38
rs1049196473 Chr.16: 11020390-11020390 on Build GRCh38
rs950018419 Chr.16: 11020357-11020357 on Build GRCh38
rs759968559 Chr.16: 11020252-11020252 on Build GRCh38
rs189251752 Chr.16: 11020479-11020479 on Build GRCh38
rs752705678 Chr.16: 11020348-11020348 on Build GRCh38
rs766675571 Chr.16: 11020250-11020250 on Build GRCh38
rs766621712 Chr.16: 11020287-11020289 on Build GRCh38
rs74163614 Chr.16: 11020313-11020313 on Build GRCh38
rs950251021 Chr.16: 11020133-11020133 on Build GRCh38
rs201072027 Chr.16: 11020287-11020287 on Build GRCh38
rs371915032 Chr.16: 11020216-11020216 on Build GRCh38
rs941812975 Chr.16: 11020374-11020374 on Build GRCh38
rs756250093 Chr.16: 11020268-11020268 on Build GRCh38
rs536862590 Chr.16: 11020102-11020102 on Build GRCh38
rs374800849 Chr.16: 11020359-11020359 on Build GRCh38
rs981400818 Chr.16: 11020509-11020509 on Build GRCh38
rs777429548 Chr.16: 11020361-11020361 on Build GRCh38
rs370546327 Chr.16: 11020299-11020299 on Build GRCh38
rs1000930475 Chr.16: 11020483-11020483 on Build GRCh38
rs1039412956 Chr.16: 11020418-11020418 on Build GRCh38
rs573022662 Chr.16: 11020498-11020498 on Build GRCh38
rs558543797 Chr.16: 11020507-11020507 on Build GRCh38
rs796504796 Chr.16: 11020224-11020224 on Build GRCh38
rs899535245 Chr.16: 11020422-11020422 on Build GRCh38
rs756345867 Chr.16: 11020203-11020203 on Build GRCh38
rs772546016 Chr.16: 11020343-11020343 on Build GRCh38
rs200731011 Chr.16: 11020241-11020241 on Build GRCh38
rs761944553 Chr.16: 11020339-11020339 on Build GRCh38
rs201461895 Chr.16: 11020198-11020198 on Build GRCh38
rs768597834 Chr.16: 11020221-11020221 on Build GRCh38
rs567170949 Chr.16: 11020456-11020456 on Build GRCh38
rs201325973 Chr.16: 11020257-11020257 on Build GRCh38
rs201881116 Chr.16: 11020239-11020239 on Build GRCh38
rs200984316 Chr.16: 11020365-11020365 on Build GRCh38
rs751592742 Chr.16: 11020305-11020305 on Build GRCh38
rs895771736 Chr.16: 11020453-11020453 on Build GRCh38
rs548500656 Chr.16: 11020508-11020508 on Build GRCh38
rs575409948 Chr.16: 11020396-11020396 on Build GRCh38
rs80034801 Chr.16: 11020290-11020290 on Build GRCh38
rs200120297 Chr.16: 11020240-11020240 on Build GRCh38
rs200372176 Chr.16: 11020293-11020293 on Build GRCh38
rs184454129 Chr.16: 11020444-11020444 on Build GRCh38
rs985379804 Chr.16: 11020522-11020522 on Build GRCh38
rs536037957 Chr.16: 11020473-11020473 on Build GRCh38
rs111322660 Chr.16: 11020121-11020121 on Build GRCh38
rs748925258 Chr.16: 11020314-11020314 on Build GRCh38
rs76837940 Chr.16: 11020251-11020251 on Build GRCh38
rs564328177 Chr.16: 11020308-11020308 on Build GRCh38
rs753191649 Chr.16: 11020267-11020267 on Build GRCh38
rs888207544 Chr.16: 11020398-11020398 on Build GRCh38
rs575419018 Chr.16: 11020513-11020513 on Build GRCh38
rs199544900 Chr.16: 11020171-11020171 on Build GRCh38
rs72650660 Chr.16: 11020335-11020335 on Build GRCh38
rs760006232 Chr.16: 11020322-11020322 on Build GRCh38
rs201763346 Chr.16: 11020217-11020217 on Build GRCh38

View Full Product Details