Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 22007936-22008472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf52
Entrez Gene ID
730094
Gene Name
chromosome 16 open reading frame 52
Gene Aliases
-
UniGene
Hs.498890
Gene Chromosome Location
Chr.16: 22008135-22084654 on Build GRCh38
Transcripts
NM_001164579.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6988554 c.62C>T p.A21V Chr.16: 22008363-22008363 on Build GRCh38
6389263 c.76C>A p.P26T Chr.16: 22008377-22008377 on Build GRCh38
4769888 c.60C>G p.I20M Chr.16: 22008361-22008361 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1000154987 Chr.16: 22008034-22008034 on Build GRCh38
rs1014964980 Chr.16: 22008220-22008220 on Build GRCh38
rs945169149 Chr.16: 22008288-22008288 on Build GRCh38
rs575652054 Chr.16: 22008448-22008448 on Build GRCh38
rs865950864 Chr.16: 22008029-22008029 on Build GRCh38
rs991195157 Chr.16: 22008122-22008122 on Build GRCh38
rs977979323 Chr.16: 22008290-22008290 on Build GRCh38
rs1029020160 Chr.16: 22008054-22008054 on Build GRCh38
rs368259073 Chr.16: 22008332-22008332 on Build GRCh38
rs771934088 Chr.16: 22008394-22008394 on Build GRCh38
rs901836413 Chr.16: 22008036-22008036 on Build GRCh38
rs981394781 Chr.16: 22007983-22007984 on Build GRCh38
rs751264997 Chr.16: 22008328-22008328 on Build GRCh38
rs1035998835 Chr.16: 22008311-22008311 on Build GRCh38
rs928604907 Chr.16: 22008116-22008116 on Build GRCh38
rs770601867 Chr.16: 22008356-22008356 on Build GRCh38
rs935550068 Chr.16: 22008173-22008173 on Build GRCh38
rs952152859 Chr.16: 22008057-22008057 on Build GRCh38
rs1002739855 Chr.16: 22008287-22008287 on Build GRCh38
rs758868178 Chr.16: 22008274-22008275 on Build GRCh38
rs748745731 Chr.16: 22008453-22008453 on Build GRCh38
rs747663382 Chr.16: 22008407-22008408 on Build GRCh38
rs777480153 Chr.16: 22008449-22008449 on Build GRCh38
rs755333736 Chr.16: 22008426-22008426 on Build GRCh38
rs780836573 Chr.16: 22008412-22008413 on Build GRCh38
rs759307764 Chr.16: 22008376-22008376 on Build GRCh38
rs1006279474 Chr.16: 22008080-22008080 on Build GRCh38
rs968378922 Chr.16: 22008380-22008380 on Build GRCh38
rs907777359 Chr.16: 22007959-22007959 on Build GRCh38
rs1041498853 Chr.16: 22008141-22008141 on Build GRCh38
rs754650320 Chr.16: 22008394-22008395 on Build GRCh38
rs996223772 Chr.16: 22008040-22008040 on Build GRCh38
rs1041017557 Chr.16: 22008180-22008180 on Build GRCh38
rs914260443 Chr.16: 22008134-22008134 on Build GRCh38
rs765342238 Chr.16: 22008413-22008413 on Build GRCh38
rs961912364 Chr.16: 22008086-22008086 on Build GRCh38
rs772828934 Chr.16: 22008282-22008282 on Build GRCh38
rs1006227330 Chr.16: 22008217-22008217 on Build GRCh38
rs769381807 Chr.16: 22008281-22008281 on Build GRCh38
rs780642452 Chr.16: 22008408-22008409 on Build GRCh38
rs1040634356 Chr.16: 22008035-22008035 on Build GRCh38
rs931955725 Chr.16: 22008192-22008192 on Build GRCh38
rs980670328 Chr.16: 22007972-22007972 on Build GRCh38
rs959045010 Chr.16: 22007987-22007988 on Build GRCh38
rs981725682 Chr.16: 22008088-22008088 on Build GRCh38
rs571550762 Chr.16: 22008016-22008016 on Build GRCh38
rs960368696 Chr.16: 22008236-22008236 on Build GRCh38
rs538496858 Chr.16: 22008018-22008018 on Build GRCh38
rs964966038 Chr.16: 22008427-22008427 on Build GRCh38
rs765662882 Chr.16: 22008410-22008411 on Build GRCh38
rs781678242 Chr.16: 22008271-22008271 on Build GRCh38
rs554208333 Chr.16: 22008431-22008431 on Build GRCh38
rs958603161 Chr.16: 22008119-22008119 on Build GRCh38
rs777539958 Chr.16: 22008413-22008414 on Build GRCh38
rs867766356 Chr.16: 22008111-22008111 on Build GRCh38
rs924069976 Chr.16: 22008152-22008152 on Build GRCh38
rs1017810544 Chr.16: 22007963-22007963 on Build GRCh38
rs774026195 Chr.16: 22008361-22008361 on Build GRCh38
rs751878267 Chr.16: 22008416-22008416 on Build GRCh38
rs1055943810 Chr.16: 22008002-22008002 on Build GRCh38
rs902120512 Chr.16: 22008188-22008188 on Build GRCh38
rs536353754 Chr.16: 22008405-22008405 on Build GRCh38
rs796962265 Chr.16: 22008408-22008409 on Build GRCh38
rs1043896120 Chr.16: 22008027-22008027 on Build GRCh38
rs937063035 Chr.16: 22007996-22007996 on Build GRCh38
rs1029914678 Chr.16: 22008443-22008443 on Build GRCh38
rs1033285687 Chr.16: 22008110-22008110 on Build GRCh38
rs553928957 Chr.16: 22008084-22008084 on Build GRCh38
rs749296477 Chr.16: 22008418-22008419 on Build GRCh38
rs991818706 Chr.16: 22008274-22008274 on Build GRCh38
rs961911169 Chr.16: 22007975-22007975 on Build GRCh38
rs977168962 Chr.16: 22008396-22008396 on Build GRCh38
rs201044196 Chr.16: 22008326-22008326 on Build GRCh38
rs750498838 Chr.16: 22008414-22008414 on Build GRCh38
rs372729240 Chr.16: 22008409-22008409 on Build GRCh38
rs781770795 Chr.16: 22008429-22008429 on Build GRCh38
rs747696648 Chr.16: 22008280-22008280 on Build GRCh38
rs914355010 Chr.16: 22008009-22008009 on Build GRCh38
rs962937273 Chr.16: 22007965-22007965 on Build GRCh38
rs947092065 Chr.16: 22008136-22008136 on Build GRCh38
rs958626940 Chr.16: 22008317-22008317 on Build GRCh38
rs377033405 Chr.16: 22008411-22008411 on Build GRCh38
rs776782299 Chr.16: 22008409-22008410 on Build GRCh38

View Full Product Details