Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 22082575-22083079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf52
Entrez Gene ID
730094
Gene Name
chromosome 16 open reading frame 52
Gene Aliases
-
UniGene
Hs.498890
Gene Chromosome Location
Chr.16: 22008135-22084654 on Build GRCh38
Transcripts
NM_001164579.1

SNP Detail

SNP ID Chromosome Location
rs376936384 Chr.16: 22083047-22083047 on Build GRCh38
rs915660789 Chr.16: 22082707-22082707 on Build GRCh38
rs942881783 Chr.16: 22083015-22083015 on Build GRCh38
rs965612664 Chr.16: 22082748-22082748 on Build GRCh38
rs980564089 Chr.16: 22082848-22082848 on Build GRCh38
rs997882990 Chr.16: 22082752-22082752 on Build GRCh38
rs913200675 Chr.16: 22082913-22082913 on Build GRCh38
rs371745408 Chr.16: 22083033-22083033 on Build GRCh38
rs1039883774 Chr.16: 22083020-22083020 on Build GRCh38
rs147556437 Chr.16: 22082653-22082653 on Build GRCh38
rs909715564 Chr.16: 22082816-22082816 on Build GRCh38
rs1007554387 Chr.16: 22082632-22082632 on Build GRCh38
rs904601386 Chr.16: 22082987-22082987 on Build GRCh38
rs959693078 Chr.16: 22082882-22082882 on Build GRCh38
rs774920789 Chr.16: 22082941-22082941 on Build GRCh38
rs142779262 Chr.16: 22083046-22083046 on Build GRCh38
rs533940218 Chr.16: 22082983-22082983 on Build GRCh38
rs980887627 Chr.16: 22082991-22082991 on Build GRCh38
rs951224154 Chr.16: 22082780-22082780 on Build GRCh38
rs925494746 Chr.16: 22083012-22083012 on Build GRCh38
rs969810425 Chr.16: 22082954-22082954 on Build GRCh38
rs983858905 Chr.16: 22082789-22082789 on Build GRCh38
rs138732856 Chr.16: 22082939-22082939 on Build GRCh38
rs775255800 Chr.16: 22082681-22082681 on Build GRCh38
rs946128660 Chr.16: 22082955-22082955 on Build GRCh38
rs61271595 Chr.16: 22082654-22082654 on Build GRCh38
rs1043105725 Chr.16: 22082970-22082970 on Build GRCh38
rs531841279 Chr.16: 22082806-22082806 on Build GRCh38
rs993461090 Chr.16: 22082876-22082876 on Build GRCh38
rs936963236 Chr.16: 22082610-22082610 on Build GRCh38
rs762747578 Chr.16: 22082798-22082798 on Build GRCh38
rs1018579066 Chr.16: 22082716-22082716 on Build GRCh38
rs865894111 Chr.16: 22082775-22082775 on Build GRCh38
rs142007625 Chr.16: 22082883-22082883 on Build GRCh38
rs766351596 Chr.16: 22082712-22082712 on Build GRCh38
rs949856603 Chr.16: 22082711-22082711 on Build GRCh38
rs189749407 Chr.16: 22082670-22082670 on Build GRCh38
rs150288212 Chr.16: 22082640-22082640 on Build GRCh38
rs568498075 Chr.16: 22082881-22082881 on Build GRCh38
rs1051811685 Chr.16: 22082749-22082749 on Build GRCh38
rs969211016 Chr.16: 22082839-22082839 on Build GRCh38
rs145860074 Chr.16: 22082911-22082911 on Build GRCh38
rs530325641 Chr.16: 22082797-22082797 on Build GRCh38
rs550230217 Chr.16: 22082864-22082864 on Build GRCh38

View Full Product Details