Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23418622-23419126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG7
Entrez Gene ID
91949
Gene Name
component of oligomeric golgi complex 7
Gene Aliases
CDG2E
UniGene
Hs.185807
Gene Chromosome Location
Chr.16: 23388493-23453215 on Build GRCh38
Transcripts
NM_153603.3 XM_017023870.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5620521 c.1088G>T p.G363V Chr.16: 23418749-23418749 on Build GRCh38
6393694 c.1123A>G p.S375G Chr.16: 23418714-23418714 on Build GRCh38
5775750 c.1060C>T p.P354S Chr.16: 23418777-23418777 on Build GRCh38
4986094 c.1019A>G p.N340S Chr.16: 23418818-23418818 on Build GRCh38
968766 c.1089C>T p.G363G Chr.16: 23418748-23418748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs114661874 Chr.16: 23418724-23418724 on Build GRCh38
rs1027581035 Chr.16: 23418834-23418834 on Build GRCh38
rs1034933858 Chr.16: 23418949-23418950 on Build GRCh38
rs1013321800 Chr.16: 23419032-23419032 on Build GRCh38
rs969050507 Chr.16: 23418933-23418933 on Build GRCh38
rs897925121 Chr.16: 23419003-23419003 on Build GRCh38
rs898119862 Chr.16: 23418862-23418862 on Build GRCh38
rs200768125 Chr.16: 23418794-23418794 on Build GRCh38
rs878853109 Chr.16: 23418702-23418702 on Build GRCh38
rs761298707 Chr.16: 23418764-23418764 on Build GRCh38
rs77369028 Chr.16: 23419013-23419013 on Build GRCh38
rs762609553 Chr.16: 23418902-23418902 on Build GRCh38
rs575790381 Chr.16: 23418946-23418946 on Build GRCh38
rs200620098 Chr.16: 23418708-23418708 on Build GRCh38
rs201747640 Chr.16: 23418803-23418803 on Build GRCh38
rs745893566 Chr.16: 23418854-23418854 on Build GRCh38
rs185250898 Chr.16: 23418876-23418876 on Build GRCh38
rs375684381 Chr.16: 23418852-23418852 on Build GRCh38
rs1036464583 Chr.16: 23418992-23418992 on Build GRCh38
rs994816379 Chr.16: 23418818-23418818 on Build GRCh38
rs889092441 Chr.16: 23418790-23418790 on Build GRCh38
rs930692316 Chr.16: 23419006-23419006 on Build GRCh38
rs750074145 Chr.16: 23418677-23418677 on Build GRCh38
rs897895682 Chr.16: 23418804-23418804 on Build GRCh38
rs987960521 Chr.16: 23418991-23418991 on Build GRCh38
rs746762111 Chr.16: 23418691-23418691 on Build GRCh38
rs758072796 Chr.16: 23418782-23418782 on Build GRCh38
rs767516970 Chr.16: 23419083-23419083 on Build GRCh38
rs776014966 Chr.16: 23418729-23418729 on Build GRCh38
rs910629189 Chr.16: 23419072-23419072 on Build GRCh38
rs756756975 Chr.16: 23418654-23418654 on Build GRCh38
rs1024636972 Chr.16: 23418858-23418858 on Build GRCh38
rs778477301 Chr.16: 23418669-23418669 on Build GRCh38
rs765976652 Chr.16: 23418784-23418784 on Build GRCh38
rs977204872 Chr.16: 23418666-23418666 on Build GRCh38
rs568236280 Chr.16: 23418653-23418653 on Build GRCh38
rs761056859 Chr.16: 23418746-23418746 on Build GRCh38
rs763659623 Chr.16: 23419029-23419029 on Build GRCh38
rs781052051 Chr.16: 23418837-23418837 on Build GRCh38
rs751396156 Chr.16: 23418802-23418802 on Build GRCh38
rs912116576 Chr.16: 23418943-23418943 on Build GRCh38
rs376934637 Chr.16: 23418853-23418853 on Build GRCh38
rs764545607 Chr.16: 23418747-23418747 on Build GRCh38
rs535729641 Chr.16: 23418887-23418887 on Build GRCh38
rs746091664 Chr.16: 23418718-23418718 on Build GRCh38
rs74012174 Chr.16: 23418689-23418689 on Build GRCh38
rs750544067 Chr.16: 23419086-23419086 on Build GRCh38
rs1031445386 Chr.16: 23418896-23418896 on Build GRCh38
rs1036433681 Chr.16: 23418791-23418791 on Build GRCh38
rs754780917 Chr.16: 23418807-23418807 on Build GRCh38
rs1024250717 Chr.16: 23419069-23419069 on Build GRCh38
rs749378045 Chr.16: 23418709-23418709 on Build GRCh38
rs957081254 Chr.16: 23418898-23418898 on Build GRCh38
rs956643208 Chr.16: 23418975-23418975 on Build GRCh38
rs757481553 Chr.16: 23418942-23418942 on Build GRCh38
rs75523257 Chr.16: 23418692-23418692 on Build GRCh38
rs915103392 Chr.16: 23419008-23419008 on Build GRCh38
rs748110576 Chr.16: 23418846-23418846 on Build GRCh38
rs946554801 Chr.16: 23419009-23419009 on Build GRCh38
rs771031254 Chr.16: 23418711-23418711 on Build GRCh38
rs150840793 Chr.16: 23418955-23418955 on Build GRCh38
rs1050959808 Chr.16: 23418771-23418771 on Build GRCh38
rs371201483 Chr.16: 23418878-23418878 on Build GRCh38
rs920901515 Chr.16: 23418906-23418906 on Build GRCh38
rs764735319 Chr.16: 23418765-23418765 on Build GRCh38
rs116838850 Chr.16: 23418911-23418911 on Build GRCh38
rs749881280 Chr.16: 23418770-23418770 on Build GRCh38
rs558338172 Chr.16: 23418962-23418962 on Build GRCh38
rs751766541 Chr.16: 23418905-23418905 on Build GRCh38
rs944861338 Chr.16: 23418783-23418783 on Build GRCh38
rs1012794634 Chr.16: 23418855-23418855 on Build GRCh38
rs907093731 Chr.16: 23419080-23419080 on Build GRCh38
rs539939894 Chr.16: 23418748-23418748 on Build GRCh38

View Full Product Details