Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23524056-23524551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EARS2
Entrez Gene ID
124454
Gene Name
glutamyl-tRNA synthetase 2, mitochondrial
Gene Aliases
COXPD12, MSE1, gluRS
UniGene
Hs.620541
Gene Chromosome Location
Chr.16: 23522013-23557375 on Build GRCh38
Transcripts
NM_001083614.1 NR_003501.1 XM_011545738.1 XM_011545739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5793887 c.1537G>C p.V513L Chr.16: 23524406-23524406 on Build GRCh38
702980 c.1507G>C p.E503Q Chr.16: 23524436-23524436 on Build GRCh38
7365791 c.1546C>T p.R516W Chr.16: 23524397-23524397 on Build GRCh38
702981 c.1541G>A p.R514Q Chr.16: 23524402-23524402 on Build GRCh38
968787 c.1547G>A p.R516Q Chr.16: 23524396-23524396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150304027 Chr.16: 23524343-23524343 on Build GRCh38
rs1014557762 Chr.16: 23524254-23524254 on Build GRCh38
rs748985956 Chr.16: 23524407-23524407 on Build GRCh38
rs935226177 Chr.16: 23524097-23524097 on Build GRCh38
rs781726617 Chr.16: 23524391-23524391 on Build GRCh38
rs201727231 Chr.16: 23524396-23524396 on Build GRCh38
rs767798450 Chr.16: 23524456-23524457 on Build GRCh38
rs547044329 Chr.16: 23524292-23524292 on Build GRCh38
rs199673118 Chr.16: 23524472-23524472 on Build GRCh38
rs570258433 Chr.16: 23524115-23524115 on Build GRCh38
rs778177308 Chr.16: 23524080-23524080 on Build GRCh38
rs138487221 Chr.16: 23524115-23524115 on Build GRCh38
rs769364311 Chr.16: 23524188-23524188 on Build GRCh38
rs372659806 Chr.16: 23524480-23524480 on Build GRCh38
rs769320749 Chr.16: 23524397-23524397 on Build GRCh38
rs950129619 Chr.16: 23524129-23524129 on Build GRCh38
rs774841172 Chr.16: 23524432-23524432 on Build GRCh38
rs566933362 Chr.16: 23524294-23524294 on Build GRCh38
rs745678462 Chr.16: 23524452-23524452 on Build GRCh38
rs886051830 Chr.16: 23524216-23524216 on Build GRCh38
rs761966397 Chr.16: 23524355-23524355 on Build GRCh38
rs893379782 Chr.16: 23524498-23524498 on Build GRCh38
rs569642012 Chr.16: 23524357-23524357 on Build GRCh38
rs1054029019 Chr.16: 23524136-23524136 on Build GRCh38
rs1027601915 Chr.16: 23524354-23524354 on Build GRCh38
rs201144681 Chr.16: 23524501-23524501 on Build GRCh38
rs751942624 Chr.16: 23524388-23524388 on Build GRCh38
rs1029191438 Chr.16: 23524201-23524201 on Build GRCh38
rs1008590057 Chr.16: 23524230-23524230 on Build GRCh38
rs949419846 Chr.16: 23524365-23524365 on Build GRCh38
rs1028683326 Chr.16: 23524170-23524170 on Build GRCh38
rs759447363 Chr.16: 23524342-23524342 on Build GRCh38
rs755506732 Chr.16: 23524389-23524389 on Build GRCh38
rs916684760 Chr.16: 23524361-23524361 on Build GRCh38
rs961968725 Chr.16: 23524235-23524236 on Build GRCh38
rs997021215 Chr.16: 23524192-23524192 on Build GRCh38
rs141499212 Chr.16: 23524280-23524280 on Build GRCh38
rs765449250 Chr.16: 23524495-23524495 on Build GRCh38
rs916011586 Chr.16: 23524392-23524392 on Build GRCh38
rs527418211 Chr.16: 23524289-23524289 on Build GRCh38
rs995912366 Chr.16: 23524156-23524156 on Build GRCh38
rs558487697 Chr.16: 23524402-23524402 on Build GRCh38
rs893636562 Chr.16: 23524141-23524141 on Build GRCh38
rs376807600 Chr.16: 23524403-23524403 on Build GRCh38
rs935282470 Chr.16: 23524427-23524427 on Build GRCh38
rs770554002 Chr.16: 23524422-23524422 on Build GRCh38
rs774079110 Chr.16: 23524433-23524433 on Build GRCh38
rs546116107 Chr.16: 23524195-23524195 on Build GRCh38
rs755731108 Chr.16: 23524467-23524475 on Build GRCh38
rs755445330 Chr.16: 23524334-23524335 on Build GRCh38
rs1045696625 Chr.16: 23524135-23524135 on Build GRCh38
rs1018598540 Chr.16: 23524231-23524231 on Build GRCh38
rs780381864 Chr.16: 23524191-23524191 on Build GRCh38
rs1008533477 Chr.16: 23524205-23524205 on Build GRCh38
rs1468138 Chr.16: 23524359-23524359 on Build GRCh38
rs982525766 Chr.16: 23524370-23524370 on Build GRCh38
rs369770210 Chr.16: 23524468-23524468 on Build GRCh38
rs752357109 Chr.16: 23524463-23524464 on Build GRCh38
rs200936622 Chr.16: 23524356-23524356 on Build GRCh38

View Full Product Details