Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23534474-23534976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EARS2
Entrez Gene ID
124454
Gene Name
glutamyl-tRNA synthetase 2, mitochondrial
Gene Aliases
COXPD12, MSE1, gluRS
UniGene
Hs.620541
Gene Chromosome Location
Chr.16: 23522013-23557375 on Build GRCh38
Transcripts
NM_001083614.1 NM_001308211.1 NR_003501.1 XM_011545738.1 XM_011545739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5620733 c.911C>T p.P304L Chr.16: 23534935-23534935 on Build GRCh38
5513174 c.958+5G>A p.? Chr.16: 23534883-23534883 on Build GRCh38
5980943 c.913G>A p.D305N Chr.16: 23534933-23534933 on Build GRCh38
3279758 c.912C>T p.P304P Chr.16: 23534934-23534934 on Build GRCh38
3888229 c.917C>T p.S306F Chr.16: 23534929-23534929 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs549370113 Chr.16: 23534788-23534788 on Build GRCh38
rs763029240 Chr.16: 23534920-23534920 on Build GRCh38
rs773325130 Chr.16: 23534915-23534915 on Build GRCh38
rs561931446 Chr.16: 23534526-23534526 on Build GRCh38
rs75091684 Chr.16: 23534657-23534657 on Build GRCh38
rs1027870378 Chr.16: 23534806-23534806 on Build GRCh38
rs753022070 Chr.16: 23534934-23534934 on Build GRCh38
rs770099559 Chr.16: 23534910-23534910 on Build GRCh38
rs933679390 Chr.16: 23534917-23534917 on Build GRCh38
rs112190633 Chr.16: 23534758-23534758 on Build GRCh38
rs1033760693 Chr.16: 23534701-23534701 on Build GRCh38
rs372273855 Chr.16: 23534845-23534845 on Build GRCh38
rs542411959 Chr.16: 23534506-23534506 on Build GRCh38
rs777436163 Chr.16: 23534868-23534868 on Build GRCh38
rs964900333 Chr.16: 23534882-23534882 on Build GRCh38
rs756733389 Chr.16: 23534879-23534879 on Build GRCh38
rs746838793 Chr.16: 23534897-23534897 on Build GRCh38
rs762973093 Chr.16: 23534839-23534839 on Build GRCh38
rs546602781 Chr.16: 23534700-23534700 on Build GRCh38
rs767860727 Chr.16: 23534862-23534862 on Build GRCh38
rs569523339 Chr.16: 23534812-23534812 on Build GRCh38
rs747769437 Chr.16: 23534601-23534601 on Build GRCh38
rs753113708 Chr.16: 23534864-23534864 on Build GRCh38
rs979601199 Chr.16: 23534850-23534850 on Build GRCh38
rs1038144322 Chr.16: 23534681-23534681 on Build GRCh38
rs975609725 Chr.16: 23534884-23534885 on Build GRCh38
rs920964901 Chr.16: 23534609-23534609 on Build GRCh38
rs760017945 Chr.16: 23534932-23534932 on Build GRCh38
rs879567788 Chr.16: 23534816-23534816 on Build GRCh38
rs935973795 Chr.16: 23534608-23534608 on Build GRCh38
rs560084947 Chr.16: 23534721-23534721 on Build GRCh38
rs755736432 Chr.16: 23534867-23534867 on Build GRCh38
rs771761807 Chr.16: 23534636-23534636 on Build GRCh38
rs1013575782 Chr.16: 23534763-23534763 on Build GRCh38
rs917369842 Chr.16: 23534521-23534521 on Build GRCh38
rs1003954803 Chr.16: 23534725-23534725 on Build GRCh38
rs939868175 Chr.16: 23534663-23534663 on Build GRCh38
rs575391276 Chr.16: 23534574-23534574 on Build GRCh38
rs553998430 Chr.16: 23534508-23534508 on Build GRCh38
rs773309672 Chr.16: 23534837-23534837 on Build GRCh38
rs565383416 Chr.16: 23534935-23534935 on Build GRCh38
rs752019143 Chr.16: 23534844-23534844 on Build GRCh38
rs920822371 Chr.16: 23534504-23534504 on Build GRCh38
rs539512674 Chr.16: 23534940-23534940 on Build GRCh38
rs34451686 Chr.16: 23534929-23534930 on Build GRCh38
rs991577629 Chr.16: 23534520-23534520 on Build GRCh38
rs991651067 Chr.16: 23534831-23534831 on Build GRCh38
rs925712060 Chr.16: 23534546-23534546 on Build GRCh38
rs117775618 Chr.16: 23534688-23534688 on Build GRCh38
rs979567058 Chr.16: 23534543-23534543 on Build GRCh38
rs774553761 Chr.16: 23534929-23534929 on Build GRCh38
rs779972250 Chr.16: 23534896-23534896 on Build GRCh38
rs996687495 Chr.16: 23534668-23534668 on Build GRCh38
rs966928471 Chr.16: 23534820-23534820 on Build GRCh38
rs139429186 Chr.16: 23534591-23534591 on Build GRCh38
rs201958550 Chr.16: 23534953-23534953 on Build GRCh38
rs777216534 Chr.16: 23534785-23534785 on Build GRCh38
rs34228465 Chr.16: 23534872-23534873 on Build GRCh38
rs759775898 Chr.16: 23534791-23534791 on Build GRCh38
rs772192769 Chr.16: 23534888-23534888 on Build GRCh38
rs543979506 Chr.16: 23534884-23534884 on Build GRCh38
rs374610062 Chr.16: 23534876-23534876 on Build GRCh38
rs368574794 Chr.16: 23534933-23534933 on Build GRCh38
rs933670599 Chr.16: 23534615-23534615 on Build GRCh38
rs538101009 Chr.16: 23534843-23534843 on Build GRCh38
rs200441317 Chr.16: 23534883-23534883 on Build GRCh38
rs770916034 Chr.16: 23534926-23534926 on Build GRCh38
rs1054390244 Chr.16: 23534648-23534648 on Build GRCh38
rs950129905 Chr.16: 23534522-23534522 on Build GRCh38

View Full Product Details